Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 64951520-64951783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHURC1-FNTB
Entrez Gene ID
100529261
Gene Name
CHURC1-FNTB readthrough
Gene Aliases
-
UniGene
Hs.325531
Gene Chromosome Location
Chr.14: 64914361-65062655 on Build GRCh38
Transcripts
NM_001202558.1 NM_001202559.1 NM_001308154.1 NM_198686.2 XM_017021277.1

Gene Symbol
RAB15
Entrez Gene ID
376267
Gene Name
RAB15, member RAS oncogene family
Gene Aliases
-
UniGene
Hs.512492
Gene Chromosome Location
Chr.14: 64945814-64972197 on Build GRCh38
Transcripts
NM_001202558.1 NM_001202559.1 NM_001308154.1 NM_198686.2 XM_017021277.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5682765 c.245A>C p.Q82P Chr.14: 64951604-64951604 on Build GRCh38
957087 c.206_207delGA p.R69fs*20 Chr.14: 64951642-64951643 on Build GRCh38
4051897 c.235C>T p.R79W Chr.14: 64951614-64951614 on Build GRCh38
7348627 c.238C>T p.R80W Chr.14: 64951611-64951611 on Build GRCh38
4993751 c.186-3C>T p.? Chr.14: 64951666-64951666 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202121487 Chr.14: 64951573-64951573 on Build GRCh38
rs192700550 Chr.14: 64951713-64951713 on Build GRCh38
rs878968224 Chr.14: 64951582-64951582 on Build GRCh38
rs918193754 Chr.14: 64951607-64951607 on Build GRCh38
rs201092247 Chr.14: 64951549-64951549 on Build GRCh38
rs376148325 Chr.14: 64951557-64951557 on Build GRCh38
rs374668849 Chr.14: 64951700-64951700 on Build GRCh38
rs1007476459 Chr.14: 64951634-64951634 on Build GRCh38
rs774560080 Chr.14: 64951690-64951690 on Build GRCh38
rs375263502 Chr.14: 64951681-64951681 on Build GRCh38
rs17752452 Chr.14: 64951757-64951757 on Build GRCh38
rs767354797 Chr.14: 64951563-64951563 on Build GRCh38
rs917374625 Chr.14: 64951548-64951548 on Build GRCh38
rs369763424 Chr.14: 64951686-64951686 on Build GRCh38
rs781495059 Chr.14: 64951670-64951670 on Build GRCh38
rs767857187 Chr.14: 64951571-64951571 on Build GRCh38
rs748552099 Chr.14: 64951677-64951677 on Build GRCh38
rs759561426 Chr.14: 64951562-64951562 on Build GRCh38
rs756782637 Chr.14: 64951629-64951629 on Build GRCh38
rs753445318 Chr.14: 64951613-64951613 on Build GRCh38
rs766518800 Chr.14: 64951555-64951555 on Build GRCh38
rs7157667 Chr.14: 64951547-64951547 on Build GRCh38
rs759523101 Chr.14: 64951691-64951691 on Build GRCh38
rs778442280 Chr.14: 64951647-64951647 on Build GRCh38
rs777403701 Chr.14: 64951612-64951612 on Build GRCh38
rs547074619 Chr.14: 64951669-64951669 on Build GRCh38
rs372423854 Chr.14: 64951587-64951587 on Build GRCh38
rs1040106599 Chr.14: 64951755-64951755 on Build GRCh38
rs752712009 Chr.14: 64951588-64951588 on Build GRCh38
rs773590414 Chr.14: 64951684-64951684 on Build GRCh38
rs145781570 Chr.14: 64951714-64951714 on Build GRCh38
rs755804668 Chr.14: 64951600-64951600 on Build GRCh38
rs769270248 Chr.14: 64951666-64951666 on Build GRCh38
rs745337121 Chr.14: 64951651-64951651 on Build GRCh38
rs956203702 Chr.14: 64951758-64951758 on Build GRCh38

View Full Product Details