Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 925351-925624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD52
Entrez Gene ID
5893
Gene Name
RAD52 homolog, DNA repair protein
Gene Aliases
-
UniGene
Hs.410355
Gene Chromosome Location
Chr.12: 911028-991195 on Build GRCh38
Transcripts
NM_001297419.1 NM_001297420.1 NM_001297421.1 NM_001297422.1 NM_134424.3 NR_123713.1 XM_005253720.4 XM_005253721.2 XM_011520990.2 XM_011520991.2 XM_011520993.2 XM_011520995.2 XM_017019769.1 XM_017019770.1 XM_017019771.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7227498 c.619-1G>A p.? Chr.12: 925526-925526 on Build GRCh38
6952298 c.538C>T p.R180C Chr.12: 925455-925455 on Build GRCh38
415663 c.475G>T p.G159W Chr.12: 925518-925518 on Build GRCh38
7087223 c.626G>T p.W209L Chr.12: 925518-925518 on Build GRCh38
7415635 c.633C>T p.C211C Chr.12: 925511-925511 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs912061045 Chr.12: 925471-925471 on Build GRCh38
rs370821139 Chr.12: 925413-925413 on Build GRCh38
rs781318903 Chr.12: 925446-925446 on Build GRCh38
rs774802853 Chr.12: 925460-925460 on Build GRCh38
rs774712903 Chr.12: 925401-925401 on Build GRCh38
rs879705768 Chr.12: 925397-925398 on Build GRCh38
rs759881543 Chr.12: 925402-925402 on Build GRCh38
rs577090674 Chr.12: 925455-925455 on Build GRCh38
rs771344880 Chr.12: 925400-925400 on Build GRCh38
rs772724840 Chr.12: 925405-925406 on Build GRCh38
rs766193263 Chr.12: 925541-925541 on Build GRCh38
rs1056037095 Chr.12: 925393-925393 on Build GRCh38
rs574333345 Chr.12: 925513-925513 on Build GRCh38
rs951810349 Chr.12: 925456-925456 on Build GRCh38
rs558702311 Chr.12: 925438-925438 on Build GRCh38
rs757281589 Chr.12: 925554-925554 on Build GRCh38
rs748370290 Chr.12: 925447-925447 on Build GRCh38
rs879912288 Chr.12: 925396-925396 on Build GRCh38
rs750351119 Chr.12: 925522-925522 on Build GRCh38
rs200127435 Chr.12: 925485-925485 on Build GRCh38
rs779195700 Chr.12: 925490-925490 on Build GRCh38
rs774739534 Chr.12: 925444-925445 on Build GRCh38
rs779340661 Chr.12: 925557-925557 on Build GRCh38
rs757271884 Chr.12: 925425-925425 on Build GRCh38
rs771434917 Chr.12: 925458-925458 on Build GRCh38
rs763779337 Chr.12: 925493-925493 on Build GRCh38
rs928526645 Chr.12: 925550-925550 on Build GRCh38
rs138828874 Chr.12: 925491-925491 on Build GRCh38
rs372461582 Chr.12: 925454-925454 on Build GRCh38
rs763689571 Chr.12: 925410-925410 on Build GRCh38
rs765196587 Chr.12: 925431-925431 on Build GRCh38
rs756253872 Chr.12: 925451-925451 on Build GRCh38
rs375873399 Chr.12: 925403-925403 on Build GRCh38
rs777906261 Chr.12: 925548-925548 on Build GRCh38
rs770515460 Chr.12: 925474-925474 on Build GRCh38
rs774157236 Chr.12: 925554-925555 on Build GRCh38
rs756343839 Chr.12: 925546-925546 on Build GRCh38
rs750281609 Chr.12: 925572-925572 on Build GRCh38
rs746359027 Chr.12: 925563-925563 on Build GRCh38
rs552708755 Chr.12: 925510-925510 on Build GRCh38
rs985877199 Chr.12: 925465-925465 on Build GRCh38
rs113335624 Chr.12: 925545-925545 on Build GRCh38
rs753952482 Chr.12: 925549-925549 on Build GRCh38
rs984891328 Chr.12: 925472-925472 on Build GRCh38
rs563418439 Chr.12: 925584-925584 on Build GRCh38
rs999857714 Chr.12: 925430-925430 on Build GRCh38
rs377004121 Chr.12: 925530-925530 on Build GRCh38
rs758161957 Chr.12: 925445-925445 on Build GRCh38
rs746245343 Chr.12: 925462-925462 on Build GRCh38

View Full Product Details