Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 24061117-24061368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYOM3
Entrez Gene ID
127294
Gene Name
myomesin 3
Gene Aliases
-
UniGene
Hs.523413
Gene Chromosome Location
Chr.1: 24056041-24112175 on Build GRCh38
Transcripts
NM_152372.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4705199 c.626C>A p.A209E Chr.1: 24061297-24061297 on Build GRCh38
7378704 c.640C>T p.Q214* Chr.1: 24061283-24061283 on Build GRCh38
1296094 c.3940G>A p.E1314K Chr.1: 24061304-24061304 on Build GRCh38
7222301 c.3935-7C>A p.? Chr.1: 24061316-24061316 on Build GRCh38
4810932 c.619G>A p.E207K Chr.1: 24061304-24061304 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992248342 Chr.1: 24061232-24061232 on Build GRCh38
rs1021660515 Chr.1: 24061152-24061152 on Build GRCh38
rs7553075 Chr.1: 24061332-24061332 on Build GRCh38
rs781467557 Chr.1: 24061229-24061229 on Build GRCh38
rs765173786 Chr.1: 24061312-24061312 on Build GRCh38
rs1011783705 Chr.1: 24061145-24061145 on Build GRCh38
rs768368580 Chr.1: 24061225-24061225 on Build GRCh38
rs770233156 Chr.1: 24061242-24061242 on Build GRCh38
rs773764958 Chr.1: 24061151-24061151 on Build GRCh38
rs568664028 Chr.1: 24061327-24061327 on Build GRCh38
rs905556765 Chr.1: 24061182-24061182 on Build GRCh38
rs967888030 Chr.1: 24061163-24061163 on Build GRCh38
rs201826629 Chr.1: 24061284-24061284 on Build GRCh38
rs773683841 Chr.1: 24061245-24061245 on Build GRCh38
rs373323109 Chr.1: 24061321-24061321 on Build GRCh38
rs576902527 Chr.1: 24061186-24061186 on Build GRCh38
rs746627664 Chr.1: 24061219-24061219 on Build GRCh38
rs201550622 Chr.1: 24061348-24061348 on Build GRCh38
rs780844720 Chr.1: 24061342-24061342 on Build GRCh38
rs759621205 Chr.1: 24061273-24061273 on Build GRCh38
rs779655722 Chr.1: 24061213-24061213 on Build GRCh38
rs774551825 Chr.1: 24061266-24061266 on Build GRCh38
rs750356636 Chr.1: 24061313-24061313 on Build GRCh38
rs757824389 Chr.1: 24061207-24061207 on Build GRCh38
rs754758387 Chr.1: 24061314-24061316 on Build GRCh38
rs749904998 Chr.1: 24061201-24061201 on Build GRCh38
rs918896689 Chr.1: 24061292-24061292 on Build GRCh38
rs753650248 Chr.1: 24061279-24061279 on Build GRCh38
rs767861269 Chr.1: 24061275-24061275 on Build GRCh38
rs770907850 Chr.1: 24061258-24061258 on Build GRCh38
rs763551551 Chr.1: 24061256-24061256 on Build GRCh38
rs748542320 Chr.1: 24061239-24061239 on Build GRCh38
rs549160937 Chr.1: 24061333-24061333 on Build GRCh38

View Full Product Details