Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 92593203-92593440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF11
Entrez Gene ID
3832
Gene Name
kinesin family member 11
Gene Aliases
EG5, HKSP, KNSL1, MCLMR, TRIP5
UniGene
Hs.8878
Gene Chromosome Location
Chr.10: 92593068-92655395 on Build GRCh38
Transcripts
NM_004523.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5647534 c.30G>A p.K10K Chr.10: 92593405-92593405 on Build GRCh38
1969638 c.8C>T p.S3L Chr.10: 92593383-92593383 on Build GRCh38
288170 c.42G>A p.E14E Chr.10: 92593417-92593417 on Build GRCh38
1349820 c.5C>T p.A2V Chr.10: 92593380-92593380 on Build GRCh38
1177925 c.27G>T p.A9A Chr.10: 92593402-92593402 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs376818610 Chr.10: 92593387-92593387 on Build GRCh38
rs779024446 Chr.10: 92593356-92593357 on Build GRCh38
rs556891991 Chr.10: 92593352-92593353 on Build GRCh38
rs534736976 Chr.10: 92593348-92593348 on Build GRCh38
rs746095298 Chr.10: 92593363-92593363 on Build GRCh38
rs768491534 Chr.10: 92593374-92593374 on Build GRCh38
rs1035422524 Chr.10: 92593405-92593405 on Build GRCh38
rs552381017 Chr.10: 92593240-92593240 on Build GRCh38
rs1037026288 Chr.10: 92593276-92593276 on Build GRCh38
rs942399029 Chr.10: 92593231-92593231 on Build GRCh38
rs909397916 Chr.10: 92593232-92593232 on Build GRCh38
rs756475965 Chr.10: 92593342-92593342 on Build GRCh38
rs797045649 Chr.10: 92593375-92593377 on Build GRCh38
rs1006674521 Chr.10: 92593332-92593332 on Build GRCh38
rs1047277725 Chr.10: 92593321-92593321 on Build GRCh38
rs185175870 Chr.10: 92593299-92593299 on Build GRCh38
rs370660638 Chr.10: 92593347-92593347 on Build GRCh38
rs375202133 Chr.10: 92593373-92593373 on Build GRCh38
rs36015451 Chr.10: 92593362-92593362 on Build GRCh38
rs993827553 Chr.10: 92593296-92593296 on Build GRCh38
rs1050789609 Chr.10: 92593305-92593305 on Build GRCh38
rs890862092 Chr.10: 92593315-92593315 on Build GRCh38
rs772146436 Chr.10: 92593367-92593367 on Build GRCh38
rs752951562 Chr.10: 92593331-92593331 on Build GRCh38
rs764844734 Chr.10: 92593401-92593401 on Build GRCh38
rs778910273 Chr.10: 92593358-92593358 on Build GRCh38
rs776766098 Chr.10: 92593384-92593384 on Build GRCh38
rs758052909 Chr.10: 92593327-92593327 on Build GRCh38
rs888621985 Chr.10: 92593323-92593323 on Build GRCh38
rs896786457 Chr.10: 92593282-92593282 on Build GRCh38
rs565712763 Chr.10: 92593313-92593313 on Build GRCh38
rs757389411 Chr.10: 92593349-92593349 on Build GRCh38
rs967300114 Chr.10: 92593404-92593404 on Build GRCh38
rs775378350 Chr.10: 92593372-92593372 on Build GRCh38
rs901466346 Chr.10: 92593271-92593271 on Build GRCh38
rs886041477 Chr.10: 92593399-92593401 on Build GRCh38
rs759607487 Chr.10: 92593364-92593364 on Build GRCh38

View Full Product Details