Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 3074701-3074962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL5RA
Entrez Gene ID
3568
Gene Name
interleukin 5 receptor subunit alpha
Gene Aliases
CD125, CDw125, HSIL5R3, IL5R
UniGene
Hs.68876
Gene Chromosome Location
Chr.3: 3066324-3110414 on Build GRCh38
Transcripts
NM_000564.4 NM_001243099.1 NM_175726.3 XM_011533677.2 XM_011533678.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1043561 c.1104G>A p.W368* Chr.3: 3074854-3074854 on Build GRCh38
5399046 c.1150G>A p.D384N Chr.3: 3074808-3074808 on Build GRCh38
5350966 c.1111T>C p.L371L Chr.3: 3074847-3074847 on Build GRCh38
5653241 c.1143T>C p.N381N Chr.3: 3074815-3074815 on Build GRCh38
6055242 c.1172A>G p.Y391C Chr.3: 3074786-3074786 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745723647 Chr.3: 3074838-3074838 on Build GRCh38
rs371978826 Chr.3: 3074889-3074889 on Build GRCh38
rs766942347 Chr.3: 3074815-3074815 on Build GRCh38
rs904451337 Chr.3: 3074841-3074841 on Build GRCh38
rs757148431 Chr.3: 3074874-3074874 on Build GRCh38
rs766820017 Chr.3: 3074900-3074900 on Build GRCh38
rs770063193 Chr.3: 3074894-3074894 on Build GRCh38
rs368091203 Chr.3: 3074885-3074885 on Build GRCh38
rs773565076 Chr.3: 3074896-3074896 on Build GRCh38
rs1008977759 Chr.3: 3074899-3074899 on Build GRCh38
rs755720855 Chr.3: 3074738-3074738 on Build GRCh38
rs144801365 Chr.3: 3074730-3074730 on Build GRCh38
rs374665307 Chr.3: 3074875-3074875 on Build GRCh38
rs752119611 Chr.3: 3074816-3074816 on Build GRCh38
rs527408144 Chr.3: 3074881-3074883 on Build GRCh38
rs764469899 Chr.3: 3074757-3074757 on Build GRCh38
rs1018738827 Chr.3: 3074875-3074876 on Build GRCh38
rs149306264 Chr.3: 3074834-3074834 on Build GRCh38
rs772102514 Chr.3: 3074856-3074856 on Build GRCh38
rs776936532 Chr.3: 3074777-3074777 on Build GRCh38
rs993936122 Chr.3: 3074925-3074925 on Build GRCh38
rs780191401 Chr.3: 3074859-3074859 on Build GRCh38
rs768741095 Chr.3: 3074773-3074773 on Build GRCh38
rs753346086 Chr.3: 3074911-3074911 on Build GRCh38
rs747012271 Chr.3: 3074864-3074864 on Build GRCh38
rs763670010 Chr.3: 3074907-3074907 on Build GRCh38
rs765590204 Chr.3: 3074792-3074792 on Build GRCh38
rs754374066 Chr.3: 3074887-3074887 on Build GRCh38
rs760847323 Chr.3: 3074769-3074769 on Build GRCh38
rs778746390 Chr.3: 3074915-3074915 on Build GRCh38
rs995110846 Chr.3: 3074770-3074770 on Build GRCh38
rs938214400 Chr.3: 3074729-3074729 on Build GRCh38
rs753528516 Chr.3: 3074831-3074831 on Build GRCh38
rs776650983 Chr.3: 3074888-3074888 on Build GRCh38
rs778837894 Chr.3: 3074837-3074837 on Build GRCh38
rs760033832 Chr.3: 3074902-3074902 on Build GRCh38
rs777528348 Chr.3: 3074739-3074739 on Build GRCh38
rs141849082 Chr.3: 3074814-3074814 on Build GRCh38
rs935795300 Chr.3: 3074857-3074857 on Build GRCh38
rs756965599 Chr.3: 3074914-3074914 on Build GRCh38
rs147942670 Chr.3: 3074753-3074753 on Build GRCh38
rs913867286 Chr.3: 3074921-3074921 on Build GRCh38
rs755669379 Chr.3: 3074822-3074822 on Build GRCh38
rs2290611 Chr.3: 3074754-3074754 on Build GRCh38
rs747754751 Chr.3: 3074737-3074737 on Build GRCh38
rs775121663 Chr.3: 3074917-3074917 on Build GRCh38
rs777248198 Chr.3: 3074823-3074823 on Build GRCh38
rs371452304 Chr.3: 3074762-3074762 on Build GRCh38
rs780697711 Chr.3: 3074733-3074733 on Build GRCh38
rs754423045 Chr.3: 3074731-3074731 on Build GRCh38
rs772192826 Chr.3: 3074756-3074756 on Build GRCh38
rs548174501 Chr.3: 3074788-3074788 on Build GRCh38
rs762044640 Chr.3: 3074782-3074782 on Build GRCh38
rs752330313 Chr.3: 3074742-3074742 on Build GRCh38
rs768812463 Chr.3: 3074884-3074884 on Build GRCh38
rs759867054 Chr.3: 3074883-3074883 on Build GRCh38

View Full Product Details