Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 39510270-39510543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UGDH
Entrez Gene ID
7358
Gene Name
UDP-glucose 6-dehydrogenase
Gene Aliases
GDH, UDP-GlcDH, UDPGDH, UGD
UniGene
Hs.572518
Gene Chromosome Location
Chr.4: 39498755-39527598 on Build GRCh38
Transcripts
NM_001184700.1 NM_001184701.1 NM_003359.3 XM_005262667.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2846751 c.607G>T p.V203F Chr.4: 39510409-39510409 on Build GRCh38
6890431 c.567G>A p.Q189Q Chr.4: 39510449-39510449 on Build GRCh38
6343806 c.535C>A p.L179M Chr.4: 39510481-39510481 on Build GRCh38
5866381 c.502G>A p.A168T Chr.4: 39510514-39510514 on Build GRCh38
6535313 c.567G>C p.Q189H Chr.4: 39510449-39510449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751260541 Chr.4: 39510447-39510447 on Build GRCh38
rs748387130 Chr.4: 39510520-39510520 on Build GRCh38
rs551600129 Chr.4: 39510317-39510317 on Build GRCh38
rs763685293 Chr.4: 39510445-39510445 on Build GRCh38
rs780212743 Chr.4: 39510314-39510314 on Build GRCh38
rs768014546 Chr.4: 39510477-39510477 on Build GRCh38
rs778486297 Chr.4: 39510399-39510399 on Build GRCh38
rs144192248 Chr.4: 39510435-39510435 on Build GRCh38
rs748362439 Chr.4: 39510371-39510371 on Build GRCh38
rs754352113 Chr.4: 39510503-39510503 on Build GRCh38
rs775468206 Chr.4: 39510428-39510428 on Build GRCh38
rs200180865 Chr.4: 39510407-39510407 on Build GRCh38
rs780471919 Chr.4: 39510500-39510500 on Build GRCh38
rs756467468 Chr.4: 39510493-39510493 on Build GRCh38
rs768152502 Chr.4: 39510342-39510342 on Build GRCh38
rs948466547 Chr.4: 39510462-39510462 on Build GRCh38
rs933655472 Chr.4: 39510402-39510402 on Build GRCh38
rs776235593 Chr.4: 39510394-39510394 on Build GRCh38
rs755113560 Chr.4: 39510516-39510516 on Build GRCh38
rs770721281 Chr.4: 39510384-39510384 on Build GRCh38
rs3186529 Chr.4: 39510518-39510518 on Build GRCh38
rs774610502 Chr.4: 39510369-39510369 on Build GRCh38
rs908289507 Chr.4: 39510507-39510507 on Build GRCh38
rs750711785 Chr.4: 39510485-39510485 on Build GRCh38
rs765048353 Chr.4: 39510413-39510413 on Build GRCh38
rs565350545 Chr.4: 39510341-39510341 on Build GRCh38
rs1054657510 Chr.4: 39510409-39510409 on Build GRCh38
rs768697505 Chr.4: 39510343-39510343 on Build GRCh38
rs894232451 Chr.4: 39510439-39510439 on Build GRCh38
rs527756813 Chr.4: 39510472-39510472 on Build GRCh38
rs935837317 Chr.4: 39510310-39510310 on Build GRCh38
rs745479173 Chr.4: 39510485-39510488 on Build GRCh38

View Full Product Details