Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 115664037-115664296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VANGL1
Entrez Gene ID
81839
Gene Name
VANGL planar cell polarity protein 1
Gene Aliases
KITENIN, LPP2, STB2, STBM2
UniGene
Hs.515130
Gene Chromosome Location
Chr.1: 115641953-115698224 on Build GRCh38
Transcripts
NM_001172411.1 NM_001172412.1 NM_138959.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
894346 c.752C>T p.T251M Chr.1: 115664208-115664208 on Build GRCh38
6243690 c.636G>T p.R212R Chr.1: 115664092-115664092 on Build GRCh38
397610 c.701A>G p.H234R Chr.1: 115664157-115664157 on Build GRCh38
894345 c.725A>C p.E242A Chr.1: 115664181-115664181 on Build GRCh38
40742 c.634C>T p.R212W Chr.1: 115664090-115664090 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776892785 Chr.1: 115664076-115664076 on Build GRCh38
rs767661635 Chr.1: 115664132-115664132 on Build GRCh38
rs752268406 Chr.1: 115664223-115664223 on Build GRCh38
rs753592449 Chr.1: 115664125-115664125 on Build GRCh38
rs779577966 Chr.1: 115664119-115664119 on Build GRCh38
rs143956994 Chr.1: 115664141-115664141 on Build GRCh38
rs775707643 Chr.1: 115664168-115664168 on Build GRCh38
rs770264647 Chr.1: 115664161-115664161 on Build GRCh38
rs780511076 Chr.1: 115664238-115664238 on Build GRCh38
rs765391868 Chr.1: 115664086-115664086 on Build GRCh38
rs754663532 Chr.1: 115664131-115664131 on Build GRCh38
rs200039235 Chr.1: 115664263-115664263 on Build GRCh38
rs757915785 Chr.1: 115664230-115664230 on Build GRCh38
rs777962862 Chr.1: 115664237-115664237 on Build GRCh38
rs773992696 Chr.1: 115664069-115664069 on Build GRCh38
rs761828824 Chr.1: 115664188-115664188 on Build GRCh38
rs746227126 Chr.1: 115664153-115664153 on Build GRCh38
rs781682884 Chr.1: 115664146-115664146 on Build GRCh38
rs376477305 Chr.1: 115664170-115664170 on Build GRCh38
rs373384296 Chr.1: 115664105-115664105 on Build GRCh38
rs771587724 Chr.1: 115664144-115664144 on Build GRCh38
rs764218954 Chr.1: 115664093-115664093 on Build GRCh38
rs929609479 Chr.1: 115664179-115664179 on Build GRCh38
rs772261356 Chr.1: 115664264-115664264 on Build GRCh38
rs377435467 Chr.1: 115664243-115664243 on Build GRCh38
rs201630629 Chr.1: 115664208-115664208 on Build GRCh38
rs777148076 Chr.1: 115664231-115664231 on Build GRCh38
rs148388816 Chr.1: 115664200-115664200 on Build GRCh38
rs538250549 Chr.1: 115664244-115664244 on Build GRCh38
rs555914402 Chr.1: 115664215-115664215 on Build GRCh38
rs1009946726 Chr.1: 115664091-115664091 on Build GRCh38
rs368722418 Chr.1: 115664221-115664221 on Build GRCh38
rs121918218 Chr.1: 115664171-115664171 on Build GRCh38
rs868017676 Chr.1: 115664276-115664276 on Build GRCh38
rs751619287 Chr.1: 115664095-115664095 on Build GRCh38
rs201593132 Chr.1: 115664082-115664082 on Build GRCh38
rs771534557 Chr.1: 115664075-115664075 on Build GRCh38
rs568821247 Chr.1: 115664090-115664090 on Build GRCh38
rs201698322 Chr.1: 115664137-115664137 on Build GRCh38
rs775717716 Chr.1: 115664087-115664087 on Build GRCh38
rs577862345 Chr.1: 115664068-115664068 on Build GRCh38
rs766252819 Chr.1: 115664209-115664209 on Build GRCh38
rs143395094 Chr.1: 115664096-115664096 on Build GRCh38
rs755880701 Chr.1: 115664107-115664107 on Build GRCh38
rs750307907 Chr.1: 115664202-115664202 on Build GRCh38
rs767398438 Chr.1: 115664097-115664097 on Build GRCh38
rs199932833 Chr.1: 115664222-115664222 on Build GRCh38
rs759896002 Chr.1: 115664078-115664078 on Build GRCh38
rs373473714 Chr.1: 115664236-115664236 on Build GRCh38
rs200762200 Chr.1: 115664176-115664176 on Build GRCh38
rs774495925 Chr.1: 115664250-115664250 on Build GRCh38
rs747576456 Chr.1: 115664074-115664074 on Build GRCh38

View Full Product Details