Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 32206781-32207042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC30A6
Entrez Gene ID
55676
Gene Name
solute carrier family 30 member 6
Gene Aliases
MST103, MSTP103, ZNT6
UniGene
Hs.23248
Gene Chromosome Location
Chr.2: 32165841-32224806 on Build GRCh38
Transcripts
NM_001193513.1 NM_001193514.1 NM_001193515.1 NM_017964.3 XM_006712048.3 XM_011532959.2 XM_011532960.2 XM_011532961.2 XM_011532962.2 XM_011532963.2 XM_017004462.1 XM_017004463.1 XM_017004465.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4836766 c.814G>C p.E272Q Chr.2: 32206931-32206931 on Build GRCh38
5911726 c.769-5C>T p.? Chr.2: 32206881-32206881 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372714720 Chr.2: 32206964-32206964 on Build GRCh38
rs201122071 Chr.2: 32206888-32206888 on Build GRCh38
rs1004087671 Chr.2: 32207004-32207004 on Build GRCh38
rs142989123 Chr.2: 32206832-32206832 on Build GRCh38
rs375583122 Chr.2: 32206843-32206843 on Build GRCh38
rs143049848 Chr.2: 32206897-32206897 on Build GRCh38
rs1001990525 Chr.2: 32206973-32206973 on Build GRCh38
rs746420319 Chr.2: 32206844-32206844 on Build GRCh38
rs993634003 Chr.2: 32206884-32206884 on Build GRCh38
rs367958197 Chr.2: 32206831-32206831 on Build GRCh38
rs913818351 Chr.2: 32206819-32206819 on Build GRCh38
rs1032088748 Chr.2: 32206975-32206975 on Build GRCh38
rs780420872 Chr.2: 32206942-32206942 on Build GRCh38
rs775691082 Chr.2: 32206849-32206849 on Build GRCh38
rs369635825 Chr.2: 32206830-32206830 on Build GRCh38
rs968367646 Chr.2: 32206826-32206826 on Build GRCh38
rs915708921 Chr.2: 32206889-32206889 on Build GRCh38
rs749716480 Chr.2: 32206840-32206840 on Build GRCh38
rs776999523 Chr.2: 32206883-32206883 on Build GRCh38
rs888402499 Chr.2: 32206959-32206963 on Build GRCh38
rs779294861 Chr.2: 32206928-32206928 on Build GRCh38
rs754478167 Chr.2: 32206913-32206913 on Build GRCh38
rs772694486 Chr.2: 32206938-32206938 on Build GRCh38
rs374425016 Chr.2: 32206803-32206803 on Build GRCh38
rs569795957 Chr.2: 32206895-32206895 on Build GRCh38
rs141245207 Chr.2: 32206918-32206918 on Build GRCh38
rs988092896 Chr.2: 32206806-32206806 on Build GRCh38
rs751134791 Chr.2: 32206905-32206905 on Build GRCh38
rs772432006 Chr.2: 32206847-32206847 on Build GRCh38
rs762986625 Chr.2: 32206898-32206898 on Build GRCh38
rs770109658 Chr.2: 32206971-32206971 on Build GRCh38
rs771285763 Chr.2: 32206842-32206842 on Build GRCh38
rs746200447 Chr.2: 32206937-32206937 on Build GRCh38
rs370420771 Chr.2: 32206961-32206961 on Build GRCh38
rs766200741 Chr.2: 32206903-32206903 on Build GRCh38
rs773108052 Chr.2: 32206980-32206980 on Build GRCh38
rs769066799 Chr.2: 32206859-32206859 on Build GRCh38
rs372538540 Chr.2: 32206851-32206851 on Build GRCh38
rs762639024 Chr.2: 32206983-32206983 on Build GRCh38
rs200170269 Chr.2: 32206925-32206925 on Build GRCh38
rs977126368 Chr.2: 32206828-32206828 on Build GRCh38
rs752372002 Chr.2: 32206924-32206924 on Build GRCh38
rs747485262 Chr.2: 32206954-32206954 on Build GRCh38

View Full Product Details