Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 102373335-102373582 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLCO6A1
Entrez Gene ID
133482
Gene Name
solute carrier organic anion transporter family member 6A1
Gene Aliases
CT48, GST, OATP-I, OATP6A1, OATPY
UniGene
Hs.388874
Gene Chromosome Location
Chr.5: 102371782-102502213 on Build GRCh38
Transcripts
NM_001289002.1 NM_001289004.1 NM_001308014.1 NM_173488.4 XM_005271874.3 XM_011543147.2 XM_011543148.2 XM_011543150.1 XM_011543153.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3946701 c.2080C>A p.R694S Chr.5: 102373432-102373432 on Build GRCh38
3409645 c.2142A>G p.K714K Chr.5: 102373370-102373370 on Build GRCh38
4477139 c.2125C>T p.P709S Chr.5: 102373387-102373387 on Build GRCh38
5866988 c.2146G>A p.E716K Chr.5: 102373366-102373366 on Build GRCh38
4476891 c.2104C>T p.P702S Chr.5: 102373408-102373408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372542541 Chr.5: 102373514-102373514 on Build GRCh38
rs769313361 Chr.5: 102373414-102373414 on Build GRCh38
rs755111748 Chr.5: 102373376-102373376 on Build GRCh38
rs1006265937 Chr.5: 102373375-102373375 on Build GRCh38
rs774754229 Chr.5: 102373515-102373515 on Build GRCh38
rs200960054 Chr.5: 102373427-102373427 on Build GRCh38
rs755866731 Chr.5: 102373476-102373476 on Build GRCh38
rs375156373 Chr.5: 102373478-102373478 on Build GRCh38
rs757674943 Chr.5: 102373468-102373468 on Build GRCh38
rs753026095 Chr.5: 102373529-102373530 on Build GRCh38
rs765670709 Chr.5: 102373379-102373379 on Build GRCh38
rs759739946 Chr.5: 102373446-102373446 on Build GRCh38
rs776837032 Chr.5: 102373388-102373389 on Build GRCh38
rs780805267 Chr.5: 102373469-102373469 on Build GRCh38
rs775487566 Chr.5: 102373419-102373419 on Build GRCh38
rs758961356 Chr.5: 102373386-102373386 on Build GRCh38
rs766374191 Chr.5: 102373386-102373388 on Build GRCh38
rs369430600 Chr.5: 102373438-102373438 on Build GRCh38
rs990011796 Chr.5: 102373440-102373440 on Build GRCh38
rs565584698 Chr.5: 102373387-102373387 on Build GRCh38
rs758789785 Chr.5: 102373530-102373531 on Build GRCh38
rs372939815 Chr.5: 102373369-102373369 on Build GRCh38
rs764119102 Chr.5: 102373362-102373362 on Build GRCh38
rs567010617 Chr.5: 102373435-102373435 on Build GRCh38
rs146381310 Chr.5: 102373370-102373370 on Build GRCh38
rs948084944 Chr.5: 102373470-102373470 on Build GRCh38
rs765432027 Chr.5: 102373441-102373441 on Build GRCh38
rs76193868 Chr.5: 102373384-102373384 on Build GRCh38
rs745575902 Chr.5: 102373391-102373391 on Build GRCh38
rs773054786 Chr.5: 102373376-102373381 on Build GRCh38
rs760759499 Chr.5: 102373379-102373381 on Build GRCh38
rs768501310 Chr.5: 102373430-102373430 on Build GRCh38
rs768590757 Chr.5: 102373489-102373489 on Build GRCh38
rs925469552 Chr.5: 102373374-102373374 on Build GRCh38
rs543024661 Chr.5: 102373550-102373550 on Build GRCh38
rs780528826 Chr.5: 102373389-102373389 on Build GRCh38
rs775702422 Chr.5: 102373541-102373541 on Build GRCh38
rs369551604 Chr.5: 102373507-102373507 on Build GRCh38
rs146658765 Chr.5: 102373454-102373454 on Build GRCh38
rs1017651439 Chr.5: 102373523-102373523 on Build GRCh38
rs756816051 Chr.5: 102373388-102373388 on Build GRCh38
rs753125945 Chr.5: 102373442-102373442 on Build GRCh38
rs539137924 Chr.5: 102373431-102373431 on Build GRCh38
rs750601411 Chr.5: 102373422-102373422 on Build GRCh38
rs369810298 Chr.5: 102373532-102373532 on Build GRCh38
rs759528774 Chr.5: 102373531-102373531 on Build GRCh38
rs769220825 Chr.5: 102373432-102373432 on Build GRCh38
rs773495931 Chr.5: 102373368-102373368 on Build GRCh38
rs377090264 Chr.5: 102373530-102373530 on Build GRCh38
rs143214362 Chr.5: 102373434-102373434 on Build GRCh38
rs752093945 Chr.5: 102373457-102373457 on Build GRCh38
rs200400953 Chr.5: 102373480-102373480 on Build GRCh38
rs765489946 Chr.5: 102373494-102373494 on Build GRCh38
rs758765105 Chr.5: 102373444-102373444 on Build GRCh38
rs748153829 Chr.5: 102373513-102373513 on Build GRCh38

View Full Product Details