Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40220561-40220790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUB1B
Entrez Gene ID
701
Gene Name
BUB1 mitotic checkpoint serine/threonine kinase B
Gene Aliases
BUB1beta, BUBR1, Bub1A, MAD3L, MVA1, SSK1, hBUBR1
UniGene
Hs.513645
Gene Chromosome Location
Chr.15: 40161009-40221136 on Build GRCh38
Transcripts
NM_001211.5 NM_001128628.2 NM_001128629.2

Gene Symbol
BUB1B-PAK6
Entrez Gene ID
106821730
Gene Name
BUB1B-PAK6 readthrough
Gene Aliases
-
UniGene
Hs.732375
Gene Chromosome Location
Chr.15: 40217428-40277487 on Build GRCh38
Transcripts
NM_001211.5 NM_001128628.2 NM_001128629.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6573453 c.3016G>C p.E1006Q Chr.15: 40220622-40220622 on Build GRCh38
3401695 c.3177G>A p.G1059G Chr.15: 40220741-40220741 on Build GRCh38
6478474 c.3037C>T p.R1013W Chr.15: 40220601-40220601 on Build GRCh38
1372607 c.3112G>A p.V1038I Chr.15: 40220718-40220718 on Build GRCh38
5019565 c.3136A>C p.N1046H Chr.15: 40220700-40220700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35611758 Chr.15: 40220705-40220705 on Build GRCh38
rs764992665 Chr.15: 40220731-40220731 on Build GRCh38
rs778324895 Chr.15: 40220675-40220675 on Build GRCh38
rs781682685 Chr.15: 40220649-40220649 on Build GRCh38
rs902224077 Chr.15: 40220621-40220621 on Build GRCh38
rs776542291 Chr.15: 40220719-40220719 on Build GRCh38
rs770219125 Chr.15: 40220680-40220680 on Build GRCh38
rs966176922 Chr.15: 40220642-40220642 on Build GRCh38
rs376467292 Chr.15: 40220753-40220753 on Build GRCh38
rs35008769 Chr.15: 40220720-40220721 on Build GRCh38
rs28989185 Chr.15: 40220641-40220641 on Build GRCh38
rs781600621 Chr.15: 40220619-40220619 on Build GRCh38
rs767561425 Chr.15: 40220768-40220768 on Build GRCh38
rs770279504 Chr.15: 40220669-40220669 on Build GRCh38
rs34700927 Chr.15: 40220700-40220700 on Build GRCh38
rs370506288 Chr.15: 40220626-40220626 on Build GRCh38
rs761776038 Chr.15: 40220730-40220730 on Build GRCh38
rs771709698 Chr.15: 40220682-40220682 on Build GRCh38
rs554186151 Chr.15: 40220742-40220742 on Build GRCh38
rs984548668 Chr.15: 40220762-40220762 on Build GRCh38
rs771453874 Chr.15: 40220600-40220600 on Build GRCh38
rs775040728 Chr.15: 40220699-40220699 on Build GRCh38
rs751744682 Chr.15: 40220752-40220752 on Build GRCh38
rs748723428 Chr.15: 40220655-40220655 on Build GRCh38
rs374968269 Chr.15: 40220634-40220634 on Build GRCh38
rs976402578 Chr.15: 40220676-40220676 on Build GRCh38
rs750610809 Chr.15: 40220628-40220628 on Build GRCh38
rs34998711 Chr.15: 40220617-40220617 on Build GRCh38
rs576684184 Chr.15: 40220711-40220711 on Build GRCh38
rs745415828 Chr.15: 40220766-40220767 on Build GRCh38
rs869239471 Chr.15: 40220659-40220660 on Build GRCh38
rs772941176 Chr.15: 40220738-40220738 on Build GRCh38
rs758666856 Chr.15: 40220632-40220632 on Build GRCh38
rs755017710 Chr.15: 40220644-40220644 on Build GRCh38
rs764057313 Chr.15: 40220607-40220607 on Build GRCh38
rs766555413 Chr.15: 40220748-40220748 on Build GRCh38
rs201246315 Chr.15: 40220601-40220601 on Build GRCh38
rs79104296 Chr.15: 40220643-40220643 on Build GRCh38
rs377612791 Chr.15: 40220602-40220602 on Build GRCh38

View Full Product Details