Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 50818777-50819038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFAP2B
Entrez Gene ID
7021
Gene Name
transcription factor AP-2 beta
Gene Aliases
AP-2B, AP2-B
UniGene
Hs.33102
Gene Chromosome Location
Chr.6: 50817692-50847619 on Build GRCh38
Transcripts
NM_003221.3 XM_011514837.2 XM_017011233.1 XM_017011234.1 XM_017011235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1080127 c.54G>T p.E18D Chr.6: 50818945-50818945 on Build GRCh38
5040674 c.40T>A p.W14R Chr.6: 50818931-50818931 on Build GRCh38
6174373 c.3G>T p.M1I Chr.6: 50818894-50818894 on Build GRCh38
5497835 c.4C>T p.H2Y Chr.6: 50818895-50818895 on Build GRCh38
299547 c.81+7C>T p.? Chr.6: 50818979-50818979 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781168856 Chr.6: 50819004-50819004 on Build GRCh38
rs148262187 Chr.6: 50818966-50818966 on Build GRCh38
rs371608614 Chr.6: 50818939-50818939 on Build GRCh38
rs1044221052 Chr.6: 50818886-50818886 on Build GRCh38
rs941552391 Chr.6: 50818995-50818995 on Build GRCh38
rs769420136 Chr.6: 50818964-50818966 on Build GRCh38
rs757855620 Chr.6: 50818996-50818996 on Build GRCh38
rs576874296 Chr.6: 50818854-50818854 on Build GRCh38
rs140236023 Chr.6: 50818950-50818950 on Build GRCh38
rs908688552 Chr.6: 50818942-50818942 on Build GRCh38
rs545478132 Chr.6: 50818855-50818855 on Build GRCh38
rs376735779 Chr.6: 50818899-50818899 on Build GRCh38
rs748140993 Chr.6: 50818893-50818893 on Build GRCh38
rs2272903 Chr.6: 50818858-50818858 on Build GRCh38
rs765975460 Chr.6: 50818999-50818999 on Build GRCh38
rs886061563 Chr.6: 50818809-50818809 on Build GRCh38
rs375744745 Chr.6: 50818883-50818883 on Build GRCh38
rs775800824 Chr.6: 50818979-50818979 on Build GRCh38
rs147456896 Chr.6: 50818920-50818920 on Build GRCh38
rs761236447 Chr.6: 50818986-50818986 on Build GRCh38
rs779698516 Chr.6: 50818873-50818873 on Build GRCh38
rs759841740 Chr.6: 50818969-50818969 on Build GRCh38
rs758043124 Chr.6: 50818863-50818863 on Build GRCh38
rs368444508 Chr.6: 50818901-50818901 on Build GRCh38
rs749335365 Chr.6: 50818919-50818919 on Build GRCh38
rs745825358 Chr.6: 50818875-50818886 on Build GRCh38
rs754760121 Chr.6: 50818882-50818882 on Build GRCh38
rs542117821 Chr.6: 50818987-50818987 on Build GRCh38
rs561980561 Chr.6: 50818988-50818988 on Build GRCh38
rs905725590 Chr.6: 50818888-50818888 on Build GRCh38
rs751171700 Chr.6: 50819001-50819001 on Build GRCh38
rs555045308 Chr.6: 50818798-50818798 on Build GRCh38
rs767918301 Chr.6: 50818978-50818978 on Build GRCh38
rs371429150 Chr.6: 50819003-50819003 on Build GRCh38
rs773487751 Chr.6: 50818902-50818902 on Build GRCh38
rs746783037 Chr.6: 50818877-50818877 on Build GRCh38
rs1019064015 Chr.6: 50818994-50818994 on Build GRCh38
rs769853484 Chr.6: 50818897-50818897 on Build GRCh38
rs753154944 Chr.6: 50818845-50818845 on Build GRCh38
rs1009895753 Chr.6: 50818984-50818984 on Build GRCh38
rs891431854 Chr.6: 50818972-50818972 on Build GRCh38

View Full Product Details