Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 86380647-86380906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WWP1
Entrez Gene ID
11059
Gene Name
WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases
AIP5, Tiul1, hSDRP1
UniGene
Hs.655189
Gene Chromosome Location
Chr.8: 86342711-86468357 on Build GRCh38
Transcripts
NM_007013.3 XM_005250760.3 XM_005250761.3 XM_011516802.2 XM_017012993.1 XM_017012994.1 XM_017012995.1 XM_017012996.1 XM_017012997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5552271 c.139G>C p.D47H Chr.8: 86380794-86380794 on Build GRCh38
5875069 c.179_180CT>TC p.S60F Chr.8: 86380834-86380835 on Build GRCh38
5555414 c.110G>A p.G37E Chr.8: 86380765-86380765 on Build GRCh38
6454244 c.199C>T p.Q67* Chr.8: 86380854-86380854 on Build GRCh38
6181870 c.154A>G p.K52E Chr.8: 86380809-86380809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553219752 Chr.8: 86380764-86380764 on Build GRCh38
rs764215672 Chr.8: 86380815-86380815 on Build GRCh38
rs757827524 Chr.8: 86380743-86380747 on Build GRCh38
rs779336984 Chr.8: 86380833-86380833 on Build GRCh38
rs1047871419 Chr.8: 86380706-86380706 on Build GRCh38
rs757550098 Chr.8: 86380823-86380823 on Build GRCh38
rs754013780 Chr.8: 86380819-86380819 on Build GRCh38
rs780614026 Chr.8: 86380747-86380748 on Build GRCh38
rs200829163 Chr.8: 86380715-86380715 on Build GRCh38
rs775669291 Chr.8: 86380717-86380717 on Build GRCh38
rs573309707 Chr.8: 86380808-86380808 on Build GRCh38
rs745725963 Chr.8: 86380695-86380695 on Build GRCh38
rs899766326 Chr.8: 86380826-86380826 on Build GRCh38
rs772123139 Chr.8: 86380696-86380696 on Build GRCh38
rs747027118 Chr.8: 86380716-86380719 on Build GRCh38
rs900507117 Chr.8: 86380773-86380773 on Build GRCh38
rs761421877 Chr.8: 86380763-86380763 on Build GRCh38
rs199755621 Chr.8: 86380713-86380713 on Build GRCh38
rs918163897 Chr.8: 86380680-86380680 on Build GRCh38
rs765269062 Chr.8: 86380697-86380697 on Build GRCh38
rs774682145 Chr.8: 86380766-86380766 on Build GRCh38
rs545485029 Chr.8: 86380814-86380814 on Build GRCh38
rs767773883 Chr.8: 86380807-86380807 on Build GRCh38
rs776053128 Chr.8: 86380732-86380732 on Build GRCh38
rs746102182 Chr.8: 86380863-86380863 on Build GRCh38
rs201729551 Chr.8: 86380681-86380681 on Build GRCh38
rs759833865 Chr.8: 86380805-86380805 on Build GRCh38

View Full Product Details