Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 16208442-16208715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT15
Entrez Gene ID
117248
Gene Name
polypeptide N-acetylgalactosaminyltransferase 15
Gene Aliases
GALNACT15, GALNT13, GALNT7, GALNTL2, PIH5, pp-GalNAc-T15
UniGene
Hs.411308
Gene Chromosome Location
Chr.3: 16174322-16245190 on Build GRCh38
Transcripts
NM_001319051.1 NM_054110.4 XM_005264852.4 XM_017005682.1 XM_017005683.1 XM_017005684.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5974619 c.1018C>T p.H340Y Chr.3: 16208609-16208609 on Build GRCh38
3589517 c.972C>T p.P324P Chr.3: 16208563-16208563 on Build GRCh38
4415176 c.970C>G p.P324A Chr.3: 16208561-16208561 on Build GRCh38
5572578 c.965A>C p.Y322S Chr.3: 16208556-16208556 on Build GRCh38
5034798 c.1079+4A>T p.? Chr.3: 16208674-16208674 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754940195 Chr.3: 16208670-16208670 on Build GRCh38
rs771511992 Chr.3: 16208585-16208585 on Build GRCh38
rs538260003 Chr.3: 16208471-16208471 on Build GRCh38
rs140097676 Chr.3: 16208660-16208660 on Build GRCh38
rs112698459 Chr.3: 16208521-16208521 on Build GRCh38
rs778214547 Chr.3: 16208578-16208578 on Build GRCh38
rs758266927 Chr.3: 16208560-16208560 on Build GRCh38
rs12634179 Chr.3: 16208561-16208561 on Build GRCh38
rs747808637 Chr.3: 16208500-16208500 on Build GRCh38
rs746532933 Chr.3: 16208489-16208489 on Build GRCh38
rs752256158 Chr.3: 16208675-16208675 on Build GRCh38
rs780743995 Chr.3: 16208674-16208674 on Build GRCh38
rs867286394 Chr.3: 16208479-16208479 on Build GRCh38
rs774899159 Chr.3: 16208587-16208587 on Build GRCh38
rs764306829 Chr.3: 16208618-16208618 on Build GRCh38
rs750178429 Chr.3: 16208622-16208622 on Build GRCh38
rs748995005 Chr.3: 16208689-16208689 on Build GRCh38
rs760829679 Chr.3: 16208610-16208610 on Build GRCh38
rs750987843 Chr.3: 16208562-16208562 on Build GRCh38
rs199803852 Chr.3: 16208612-16208612 on Build GRCh38
rs377237497 Chr.3: 16208520-16208520 on Build GRCh38
rs150527566 Chr.3: 16208604-16208604 on Build GRCh38
rs765002678 Chr.3: 16208531-16208531 on Build GRCh38
rs769580462 Chr.3: 16208573-16208573 on Build GRCh38
rs779484394 Chr.3: 16208485-16208485 on Build GRCh38
rs774250982 Chr.3: 16208512-16208512 on Build GRCh38
rs763493649 Chr.3: 16208507-16208507 on Build GRCh38
rs774152107 Chr.3: 16208671-16208671 on Build GRCh38
rs372855028 Chr.3: 16208508-16208508 on Build GRCh38
rs766139523 Chr.3: 16208655-16208655 on Build GRCh38
rs758389934 Chr.3: 16208483-16208483 on Build GRCh38
rs762844702 Chr.3: 16208564-16208564 on Build GRCh38
rs755785090 Chr.3: 16208677-16208677 on Build GRCh38
rs191212291 Chr.3: 16208468-16208468 on Build GRCh38
rs372600104 Chr.3: 16208589-16208589 on Build GRCh38
rs540396167 Chr.3: 16208668-16208668 on Build GRCh38
rs766217552 Chr.3: 16208675-16208675 on Build GRCh38
rs754493057 Chr.3: 16208565-16208565 on Build GRCh38
rs773782318 Chr.3: 16208504-16208504 on Build GRCh38
rs370149856 Chr.3: 16208572-16208572 on Build GRCh38
rs770254825 Chr.3: 16208503-16208503 on Build GRCh38
rs377656578 Chr.3: 16208478-16208478 on Build GRCh38
rs772185988 Chr.3: 16208603-16208603 on Build GRCh38
rs777586328 Chr.3: 16208688-16208688 on Build GRCh38
rs958216507 Chr.3: 16208629-16208629 on Build GRCh38
rs762760508 Chr.3: 16208644-16208644 on Build GRCh38
rs780610574 Chr.3: 16208569-16208569 on Build GRCh38
rs750233423 Chr.3: 16208547-16208547 on Build GRCh38
rs200511414 Chr.3: 16208579-16208579 on Build GRCh38
rs1505609 Chr.3: 16208491-16208491 on Build GRCh38

View Full Product Details