Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 53238088-53238349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQSEC2
Entrez Gene ID
23096
Gene Name
IQ motif and Sec7 domain 2
Gene Aliases
BRAG1, MRX1, MRX78
UniGene
Hs.496138
Gene Chromosome Location
Chr.X: 53225784-53321350 on Build GRCh38
Transcripts
NM_001111125.2 NM_015075.1 XM_006724579.2 XM_006724580.3 XM_006724581.3 XM_006724582.3 XM_006724583.3 XM_006724584.2 XM_011530773.2 XM_011530774.2 XM_011530776.1 XM_011530777.1 XM_017029359.1 XM_017029360.1

SNP Detail

SNP ID Chromosome Location
rs782263065 Chr.X: 53238244-53238244 on Build GRCh38
rs782715847 Chr.X: 53238180-53238180 on Build GRCh38
rs143966238 Chr.X: 53238127-53238127 on Build GRCh38
rs371502883 Chr.X: 53238310-53238310 on Build GRCh38
rs367700660 Chr.X: 53238229-53238229 on Build GRCh38
rs782193491 Chr.X: 53238311-53238311 on Build GRCh38
rs782437696 Chr.X: 53238179-53238179 on Build GRCh38
rs782569979 Chr.X: 53238193-53238193 on Build GRCh38
rs782735027 Chr.X: 53238149-53238149 on Build GRCh38
rs782123939 Chr.X: 53238130-53238130 on Build GRCh38
rs781899074 Chr.X: 53238191-53238191 on Build GRCh38
rs782632137 Chr.X: 53238216-53238216 on Build GRCh38
rs973062974 Chr.X: 53238256-53238256 on Build GRCh38
rs797045140 Chr.X: 53238308-53238309 on Build GRCh38
rs782264682 Chr.X: 53238325-53238325 on Build GRCh38
rs368127921 Chr.X: 53238123-53238123 on Build GRCh38
rs782137079 Chr.X: 53238119-53238119 on Build GRCh38
rs797045627 Chr.X: 53238217-53238217 on Build GRCh38
rs375024113 Chr.X: 53238173-53238173 on Build GRCh38
rs199643273 Chr.X: 53238263-53238263 on Build GRCh38

View Full Product Details