Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 45136534-45136795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2F2
Entrez Gene ID
2963
Gene Name
general transcription factor IIF subunit 2
Gene Aliases
BTF4, RAP30, TF2F2, TFIIF
UniGene
Hs.654582
Gene Chromosome Location
Chr.13: 45119830-45284105 on Build GRCh38
Transcripts
NM_004128.2 XM_011535052.2 XM_011535053.2 XM_017020550.1 XM_017020551.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1662092 c.94G>C p.A32P Chr.13: 45136760-45136760 on Build GRCh38
6676698 c.87G>A p.Q29Q Chr.13: 45136753-45136753 on Build GRCh38
244840 c.100G>A p.A34T Chr.13: 45136766-45136766 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1024350838 Chr.13: 45136608-45136608 on Build GRCh38
rs936259854 Chr.13: 45136685-45136685 on Build GRCh38
rs759047531 Chr.13: 45136711-45136711 on Build GRCh38
rs746107381 Chr.13: 45136618-45136618 on Build GRCh38
rs369599150 Chr.13: 45136687-45136687 on Build GRCh38
rs958672376 Chr.13: 45136584-45136584 on Build GRCh38
rs772623280 Chr.13: 45136688-45136688 on Build GRCh38
rs190042066 Chr.13: 45136677-45136677 on Build GRCh38
rs111387853 Chr.13: 45136760-45136760 on Build GRCh38
rs764129745 Chr.13: 45136731-45136731 on Build GRCh38
rs755274273 Chr.13: 45136770-45136770 on Build GRCh38
rs776169934 Chr.13: 45136702-45136702 on Build GRCh38
rs200368894 Chr.13: 45136744-45136744 on Build GRCh38
rs773564996 Chr.13: 45136689-45136689 on Build GRCh38
rs115909751 Chr.13: 45136723-45136723 on Build GRCh38
rs569900224 Chr.13: 45136576-45136576 on Build GRCh38
rs145466997 Chr.13: 45136753-45136753 on Build GRCh38
rs139949632 Chr.13: 45136579-45136579 on Build GRCh38
rs757139282 Chr.13: 45136733-45136733 on Build GRCh38
rs762976872 Chr.13: 45136729-45136729 on Build GRCh38
rs753972000 Chr.13: 45136756-45136756 on Build GRCh38
rs373769946 Chr.13: 45136697-45136697 on Build GRCh38
rs9534041 Chr.13: 45136613-45136613 on Build GRCh38
rs185135542 Chr.13: 45136610-45136610 on Build GRCh38
rs750130020 Chr.13: 45136772-45136772 on Build GRCh38
rs3803244 Chr.13: 45136630-45136630 on Build GRCh38
rs566387212 Chr.13: 45136622-45136622 on Build GRCh38
rs764623115 Chr.13: 45136719-45136719 on Build GRCh38
rs892197498 Chr.13: 45136647-45136647 on Build GRCh38
rs571654691 Chr.13: 45136591-45136591 on Build GRCh38
rs770517057 Chr.13: 45136699-45136699 on Build GRCh38
rs747381194 Chr.13: 45136691-45136691 on Build GRCh38
rs202043550 Chr.13: 45136701-45136701 on Build GRCh38
rs561017789 Chr.13: 45136575-45136575 on Build GRCh38
rs199678448 Chr.13: 45136765-45136765 on Build GRCh38
rs370193338 Chr.13: 45136732-45136732 on Build GRCh38

View Full Product Details