Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 103122904-103123141 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPE
Entrez Gene ID
1062
Gene Name
centromere protein E
Gene Aliases
CENP-E, KIF10, MCPH13, PPP1R61
UniGene
Hs.75573
Gene Chromosome Location
Chr.4: 103105806-103198409 on Build GRCh38
Transcripts
NM_001286734.1 NM_001813.2 XM_011531544.2 XM_011531545.2 XM_011531546.2 XM_011531547.2 XM_011531548.2 XM_011531549.2 XM_017007659.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3916885 c.6995C>T p.P2332L Chr.4: 103122908-103122908 on Build GRCh38
4860826 c.6960A>C p.R2320S Chr.4: 103123054-103123054 on Build GRCh38
732076 c.6849A>C p.R2283S Chr.4: 103123054-103123054 on Build GRCh38
3916884 c.7106C>T p.P2369L Chr.4: 103122908-103122908 on Build GRCh38
5686149 c.6966T>A p.A2322A Chr.4: 103123048-103123048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61751592 Chr.4: 103123040-103123040 on Build GRCh38
rs748559290 Chr.4: 103123062-103123062 on Build GRCh38
rs749934583 Chr.4: 103123041-103123041 on Build GRCh38
rs111281600 Chr.4: 103123086-103123086 on Build GRCh38
rs758298907 Chr.4: 103123082-103123082 on Build GRCh38
rs779493574 Chr.4: 103123050-103123050 on Build GRCh38
rs373965211 Chr.4: 103123029-103123029 on Build GRCh38
rs755515319 Chr.4: 103123044-103123044 on Build GRCh38
rs751254711 Chr.4: 103123018-103123018 on Build GRCh38
rs775234859 Chr.4: 103123009-103123009 on Build GRCh38
rs758835317 Chr.4: 103123079-103123079 on Build GRCh38
rs747200493 Chr.4: 103122964-103122964 on Build GRCh38
rs748556175 Chr.4: 103122945-103122945 on Build GRCh38
rs762829111 Chr.4: 103123011-103123011 on Build GRCh38
rs747049261 Chr.4: 103123108-103123109 on Build GRCh38
rs910502686 Chr.4: 103122947-103122947 on Build GRCh38
rs199570709 Chr.4: 103122952-103122952 on Build GRCh38
rs773511630 Chr.4: 103122961-103122961 on Build GRCh38
rs747290658 Chr.4: 103123089-103123089 on Build GRCh38
rs765224508 Chr.4: 103122993-103122993 on Build GRCh38
rs560971961 Chr.4: 103122944-103122944 on Build GRCh38
rs1042931771 Chr.4: 103122979-103122979 on Build GRCh38
rs776836410 Chr.4: 103122981-103122981 on Build GRCh38
rs201826180 Chr.4: 103123014-103123014 on Build GRCh38
rs771182826 Chr.4: 103122977-103122977 on Build GRCh38
rs115015192 Chr.4: 103122988-103122988 on Build GRCh38
rs780307648 Chr.4: 103123025-103123025 on Build GRCh38

View Full Product Details