Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 52845880-52846129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDH8
Entrez Gene ID
5100
Gene Name
protocadherin 8
Gene Aliases
ARCADLIN, PAPC
UniGene
Hs.19492
Gene Chromosome Location
Chr.13: 52843974-52848640 on Build GRCh38
Transcripts
NM_002590.3 NM_032949.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5951513 c.2507C>T p.A836V Chr.13: 52845930-52845930 on Build GRCh38
6333385 c.2468C>A p.T823N Chr.13: 52845969-52845969 on Build GRCh38
5625900 c.2414C>G p.P805R Chr.13: 52846023-52846023 on Build GRCh38
1261212 c.2392G>A p.G798R Chr.13: 52846045-52846045 on Build GRCh38
1943574 c.2377G>A p.G793R Chr.13: 52846060-52846060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769279982 Chr.13: 52846028-52846028 on Build GRCh38
rs1049765204 Chr.13: 52846022-52846022 on Build GRCh38
rs964958368 Chr.13: 52846012-52846012 on Build GRCh38
rs779577577 Chr.13: 52846083-52846083 on Build GRCh38
rs752955744 Chr.13: 52846004-52846004 on Build GRCh38
rs1017907190 Chr.13: 52846001-52846001 on Build GRCh38
rs745871248 Chr.13: 52846026-52846026 on Build GRCh38
rs932673094 Chr.13: 52846021-52846021 on Build GRCh38
rs907179114 Chr.13: 52846092-52846092 on Build GRCh38
rs756049482 Chr.13: 52846081-52846081 on Build GRCh38
rs757040442 Chr.13: 52846080-52846080 on Build GRCh38
rs760057765 Chr.13: 52846089-52846089 on Build GRCh38
rs866577694 Chr.13: 52845936-52845936 on Build GRCh38
rs149674876 Chr.13: 52845930-52845930 on Build GRCh38
rs761100634 Chr.13: 52846053-52846053 on Build GRCh38
rs200182252 Chr.13: 52845928-52845928 on Build GRCh38
rs965973986 Chr.13: 52846000-52846000 on Build GRCh38
rs568650642 Chr.13: 52846023-52846023 on Build GRCh38
rs758592537 Chr.13: 52846073-52846073 on Build GRCh38
rs1042549078 Chr.13: 52846066-52846066 on Build GRCh38
rs765306356 Chr.13: 52846065-52846065 on Build GRCh38
rs759020777 Chr.13: 52845923-52845923 on Build GRCh38
rs376226669 Chr.13: 52846086-52846086 on Build GRCh38
rs1047810990 Chr.13: 52846111-52846111 on Build GRCh38
rs531179041 Chr.13: 52846030-52846030 on Build GRCh38
rs563281878 Chr.13: 52845902-52845902 on Build GRCh38
rs769332896 Chr.13: 52845927-52845927 on Build GRCh38
rs528207061 Chr.13: 52845998-52845998 on Build GRCh38
rs946797218 Chr.13: 52846061-52846061 on Build GRCh38
rs865956339 Chr.13: 52846095-52846095 on Build GRCh38
rs765681481 Chr.13: 52846100-52846100 on Build GRCh38
rs746894409 Chr.13: 52845918-52845918 on Build GRCh38
rs767864013 Chr.13: 52845932-52845932 on Build GRCh38
rs754447078 Chr.13: 52845992-52845992 on Build GRCh38
rs571170434 Chr.13: 52846088-52846088 on Build GRCh38
rs777491120 Chr.13: 52845909-52845909 on Build GRCh38
rs551287635 Chr.13: 52846087-52846087 on Build GRCh38
rs758566629 Chr.13: 52846005-52846005 on Build GRCh38
rs759405347 Chr.13: 52845920-52845920 on Build GRCh38
rs868863030 Chr.13: 52845961-52845961 on Build GRCh38
rs748712848 Chr.13: 52846042-52846043 on Build GRCh38
rs777275555 Chr.13: 52846060-52846060 on Build GRCh38
rs17857085 Chr.13: 52845937-52845937 on Build GRCh38
rs766893845 Chr.13: 52846056-52846056 on Build GRCh38
rs201133850 Chr.13: 52846099-52846099 on Build GRCh38
rs770881105 Chr.13: 52845919-52845919 on Build GRCh38
rs757089548 Chr.13: 52846008-52846008 on Build GRCh38
rs777581388 Chr.13: 52846006-52846006 on Build GRCh38
rs548016632 Chr.13: 52846002-52846002 on Build GRCh38
rs559153648 Chr.13: 52845980-52845980 on Build GRCh38
rs147568115 Chr.13: 52846113-52846117 on Build GRCh38
rs922594376 Chr.13: 52846013-52846013 on Build GRCh38
rs776109951 Chr.13: 52846113-52846113 on Build GRCh38
rs770663005 Chr.13: 52846080-52846081 on Build GRCh38
rs915305709 Chr.13: 52846049-52846049 on Build GRCh38
rs976427422 Chr.13: 52846009-52846009 on Build GRCh38
rs773578849 Chr.13: 52846051-52846051 on Build GRCh38
rs750758713 Chr.13: 52845945-52845945 on Build GRCh38
rs767018492 Chr.13: 52845991-52845991 on Build GRCh38
rs764198033 Chr.13: 52846074-52846074 on Build GRCh38
rs761223699 Chr.13: 52845989-52845989 on Build GRCh38
rs746799526 Chr.13: 52846007-52846007 on Build GRCh38
rs768224218 Chr.13: 52846043-52846043 on Build GRCh38
rs774094455 Chr.13: 52846100-52846102 on Build GRCh38

View Full Product Details