Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 3612103-3612376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKX
Entrez Gene ID
5613
Gene Name
protein kinase, X-linked
Gene Aliases
PKX1
UniGene
Hs.390788
Gene Chromosome Location
Chr.X: 3604343-3713634 on Build GRCh38
Transcripts
NM_005044.4

SNP Detail

SNP ID Chromosome Location
rs751094148 Chr.X: 3612282-3612282 on Build GRCh38
rs778444616 Chr.X: 3612133-3612133 on Build GRCh38
rs774876022 Chr.X: 3612227-3612229 on Build GRCh38
rs765742917 Chr.X: 3612281-3612281 on Build GRCh38
rs762522125 Chr.X: 3612278-3612278 on Build GRCh38
rs768718627 Chr.X: 3612350-3612350 on Build GRCh38
rs376971341 Chr.X: 3612260-3612260 on Build GRCh38
rs2287236 Chr.X: 3612153-3612153 on Build GRCh38
rs45588733 Chr.X: 3612295-3612295 on Build GRCh38
rs1032539347 Chr.X: 3612212-3612212 on Build GRCh38
rs145173948 Chr.X: 3612239-3612239 on Build GRCh38
rs149150830 Chr.X: 3612243-3612243 on Build GRCh38
rs779936306 Chr.X: 3612343-3612343 on Build GRCh38
rs141700607 Chr.X: 3612317-3612317 on Build GRCh38
rs141993152 Chr.X: 3612292-3612292 on Build GRCh38
rs766045812 Chr.X: 3612186-3612186 on Build GRCh38
rs757899721 Chr.X: 3612183-3612183 on Build GRCh38
rs780794643 Chr.X: 3612198-3612198 on Build GRCh38
rs149734060 Chr.X: 3612164-3612164 on Build GRCh38
rs993702481 Chr.X: 3612141-3612141 on Build GRCh38
rs754664396 Chr.X: 3612191-3612191 on Build GRCh38
rs149966538 Chr.X: 3612234-3612234 on Build GRCh38
rs958280376 Chr.X: 3612293-3612293 on Build GRCh38
rs139150166 Chr.X: 3612316-3612316 on Build GRCh38
rs200967884 Chr.X: 3612245-3612245 on Build GRCh38
rs146727237 Chr.X: 3612296-3612296 on Build GRCh38
rs745335498 Chr.X: 3612136-3612136 on Build GRCh38
rs757223132 Chr.X: 3612341-3612341 on Build GRCh38
rs769212571 Chr.X: 3612190-3612190 on Build GRCh38
rs747979274 Chr.X: 3612226-3612226 on Build GRCh38
rs775774808 Chr.X: 3612242-3612242 on Build GRCh38
rs765714555 Chr.X: 3612253-3612253 on Build GRCh38
rs762248323 Chr.X: 3612150-3612150 on Build GRCh38
rs747130149 Chr.X: 3612345-3612345 on Build GRCh38
rs55657449 Chr.X: 3612334-3612334 on Build GRCh38
rs756075852 Chr.X: 3612231-3612231 on Build GRCh38
rs12395404 Chr.X: 3612149-3612149 on Build GRCh38

View Full Product Details