Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15415723-15415996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL6
Entrez Gene ID
131965
Gene Name
methyltransferase like 6
Gene Aliases
-
UniGene
Hs.149487
Gene Chromosome Location
Chr.3: 15381275-15427580 on Build GRCh38
Transcripts
NM_001301790.1 NM_001301791.1 NM_001301792.1 NM_152396.3 XM_005264867.3 XM_005264870.4 XM_006712970.3 XM_006712972.3 XM_011533357.2 XM_017005718.1 XM_017005719.1 XM_017005720.1 XM_017005721.1 XM_017005722.1 XM_017005723.1 XM_017005724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7191732 c.335T>C p.I112T Chr.3: 15415833-15415833 on Build GRCh38
728631 c.452T>C p.V151A Chr.3: 15415851-15415851 on Build GRCh38
4490706 c.367C>T p.P123S Chr.3: 15415936-15415936 on Build GRCh38
7412913 c.464T>C p.M155T Chr.3: 15415839-15415839 on Build GRCh38
7412914 c.329T>C p.M110T Chr.3: 15415839-15415839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879011196 Chr.3: 15415889-15415889 on Build GRCh38
rs913481660 Chr.3: 15415775-15415775 on Build GRCh38
rs372239066 Chr.3: 15415909-15415909 on Build GRCh38
rs751792903 Chr.3: 15415911-15415911 on Build GRCh38
rs779056188 Chr.3: 15415750-15415750 on Build GRCh38
rs750570151 Chr.3: 15415780-15415780 on Build GRCh38
rs769652446 Chr.3: 15415796-15415796 on Build GRCh38
rs781044299 Chr.3: 15415958-15415958 on Build GRCh38
rs770836565 Chr.3: 15415816-15415816 on Build GRCh38
rs770875857 Chr.3: 15415937-15415937 on Build GRCh38
rs771734569 Chr.3: 15415962-15415962 on Build GRCh38
rs775315969 Chr.3: 15415840-15415840 on Build GRCh38
rs758502435 Chr.3: 15415890-15415890 on Build GRCh38
rs775229518 Chr.3: 15415963-15415963 on Build GRCh38
rs868214621 Chr.3: 15415839-15415839 on Build GRCh38
rs773982574 Chr.3: 15415820-15415820 on Build GRCh38
rs1009123274 Chr.3: 15415852-15415852 on Build GRCh38
rs760903367 Chr.3: 15415841-15415841 on Build GRCh38
rs758607784 Chr.3: 15415785-15415785 on Build GRCh38
rs780293006 Chr.3: 15415786-15415786 on Build GRCh38
rs774245490 Chr.3: 15415947-15415947 on Build GRCh38
rs374633544 Chr.3: 15415917-15415917 on Build GRCh38
rs777616158 Chr.3: 15415914-15415914 on Build GRCh38
rs750482105 Chr.3: 15415867-15415867 on Build GRCh38
rs892596636 Chr.3: 15415868-15415868 on Build GRCh38
rs757209185 Chr.3: 15415752-15415752 on Build GRCh38
rs1009732259 Chr.3: 15415884-15415884 on Build GRCh38
rs368735587 Chr.3: 15415959-15415959 on Build GRCh38
rs749091689 Chr.3: 15415931-15415931 on Build GRCh38
rs765792385 Chr.3: 15415863-15415863 on Build GRCh38
rs759163311 Chr.3: 15415834-15415834 on Build GRCh38
rs754231979 Chr.3: 15415860-15415860 on Build GRCh38
rs762378688 Chr.3: 15415862-15415862 on Build GRCh38
rs747896266 Chr.3: 15415795-15415795 on Build GRCh38
rs186541587 Chr.3: 15415859-15415859 on Build GRCh38
rs368277918 Chr.3: 15415797-15415797 on Build GRCh38
rs749249540 Chr.3: 15415810-15415810 on Build GRCh38

View Full Product Details