Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 65528950-65529183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPK
Entrez Gene ID
64105
Gene Name
centromere protein K
Gene Aliases
AF5alpha, CENP-K, FKSG14, P33, Solt
UniGene
Hs.529778
Gene Chromosome Location
Chr.5: 65488919-65563194 on Build GRCh38
Transcripts
NM_001267038.1 NM_022145.4 XM_011543534.2 XM_011543535.2 XM_011543536.2 XM_011543537.2 XM_011543538.2 XM_011543539.2 XM_011543540.2 XM_011543541.2 XM_011543542.2 XM_017009690.1 XM_017009691.1 XM_017009692.1 XM_017009693.1 XM_017009694.1 XM_017009695.1 XM_017009696.1 XM_017009697.1 XM_017009698.1 XM_017009699.1 XM_017009700.1 XM_017009701.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1069345 c.438G>C p.L146F Chr.5: 65528951-65528951 on Build GRCh38
738385 c.380G>T p.R127L Chr.5: 65529009-65529009 on Build GRCh38
1438169 c.422T>C p.V141A Chr.5: 65528967-65528967 on Build GRCh38
7302497 c.337T>C p.S113P Chr.5: 65529151-65529151 on Build GRCh38
4488144 c.327C>T p.S109S Chr.5: 65529161-65529161 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766439178 Chr.5: 65529090-65529090 on Build GRCh38
rs753691260 Chr.5: 65529142-65529142 on Build GRCh38
rs138940796 Chr.5: 65529009-65529009 on Build GRCh38
rs909927578 Chr.5: 65529119-65529119 on Build GRCh38
rs768317556 Chr.5: 65529150-65529150 on Build GRCh38
rs762682516 Chr.5: 65529040-65529040 on Build GRCh38
rs780195488 Chr.5: 65528997-65528997 on Build GRCh38
rs755202958 Chr.5: 65529100-65529100 on Build GRCh38
rs892129867 Chr.5: 65529000-65529000 on Build GRCh38
rs145897680 Chr.5: 65529143-65529143 on Build GRCh38
rs140748564 Chr.5: 65529129-65529129 on Build GRCh38
rs746826114 Chr.5: 65529018-65529018 on Build GRCh38
rs976731015 Chr.5: 65529083-65529083 on Build GRCh38
rs954261149 Chr.5: 65529019-65529019 on Build GRCh38
rs751328750 Chr.5: 65529114-65529114 on Build GRCh38
rs771237242 Chr.5: 65529075-65529078 on Build GRCh38
rs775207225 Chr.5: 65529033-65529033 on Build GRCh38
rs201048796 Chr.5: 65529055-65529055 on Build GRCh38
rs760676434 Chr.5: 65529088-65529088 on Build GRCh38
rs768023361 Chr.5: 65529078-65529078 on Build GRCh38
rs762118459 Chr.5: 65529076-65529076 on Build GRCh38
rs1056941169 Chr.5: 65529109-65529112 on Build GRCh38
rs370658786 Chr.5: 65529084-65529084 on Build GRCh38
rs923992690 Chr.5: 65529077-65529077 on Build GRCh38
rs143054455 Chr.5: 65529060-65529060 on Build GRCh38
rs748850005 Chr.5: 65529147-65529147 on Build GRCh38
rs79197620 Chr.5: 65529023-65529023 on Build GRCh38
rs984249753 Chr.5: 65529112-65529112 on Build GRCh38
rs377588006 Chr.5: 65529022-65529022 on Build GRCh38
rs755218264 Chr.5: 65529010-65529010 on Build GRCh38
rs779041711 Chr.5: 65529105-65529105 on Build GRCh38
rs757023799 Chr.5: 65529115-65529115 on Build GRCh38
rs769434490 Chr.5: 65529027-65529027 on Build GRCh38
rs867891240 Chr.5: 65528998-65528998 on Build GRCh38
rs781217896 Chr.5: 65529116-65529116 on Build GRCh38
rs185309293 Chr.5: 65529011-65529011 on Build GRCh38
rs753974698 Chr.5: 65529094-65529094 on Build GRCh38
rs745797201 Chr.5: 65529125-65529125 on Build GRCh38

View Full Product Details