Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 21517072-21517297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SKIDA1
Entrez Gene ID
387640
Gene Name
SKI/DACH domain containing 1
Gene Aliases
C10orf140, DLN-1
UniGene
Hs.350848
Gene Chromosome Location
Chr.10: 21513475-21525682 on Build GRCh38
Transcripts
NM_207371.3 XM_005252450.3 XM_011519480.2 XM_017016225.1 XM_017016226.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5472996 c.702T>C p.A234A Chr.10: 21517121-21517121 on Build GRCh38
6850820 c.714C>T p.A238A Chr.10: 21517109-21517109 on Build GRCh38
7272453 c.715G>A p.A239T Chr.10: 21517108-21517108 on Build GRCh38
6850818 c.711C>T p.A237A Chr.10: 21517112-21517112 on Build GRCh38
5483460 c.696C>T p.A232A Chr.10: 21517127-21517127 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773056898 Chr.10: 21517119-21517121 on Build GRCh38
rs868155746 Chr.10: 21517121-21517121 on Build GRCh38
rs749937295 Chr.10: 21517252-21517252 on Build GRCh38
rs370781497 Chr.10: 21517277-21517277 on Build GRCh38
rs1027699107 Chr.10: 21517276-21517276 on Build GRCh38
rs767145419 Chr.10: 21517214-21517214 on Build GRCh38
rs754621872 Chr.10: 21517125-21517133 on Build GRCh38
rs541879371 Chr.10: 21517118-21517118 on Build GRCh38
rs765891124 Chr.10: 21517124-21517125 on Build GRCh38
rs377557900 Chr.10: 21517258-21517258 on Build GRCh38
rs983418547 Chr.10: 21517279-21517279 on Build GRCh38
rs767464686 Chr.10: 21517101-21517124 on Build GRCh38
rs779528225 Chr.10: 21517265-21517265 on Build GRCh38
rs1014436836 Chr.10: 21517179-21517179 on Build GRCh38
rs906136033 Chr.10: 21517246-21517246 on Build GRCh38
rs1024868879 Chr.10: 21517229-21517229 on Build GRCh38
rs764855836 Chr.10: 21517125-21517130 on Build GRCh38
rs868442911 Chr.10: 21517124-21517124 on Build GRCh38
rs751205037 Chr.10: 21517169-21517169 on Build GRCh38
rs763904059 Chr.10: 21517127-21517127 on Build GRCh38
rs760132318 Chr.10: 21517125-21517127 on Build GRCh38
rs993815009 Chr.10: 21517107-21517124 on Build GRCh38
rs756871409 Chr.10: 21517202-21517202 on Build GRCh38

View Full Product Details