Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 206541939-206542194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM23
Entrez Gene ID
8745
Gene Name
ADAM metallopeptidase domain 23
Gene Aliases
MDC-3, MDC3
UniGene
Hs.370287
Gene Chromosome Location
Chr.2: 206443544-206621130 on Build GRCh38
Transcripts
NM_003812.3 XM_005246932.3 XM_011512086.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1482689 c.656+1G>A p.? Chr.2: 206542135-206542135 on Build GRCh38
4090556 c.612C>T p.G204G Chr.2: 206542090-206542090 on Build GRCh38
1482687 c.600A>G p.G200G Chr.2: 206542078-206542078 on Build GRCh38
3933518 c.656A>G p.H219R Chr.2: 206542134-206542134 on Build GRCh38
6090431 c.607A>G p.R203G Chr.2: 206542085-206542085 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780983131 Chr.2: 206542036-206542036 on Build GRCh38
rs568263812 Chr.2: 206541985-206541985 on Build GRCh38
rs995086915 Chr.2: 206541971-206541982 on Build GRCh38
rs767246517 Chr.2: 206542161-206542161 on Build GRCh38
rs750612641 Chr.2: 206542060-206542060 on Build GRCh38
rs924525604 Chr.2: 206542055-206542055 on Build GRCh38
rs968528046 Chr.2: 206541996-206541996 on Build GRCh38
rs764101026 Chr.2: 206542012-206542012 on Build GRCh38
rs370233704 Chr.2: 206542091-206542091 on Build GRCh38
rs1029106581 Chr.2: 206542146-206542146 on Build GRCh38
rs75766366 Chr.2: 206542023-206542023 on Build GRCh38
rs368058165 Chr.2: 206542112-206542112 on Build GRCh38
rs756318376 Chr.2: 206542088-206542088 on Build GRCh38
rs761741640 Chr.2: 206542158-206542158 on Build GRCh38
rs914385070 Chr.2: 206541967-206541967 on Build GRCh38
rs373442262 Chr.2: 206542154-206542154 on Build GRCh38
rs376542615 Chr.2: 206542002-206542002 on Build GRCh38
rs577038898 Chr.2: 206542150-206542150 on Build GRCh38
rs750181192 Chr.2: 206542166-206542166 on Build GRCh38
rs1005139516 Chr.2: 206542103-206542103 on Build GRCh38
rs377435335 Chr.2: 206542170-206542170 on Build GRCh38
rs756988710 Chr.2: 206542032-206542032 on Build GRCh38
rs759146072 Chr.2: 206542121-206542121 on Build GRCh38
rs771182635 Chr.2: 206541991-206541991 on Build GRCh38
rs901879815 Chr.2: 206542114-206542114 on Build GRCh38
rs142194045 Chr.2: 206542104-206542104 on Build GRCh38
rs769522829 Chr.2: 206542124-206542124 on Build GRCh38
rs1049245295 Chr.2: 206542048-206542048 on Build GRCh38
rs554123540 Chr.2: 206542095-206542095 on Build GRCh38
rs759098554 Chr.2: 206542167-206542168 on Build GRCh38
rs780126514 Chr.2: 206542090-206542090 on Build GRCh38
rs151048392 Chr.2: 206542125-206542125 on Build GRCh38
rs548448744 Chr.2: 206541971-206541971 on Build GRCh38
rs764194899 Chr.2: 206542151-206542151 on Build GRCh38
rs769187767 Chr.2: 206542092-206542092 on Build GRCh38

View Full Product Details