Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 52237854-52238121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFYVE9
Entrez Gene ID
9372
Gene Name
zinc finger FYVE-type containing 9
Gene Aliases
MADHIP, NSP, PPP1R173, SARA, SMADIP
UniGene
Hs.532345
Gene Chromosome Location
Chr.1: 52142001-52348664 on Build GRCh38
Transcripts
NM_004799.3 NM_007324.3 XM_011542437.2 XM_017002845.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6827056 c.630G>T p.E210D Chr.1: 52238047-52238047 on Build GRCh38
7275034 c.692C>A p.P231H Chr.1: 52238109-52238109 on Build GRCh38
3724242 c.500A>G p.Q167R Chr.1: 52237917-52237917 on Build GRCh38
4008789 c.591T>C p.F197F Chr.1: 52238008-52238008 on Build GRCh38
910616 c.532A>G p.M178V Chr.1: 52237949-52237949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561123836 Chr.1: 52238063-52238063 on Build GRCh38
rs772530964 Chr.1: 52238088-52238088 on Build GRCh38
rs944631576 Chr.1: 52237963-52237963 on Build GRCh38
rs1043912983 Chr.1: 52238055-52238055 on Build GRCh38
rs368049077 Chr.1: 52237995-52237995 on Build GRCh38
rs972257899 Chr.1: 52237921-52237921 on Build GRCh38
rs748557900 Chr.1: 52238083-52238083 on Build GRCh38
rs764847826 Chr.1: 52237999-52237999 on Build GRCh38
rs768785177 Chr.1: 52237917-52237917 on Build GRCh38
rs779975324 Chr.1: 52237940-52237940 on Build GRCh38
rs757189596 Chr.1: 52237932-52237932 on Build GRCh38
rs777599141 Chr.1: 52237909-52237909 on Build GRCh38
rs752186028 Chr.1: 52238014-52238014 on Build GRCh38
rs751348948 Chr.1: 52237916-52237916 on Build GRCh38
rs762212848 Chr.1: 52237955-52237955 on Build GRCh38
rs545208610 Chr.1: 52237933-52237933 on Build GRCh38
rs773422957 Chr.1: 52238093-52238093 on Build GRCh38
rs750424414 Chr.1: 52238042-52238042 on Build GRCh38
rs1040303269 Chr.1: 52238010-52238010 on Build GRCh38
rs774861730 Chr.1: 52237950-52237950 on Build GRCh38
rs911512386 Chr.1: 52237998-52237998 on Build GRCh38
rs772481008 Chr.1: 52237956-52237956 on Build GRCh38
rs768997160 Chr.1: 52237946-52237946 on Build GRCh38
rs761059521 Chr.1: 52237996-52237996 on Build GRCh38
rs952904640 Chr.1: 52237953-52237953 on Build GRCh38
rs563681333 Chr.1: 52238033-52238033 on Build GRCh38
rs370074863 Chr.1: 52237913-52237913 on Build GRCh38
rs142888719 Chr.1: 52237930-52237930 on Build GRCh38
rs1000657476 Chr.1: 52238081-52238081 on Build GRCh38
rs144874848 Chr.1: 52237936-52237936 on Build GRCh38
rs182750927 Chr.1: 52238037-52238037 on Build GRCh38
rs370955487 Chr.1: 52237889-52237889 on Build GRCh38
rs553338439 Chr.1: 52237893-52237893 on Build GRCh38
rs781021724 Chr.1: 52238039-52238039 on Build GRCh38
rs755993265 Chr.1: 52238044-52238044 on Build GRCh38
rs762658122 Chr.1: 52238024-52238024 on Build GRCh38
rs769878101 Chr.1: 52237938-52237938 on Build GRCh38
rs769015703 Chr.1: 52238069-52238069 on Build GRCh38
rs965979909 Chr.1: 52238008-52238008 on Build GRCh38
rs528396709 Chr.1: 52238073-52238073 on Build GRCh38
rs749580771 Chr.1: 52237942-52237942 on Build GRCh38
rs749316478 Chr.1: 52238066-52238066 on Build GRCh38
rs759041526 Chr.1: 52237880-52237880 on Build GRCh38
rs138765001 Chr.1: 52238036-52238036 on Build GRCh38
rs904617054 Chr.1: 52238074-52238074 on Build GRCh38
rs753305006 Chr.1: 52237879-52237879 on Build GRCh38
rs1042852074 Chr.1: 52238090-52238090 on Build GRCh38
rs200739181 Chr.1: 52237906-52237906 on Build GRCh38

View Full Product Details