Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 95064947-95065186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG14
Entrez Gene ID
199857
Gene Name
ALG14, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases
CMS15
UniGene
Hs.408927
Gene Chromosome Location
Chr.1: 94981565-95072974 on Build GRCh38
Transcripts
NM_001305242.1 NM_144988.3 NR_131032.1 XM_005270582.3 XM_011540897.2 NR_132786.1

Gene Symbol
LOC101928098
Entrez Gene ID
101928098
Gene Name
uncharacterized LOC101928098
Gene Aliases
-
UniGene
Hs.730090
Gene Chromosome Location
Chr.1: 95061596-95067545 on Build GRCh38
Transcripts
NM_001305242.1 NM_144988.3 NR_131032.1 XM_005270582.3 XM_011540897.2 NR_132786.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6489996 c.171G>C p.G57G Chr.1: 95064983-95064983 on Build GRCh38
7242213 c.159G>A p.L53L Chr.1: 95064995-95064995 on Build GRCh38
3419607 c.166C>T p.L56F Chr.1: 95064988-95064988 on Build GRCh38
4010742 c.141G>A p.G47G Chr.1: 95065013-95065013 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs190745630 Chr.1: 95065099-95065099 on Build GRCh38
rs772833545 Chr.1: 95065016-95065016 on Build GRCh38
rs141065009 Chr.1: 95064983-95064983 on Build GRCh38
rs780277809 Chr.1: 95064975-95064975 on Build GRCh38
rs914294107 Chr.1: 95065091-95065091 on Build GRCh38
rs188453904 Chr.1: 95065059-95065059 on Build GRCh38
rs947130234 Chr.1: 95065096-95065096 on Build GRCh38
rs759429976 Chr.1: 95065062-95065062 on Build GRCh38
rs770491182 Chr.1: 95065121-95065121 on Build GRCh38
rs954876014 Chr.1: 95065013-95065013 on Build GRCh38
rs754951451 Chr.1: 95065113-95065113 on Build GRCh38
rs942762121 Chr.1: 95065133-95065133 on Build GRCh38
rs747589839 Chr.1: 95065003-95065003 on Build GRCh38
rs72720257 Chr.1: 95065114-95065114 on Build GRCh38
rs372806617 Chr.1: 95065066-95065066 on Build GRCh38
rs6674467 Chr.1: 95065153-95065153 on Build GRCh38
rs778566822 Chr.1: 95064991-95064991 on Build GRCh38
rs139758313 Chr.1: 95065014-95065014 on Build GRCh38
rs910413063 Chr.1: 95065119-95065119 on Build GRCh38
rs761928890 Chr.1: 95065069-95065069 on Build GRCh38
rs768248972 Chr.1: 95064989-95064989 on Build GRCh38
rs764375049 Chr.1: 95065064-95065064 on Build GRCh38
rs776264274 Chr.1: 95065060-95065060 on Build GRCh38
rs12751061 Chr.1: 95065035-95065035 on Build GRCh38

View Full Product Details