Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 118395250-118395475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR44
Entrez Gene ID
54521
Gene Name
WD repeat domain 44
Gene Aliases
RAB11BP, RPH11
UniGene
Hs.98510
Gene Chromosome Location
Chr.X: 118345997-118449961 on Build GRCh38
Transcripts
NM_001184965.1 NM_001184966.1 NM_019045.4 XM_011531353.2 XM_017029600.1

SNP Detail

SNP ID Chromosome Location
rs747480816 Chr.X: 118395388-118395389 on Build GRCh38
rs778937011 Chr.X: 118395418-118395418 on Build GRCh38
rs371338318 Chr.X: 118395333-118395333 on Build GRCh38
rs777841339 Chr.X: 118395357-118395357 on Build GRCh38
rs768523158 Chr.X: 118395379-118395379 on Build GRCh38
rs765761577 Chr.X: 118395319-118395319 on Build GRCh38
rs754181976 Chr.X: 118395301-118395301 on Build GRCh38
rs755515843 Chr.X: 118395317-118395317 on Build GRCh38
rs774169231 Chr.X: 118395380-118395380 on Build GRCh38
rs766791109 Chr.X: 118395293-118395293 on Build GRCh38
rs758417491 Chr.X: 118395349-118395349 on Build GRCh38
rs749127294 Chr.X: 118395378-118395378 on Build GRCh38
rs760374300 Chr.X: 118395279-118395279 on Build GRCh38
rs974161489 Chr.X: 118395407-118395407 on Build GRCh38
rs985650508 Chr.X: 118395424-118395424 on Build GRCh38
rs757537705 Chr.X: 118395368-118395368 on Build GRCh38
rs920017010 Chr.X: 118395415-118395415 on Build GRCh38
rs1015559175 Chr.X: 118395276-118395276 on Build GRCh38
rs780037335 Chr.X: 118395373-118395373 on Build GRCh38

View Full Product Details