Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 110147865-110148138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPHP1
Entrez Gene ID
4867
Gene Name
nephrocystin 1
Gene Aliases
JBTS4, NPH1, SLSN1
UniGene
Hs.280388
Gene Chromosome Location
Chr.2: 110123336-110205062 on Build GRCh38
Transcripts
NM_000272.3 NM_001128178.1 NM_001128179.1 NM_207181.2 XM_005263676.1 XM_005263677.1 XM_005263678.2 XM_005263679.1 XM_006712551.1 XM_006712552.2 XM_011511244.1 XM_017004218.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6308897 c.1420A>G p.I474V Chr.2: 110147933-110147933 on Build GRCh38
4769956 c.1358G>A p.G453D Chr.2: 110147995-110147995 on Build GRCh38
1217696 c.1372A>G p.R458G Chr.2: 110147981-110147981 on Build GRCh38
340612 c.1428_1429TA>TC p.Y476>? Chr.2: 110147924-110147925 on Build GRCh38
4958335 c.1398T>C p.L466L Chr.2: 110147955-110147955 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768169550 Chr.2: 110147913-110147913 on Build GRCh38
rs752979255 Chr.2: 110147979-110147979 on Build GRCh38
rs140979636 Chr.2: 110148019-110148019 on Build GRCh38
rs577560349 Chr.2: 110147933-110147933 on Build GRCh38
rs375907280 Chr.2: 110148020-110148020 on Build GRCh38
rs745657805 Chr.2: 110147894-110147894 on Build GRCh38
rs777196172 Chr.2: 110148078-110148078 on Build GRCh38
rs778443933 Chr.2: 110148053-110148053 on Build GRCh38
rs763221050 Chr.2: 110148003-110148003 on Build GRCh38
rs755943785 Chr.2: 110148023-110148023 on Build GRCh38
rs768263728 Chr.2: 110148049-110148049 on Build GRCh38
rs747679629 Chr.2: 110148057-110148057 on Build GRCh38
rs759936705 Chr.2: 110147954-110147954 on Build GRCh38
rs748969566 Chr.2: 110148034-110148034 on Build GRCh38
rs748875496 Chr.2: 110147908-110147908 on Build GRCh38
rs1509417 Chr.2: 110148087-110148087 on Build GRCh38
rs764303266 Chr.2: 110148012-110148012 on Build GRCh38
rs765450176 Chr.2: 110147956-110147956 on Build GRCh38
rs771404672 Chr.2: 110148058-110148058 on Build GRCh38
rs978534998 Chr.2: 110147998-110147998 on Build GRCh38
rs1030337277 Chr.2: 110147965-110147965 on Build GRCh38
rs945386048 Chr.2: 110148086-110148086 on Build GRCh38
rs13414551 Chr.2: 110147907-110147907 on Build GRCh38
rs771446005 Chr.2: 110147926-110147926 on Build GRCh38
rs745877439 Chr.2: 110148008-110148008 on Build GRCh38
rs990050723 Chr.2: 110148046-110148046 on Build GRCh38
rs373749095 Chr.2: 110148063-110148063 on Build GRCh38
rs773810185 Chr.2: 110147920-110147920 on Build GRCh38
rs761209487 Chr.2: 110147923-110147923 on Build GRCh38
rs913978455 Chr.2: 110148054-110148054 on Build GRCh38
rs974811294 Chr.2: 110147921-110147921 on Build GRCh38
rs954734199 Chr.2: 110147984-110147984 on Build GRCh38
rs779767314 Chr.2: 110148032-110148032 on Build GRCh38
rs958211666 Chr.2: 110147951-110147951 on Build GRCh38
rs769658332 Chr.2: 110148034-110148034 on Build GRCh38
rs35932880 Chr.2: 110148107-110148108 on Build GRCh38

View Full Product Details