Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202265821-202266076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOP58
Entrez Gene ID
51602
Gene Name
NOP58 ribonucleoprotein
Gene Aliases
HSPC120, NOP5, NOP5/NOP58
UniGene
Hs.471104
Gene Chromosome Location
Chr.2: 202265716-202303661 on Build GRCh38
Transcripts
NM_015934.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
304863 c.3G>A p.M1I Chr.2: 202265944-202265944 on Build GRCh38
359621 c.34G>T p.A12S Chr.2: 202265975-202265975 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs534007673 Chr.2: 202265872-202265872 on Build GRCh38
rs753027351 Chr.2: 202265989-202265990 on Build GRCh38
rs750823552 Chr.2: 202265869-202265869 on Build GRCh38
rs749940707 Chr.2: 202265933-202265933 on Build GRCh38
rs915768304 Chr.2: 202266049-202266049 on Build GRCh38
rs374814936 Chr.2: 202265915-202265915 on Build GRCh38
rs761465787 Chr.2: 202266029-202266029 on Build GRCh38
rs779991794 Chr.2: 202265980-202265980 on Build GRCh38
rs568991461 Chr.2: 202265843-202265843 on Build GRCh38
rs371185656 Chr.2: 202265931-202265931 on Build GRCh38
rs375794713 Chr.2: 202265932-202265932 on Build GRCh38
rs535027378 Chr.2: 202265873-202265873 on Build GRCh38
rs768714477 Chr.2: 202265895-202265895 on Build GRCh38
rs1029714759 Chr.2: 202265896-202265896 on Build GRCh38
rs772249475 Chr.2: 202265903-202265903 on Build GRCh38
rs754866824 Chr.2: 202265988-202265988 on Build GRCh38
rs200002128 Chr.2: 202266009-202266009 on Build GRCh38
rs775736134 Chr.2: 202266013-202266013 on Build GRCh38
rs369397560 Chr.2: 202265891-202265891 on Build GRCh38
rs142142955 Chr.2: 202266036-202266036 on Build GRCh38
rs370710550 Chr.2: 202265986-202265986 on Build GRCh38
rs376417516 Chr.2: 202265897-202265897 on Build GRCh38
rs762189050 Chr.2: 202266010-202266010 on Build GRCh38
rs748170963 Chr.2: 202265900-202265900 on Build GRCh38
rs748343719 Chr.2: 202265989-202265989 on Build GRCh38
rs760228758 Chr.2: 202266037-202266037 on Build GRCh38
rs199514920 Chr.2: 202266000-202266000 on Build GRCh38
rs763626549 Chr.2: 202265918-202265918 on Build GRCh38
rs771593608 Chr.2: 202265913-202265913 on Build GRCh38
rs747104333 Chr.2: 202265996-202265996 on Build GRCh38
rs777314567 Chr.2: 202265998-202265998 on Build GRCh38
rs949365523 Chr.2: 202265865-202265865 on Build GRCh38
rs756006471 Chr.2: 202265948-202265948 on Build GRCh38
rs1015032964 Chr.2: 202265966-202265966 on Build GRCh38
rs148667174 Chr.2: 202265909-202265909 on Build GRCh38
rs374760196 Chr.2: 202266006-202266006 on Build GRCh38
rs578049121 Chr.2: 202265908-202265908 on Build GRCh38
rs554926408 Chr.2: 202265848-202265848 on Build GRCh38
rs777956207 Chr.2: 202265992-202265992 on Build GRCh38
rs753609540 Chr.2: 202265985-202265985 on Build GRCh38
rs373399368 Chr.2: 202265892-202265892 on Build GRCh38
rs771540480 Chr.2: 202265997-202265997 on Build GRCh38
rs536512322 Chr.2: 202265844-202265844 on Build GRCh38
rs377762537 Chr.2: 202265916-202265916 on Build GRCh38
rs767189812 Chr.2: 202266031-202266031 on Build GRCh38
rs987167021 Chr.2: 202265934-202265934 on Build GRCh38
rs756418794 Chr.2: 202266014-202266015 on Build GRCh38
rs895196230 Chr.2: 202265847-202265847 on Build GRCh38
rs764528691 Chr.2: 202266022-202266022 on Build GRCh38
rs563869366 Chr.2: 202265911-202265911 on Build GRCh38
rs867641451 Chr.2: 202265975-202265975 on Build GRCh38
rs751296013 Chr.2: 202265928-202265928 on Build GRCh38
rs902780739 Chr.2: 202265870-202265870 on Build GRCh38

View Full Product Details