Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 173233344-173233573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKX2-5
Entrez Gene ID
1482
Gene Name
NK2 homeobox 5
Gene Aliases
CHNG5, CSX, CSX1, HLHS2, NKX2.5, NKX2E, NKX4-1, VSD3
UniGene
Hs.54473
Gene Chromosome Location
Chr.5: 173232104-173235312 on Build GRCh38
Transcripts
NM_001166175.1 NM_001166176.1 NM_004387.3 XM_017009071.1

SNP Detail

SNP ID Chromosome Location
rs534163213 Chr.5: 173233501-173233501 on Build GRCh38
rs55880439 Chr.5: 173233524-173233524 on Build GRCh38
rs763540247 Chr.5: 173233522-173233524 on Build GRCh38
rs766767133 Chr.5: 173233513-173233532 on Build GRCh38
rs768660312 Chr.5: 173233525-173233525 on Build GRCh38
rs770265328 Chr.5: 173233496-173233496 on Build GRCh38
rs567753920 Chr.5: 173233515-173233515 on Build GRCh38
rs564682054 Chr.5: 173233516-173233517 on Build GRCh38
rs780682576 Chr.5: 173233524-173233524 on Build GRCh38
rs950541477 Chr.5: 173233545-173233545 on Build GRCh38
rs1013341984 Chr.5: 173233394-173233394 on Build GRCh38
rs776440889 Chr.5: 173233505-173233505 on Build GRCh38
rs777437672 Chr.5: 173233516-173233520 on Build GRCh38
rs751058601 Chr.5: 173233521-173233522 on Build GRCh38
rs200039950 Chr.5: 173233371-173233371 on Build GRCh38
rs754306771 Chr.5: 173233521-173233526 on Build GRCh38
rs558929892 Chr.5: 173233526-173233526 on Build GRCh38
rs757855884 Chr.5: 173233515-173233532 on Build GRCh38
rs767630916 Chr.5: 173233520-173233524 on Build GRCh38
rs770760092 Chr.5: 173233451-173233451 on Build GRCh38
rs756713323 Chr.5: 173233513-173233524 on Build GRCh38
rs768788951 Chr.5: 173233518-173233524 on Build GRCh38
rs957107634 Chr.5: 173233428-173233428 on Build GRCh38
rs200467566 Chr.5: 173233512-173233520 on Build GRCh38
rs762182076 Chr.5: 173233519-173233524 on Build GRCh38
rs865916303 Chr.5: 173233518-173233518 on Build GRCh38
rs556541788 Chr.5: 173233517-173233517 on Build GRCh38
rs551687864 Chr.5: 173233409-173233409 on Build GRCh38
rs3095871 Chr.5: 173233520-173233520 on Build GRCh38
rs780144291 Chr.5: 173233525-173233526 on Build GRCh38
rs571495972 Chr.5: 173233463-173233463 on Build GRCh38
rs747848277 Chr.5: 173233411-173233411 on Build GRCh38
rs868037308 Chr.5: 173233522-173233522 on Build GRCh38
rs752255164 Chr.5: 173233513-173233520 on Build GRCh38
rs766931455 Chr.5: 173233520-173233521 on Build GRCh38
rs774638512 Chr.5: 173233531-173233532 on Build GRCh38
rs747301324 Chr.5: 173233499-173233499 on Build GRCh38
rs547572698 Chr.5: 173233513-173233513 on Build GRCh38
rs770235503 Chr.5: 173233530-173233530 on Build GRCh38
rs745616594 Chr.5: 173233517-173233520 on Build GRCh38
rs749179469 Chr.5: 173233525-173233525 on Build GRCh38
rs774726585 Chr.5: 173233519-173233520 on Build GRCh38
rs56265301 Chr.5: 173233532-173233532 on Build GRCh38
rs766916370 Chr.5: 173233546-173233546 on Build GRCh38
rs759462937 Chr.5: 173233504-173233505 on Build GRCh38
rs375676786 Chr.5: 173233540-173233540 on Build GRCh38
rs763099269 Chr.5: 173233506-173233506 on Build GRCh38
rs746600877 Chr.5: 173233516-173233532 on Build GRCh38
rs536213285 Chr.5: 173233516-173233516 on Build GRCh38
rs762333634 Chr.5: 173233528-173233528 on Build GRCh38
rs760926110 Chr.5: 173233520-173233532 on Build GRCh38
rs371347562 Chr.5: 173233520-173233520 on Build GRCh38
rs764429827 Chr.5: 173233505-173233506 on Build GRCh38
rs960618881 Chr.5: 173233372-173233373 on Build GRCh38
rs550291085 Chr.5: 173233523-173233524 on Build GRCh38
rs775239026 Chr.5: 173233517-173233532 on Build GRCh38
rs757060818 Chr.5: 173233515-173233516 on Build GRCh38
rs757911285 Chr.5: 173233521-173233527 on Build GRCh38
rs745694632 Chr.5: 173233524-173233525 on Build GRCh38
rs749406949 Chr.5: 173233518-173233520 on Build GRCh38
rs768700738 Chr.5: 173233530-173233532 on Build GRCh38

View Full Product Details