Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 62802360-62802627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNH5
Entrez Gene ID
27133
Gene Name
potassium voltage-gated channel subfamily H member 5
Gene Aliases
EAG2, H-EAG2, Kv10.2, hEAG2
UniGene
Hs.27043
Gene Chromosome Location
Chr.14: 62706573-63045238 on Build GRCh38
Transcripts
NM_139318.4 NM_172375.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5441577 c.1645C>G p.P549A Chr.14: 62802506-62802506 on Build GRCh38
1255307 c.1507G>A p.A503T Chr.14: 62802470-62802470 on Build GRCh38
5441578 c.1471C>G p.P491A Chr.14: 62802506-62802506 on Build GRCh38
4051645 c.1425A>G p.R475R Chr.14: 62802552-62802552 on Build GRCh38
5740117 c.1726C>A p.L576I Chr.14: 62802425-62802425 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751812835 Chr.14: 62802526-62802527 on Build GRCh38
rs770496210 Chr.14: 62802486-62802486 on Build GRCh38
rs899330305 Chr.14: 62802473-62802473 on Build GRCh38
rs756953345 Chr.14: 62802600-62802600 on Build GRCh38
rs777712735 Chr.14: 62802597-62802597 on Build GRCh38
rs769180286 Chr.14: 62802513-62802513 on Build GRCh38
rs371585820 Chr.14: 62802557-62802557 on Build GRCh38
rs267604014 Chr.14: 62802560-62802560 on Build GRCh38
rs150842380 Chr.14: 62802485-62802485 on Build GRCh38
rs868309881 Chr.14: 62802497-62802497 on Build GRCh38
rs762196737 Chr.14: 62802526-62802526 on Build GRCh38
rs745422946 Chr.14: 62802512-62802512 on Build GRCh38
rs114074278 Chr.14: 62802471-62802471 on Build GRCh38
rs115435938 Chr.14: 62802531-62802531 on Build GRCh38
rs766461972 Chr.14: 62802439-62802439 on Build GRCh38
rs1002445202 Chr.14: 62802448-62802448 on Build GRCh38
rs754939357 Chr.14: 62802447-62802447 on Build GRCh38
rs766912404 Chr.14: 62802431-62802432 on Build GRCh38
rs922451620 Chr.14: 62802565-62802565 on Build GRCh38
rs149561461 Chr.14: 62802572-62802572 on Build GRCh38
rs766654524 Chr.14: 62802555-62802555 on Build GRCh38
rs755065139 Chr.14: 62802562-62802562 on Build GRCh38
rs751432846 Chr.14: 62802446-62802446 on Build GRCh38
rs201473849 Chr.14: 62802399-62802399 on Build GRCh38
rs747957418 Chr.14: 62802459-62802459 on Build GRCh38
rs760705689 Chr.14: 62802419-62802419 on Build GRCh38
rs943373012 Chr.14: 62802603-62802603 on Build GRCh38
rs778225685 Chr.14: 62802496-62802496 on Build GRCh38
rs988565496 Chr.14: 62802539-62802539 on Build GRCh38
rs777044154 Chr.14: 62802525-62802525 on Build GRCh38
rs763168479 Chr.14: 62802535-62802535 on Build GRCh38
rs755893047 Chr.14: 62802590-62802590 on Build GRCh38
rs781050144 Chr.14: 62802457-62802457 on Build GRCh38
rs748900455 Chr.14: 62802601-62802601 on Build GRCh38
rs374752391 Chr.14: 62802432-62802432 on Build GRCh38
rs12588427 Chr.14: 62802599-62802599 on Build GRCh38
rs543742412 Chr.14: 62802527-62802527 on Build GRCh38

View Full Product Details