Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 22946486-22946753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAUS4
Entrez Gene ID
54930
Gene Name
HAUS augmin like complex subunit 4
Gene Aliases
C14orf94
UniGene
Hs.442782
Gene Chromosome Location
Chr.14: 22946228-22957142 on Build GRCh38
Transcripts
NM_001166269.1 NM_001166270.1 NM_017815.2 NR_110002.1

Gene Symbol
LOC101926933
Entrez Gene ID
101926933
Gene Name
uncharacterized LOC101926933
Gene Aliases
-
UniGene
Hs.727321
Gene Chromosome Location
Chr.14: 22929609-22954692 on Build GRCh38
Transcripts
NM_001166269.1 NM_001166270.1 NM_017815.2 NR_110002.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5053862 c.1055G>A p.R352Q Chr.14: 22946562-22946562 on Build GRCh38
2030724 c.934C>A p.L312I Chr.14: 22946683-22946683 on Build GRCh38
1707163 c.1025C>T p.T342I Chr.14: 22946592-22946592 on Build GRCh38
7116577 c.994G>A p.E332K Chr.14: 22946623-22946623 on Build GRCh38
4049929 c.989T>C p.L330P Chr.14: 22946628-22946628 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754140448 Chr.14: 22946605-22946605 on Build GRCh38
rs771309994 Chr.14: 22946587-22946587 on Build GRCh38
rs200142609 Chr.14: 22946724-22946724 on Build GRCh38
rs759880389 Chr.14: 22946689-22946689 on Build GRCh38
rs1051130033 Chr.14: 22946620-22946620 on Build GRCh38
rs757985683 Chr.14: 22946590-22946590 on Build GRCh38
rs747678993 Chr.14: 22946562-22946562 on Build GRCh38
rs754797272 Chr.14: 22946625-22946625 on Build GRCh38
rs554534170 Chr.14: 22946536-22946536 on Build GRCh38
rs772427258 Chr.14: 22946698-22946698 on Build GRCh38
rs896266738 Chr.14: 22946716-22946716 on Build GRCh38
rs769219428 Chr.14: 22946648-22946648 on Build GRCh38
rs781048258 Chr.14: 22946634-22946634 on Build GRCh38
rs767978772 Chr.14: 22946593-22946593 on Build GRCh38
rs781151137 Chr.14: 22946555-22946555 on Build GRCh38
rs376145555 Chr.14: 22946713-22946713 on Build GRCh38
rs879078355 Chr.14: 22946585-22946585 on Build GRCh38
rs773990476 Chr.14: 22946680-22946680 on Build GRCh38
rs756218278 Chr.14: 22946508-22946508 on Build GRCh38
rs778202530 Chr.14: 22946510-22946510 on Build GRCh38
rs199534311 Chr.14: 22946520-22946520 on Build GRCh38
rs761199996 Chr.14: 22946704-22946704 on Build GRCh38
rs972752008 Chr.14: 22946549-22946549 on Build GRCh38
rs148445489 Chr.14: 22946563-22946563 on Build GRCh38
rs775861306 Chr.14: 22946703-22946703 on Build GRCh38
rs370366652 Chr.14: 22946712-22946712 on Build GRCh38
rs777217427 Chr.14: 22946653-22946653 on Build GRCh38
rs754742744 Chr.14: 22946726-22946726 on Build GRCh38
rs540346373 Chr.14: 22946612-22946612 on Build GRCh38
rs377667124 Chr.14: 22946611-22946611 on Build GRCh38
rs759916524 Chr.14: 22946591-22946591 on Build GRCh38
rs753238249 Chr.14: 22946594-22946594 on Build GRCh38
rs1033124759 Chr.14: 22946663-22946663 on Build GRCh38
rs762738399 Chr.14: 22946708-22946711 on Build GRCh38
rs1052607821 Chr.14: 22946716-22946716 on Build GRCh38
rs760687797 Chr.14: 22946600-22946600 on Build GRCh38
rs777419704 Chr.14: 22946677-22946677 on Build GRCh38
rs772856507 Chr.14: 22946569-22946569 on Build GRCh38
rs762553157 Chr.14: 22946581-22946581 on Build GRCh38
rs75188145 Chr.14: 22946524-22946524 on Build GRCh38
rs748775496 Chr.14: 22946675-22946675 on Build GRCh38
rs746961651 Chr.14: 22946530-22946530 on Build GRCh38
rs767305140 Chr.14: 22946729-22946729 on Build GRCh38
rs918717377 Chr.14: 22946589-22946589 on Build GRCh38
rs201911072 Chr.14: 22946529-22946529 on Build GRCh38
rs748051551 Chr.14: 22946642-22946642 on Build GRCh38
rs761951329 Chr.14: 22946718-22946718 on Build GRCh38
rs764199121 Chr.14: 22946604-22946604 on Build GRCh38
rs200660231 Chr.14: 22946641-22946641 on Build GRCh38
rs765620884 Chr.14: 22946719-22946719 on Build GRCh38

View Full Product Details