Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18263971-18264226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIEF2
Entrez Gene ID
125170
Gene Name
mitochondrial elongation factor 2
Gene Aliases
MID49, SMCR7
UniGene
Hs.655555
Gene Chromosome Location
Chr.17: 18260534-18265790 on Build GRCh38
Transcripts
NM_001144900.1 NM_139162.3 NM_148886.1 XM_017024190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6221571 c.580G>A p.V194M Chr.17: 18263979-18263979 on Build GRCh38
5523875 c.659C>T p.P220L Chr.17: 18264058-18264058 on Build GRCh38
704814 c.696C>A p.F232L Chr.17: 18264095-18264095 on Build GRCh38
3691405 c.658C>A p.P220T Chr.17: 18264057-18264057 on Build GRCh38
7190276 c.744C>T p.Y248Y Chr.17: 18264143-18264143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201138563 Chr.17: 18264195-18264195 on Build GRCh38
rs768197641 Chr.17: 18264027-18264027 on Build GRCh38
rs374077306 Chr.17: 18264131-18264131 on Build GRCh38
rs768055946 Chr.17: 18264057-18264057 on Build GRCh38
rs199876054 Chr.17: 18264082-18264082 on Build GRCh38
rs372507546 Chr.17: 18264124-18264124 on Build GRCh38
rs760972949 Chr.17: 18264134-18264134 on Build GRCh38
rs777504562 Chr.17: 18264174-18264174 on Build GRCh38
rs570365504 Chr.17: 18264031-18264031 on Build GRCh38
rs756349402 Chr.17: 18263991-18263991 on Build GRCh38
rs772626135 Chr.17: 18264189-18264189 on Build GRCh38
rs769549583 Chr.17: 18264112-18264112 on Build GRCh38
rs200747752 Chr.17: 18263989-18263989 on Build GRCh38
rs550456364 Chr.17: 18263993-18263993 on Build GRCh38
rs745842933 Chr.17: 18264008-18264008 on Build GRCh38
rs2605143 Chr.17: 18264191-18264191 on Build GRCh38
rs770428294 Chr.17: 18264176-18264176 on Build GRCh38
rs112403499 Chr.17: 18264175-18264175 on Build GRCh38
rs781021060 Chr.17: 18264097-18264097 on Build GRCh38
rs749138167 Chr.17: 18264049-18264049 on Build GRCh38
rs772818705 Chr.17: 18264036-18264036 on Build GRCh38
rs574879368 Chr.17: 18264156-18264156 on Build GRCh38
rs753923218 Chr.17: 18263994-18263994 on Build GRCh38
rs761038231 Chr.17: 18264061-18264061 on Build GRCh38
rs747944417 Chr.17: 18264164-18264164 on Build GRCh38
rs376412729 Chr.17: 18264153-18264153 on Build GRCh38
rs750473513 Chr.17: 18264152-18264152 on Build GRCh38
rs1017502182 Chr.17: 18264120-18264120 on Build GRCh38
rs373841311 Chr.17: 18264182-18264182 on Build GRCh38
rs765458605 Chr.17: 18264206-18264206 on Build GRCh38
rs758662136 Chr.17: 18264008-18264028 on Build GRCh38
rs757436589 Chr.17: 18264075-18264075 on Build GRCh38
rs746815006 Chr.17: 18264017-18264017 on Build GRCh38
rs3889402 Chr.17: 18264083-18264083 on Build GRCh38
rs546097250 Chr.17: 18264205-18264205 on Build GRCh38
rs754752116 Chr.17: 18264092-18264092 on Build GRCh38
rs765436333 Chr.17: 18264150-18264150 on Build GRCh38
rs202127880 Chr.17: 18264086-18264086 on Build GRCh38
rs755897737 Chr.17: 18264171-18264171 on Build GRCh38
rs772916959 Chr.17: 18264123-18264123 on Build GRCh38
rs772020431 Chr.17: 18264011-18264011 on Build GRCh38
rs369266297 Chr.17: 18264072-18264072 on Build GRCh38
rs1011779554 Chr.17: 18264197-18264197 on Build GRCh38
rs375214936 Chr.17: 18264186-18264186 on Build GRCh38
rs897104283 Chr.17: 18264050-18264050 on Build GRCh38
rs376909447 Chr.17: 18264060-18264060 on Build GRCh38
rs769390003 Chr.17: 18264035-18264035 on Build GRCh38
rs370780579 Chr.17: 18264181-18264181 on Build GRCh38
rs373199441 Chr.17: 18264155-18264155 on Build GRCh38
rs776955546 Chr.17: 18264135-18264135 on Build GRCh38
rs747845745 Chr.17: 18264102-18264102 on Build GRCh38
rs778850892 Chr.17: 18263997-18263997 on Build GRCh38
rs372335713 Chr.17: 18264032-18264032 on Build GRCh38
rs754014042 Chr.17: 18264074-18264074 on Build GRCh38
rs190023411 Chr.17: 18264154-18264154 on Build GRCh38
rs372764224 Chr.17: 18264076-18264076 on Build GRCh38
rs554628770 Chr.17: 18264147-18264147 on Build GRCh38

View Full Product Details