Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 128669350-128669623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM132C
Entrez Gene ID
92293
Gene Name
transmembrane protein 132C
Gene Aliases
PPP1R152
UniGene
Hs.49599
Gene Chromosome Location
Chr.12: 128267170-128707915 on Build GRCh38
Transcripts
NM_001136103.2 XM_011538998.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2227712 c.1340G>A p.G447E Chr.12: 128669451-128669451 on Build GRCh38
7394115 c.200C>T p.A67V Chr.12: 128669463-128669463 on Build GRCh38
4451705 c.1364A>C p.K455T Chr.12: 128669475-128669475 on Build GRCh38
936880 c.241G>A p.V81M Chr.12: 128669504-128669504 on Build GRCh38
4465220 c.1371C>T p.V457V Chr.12: 128669482-128669482 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113236544 Chr.12: 128669412-128669412 on Build GRCh38
rs1012511257 Chr.12: 128669560-128669560 on Build GRCh38
rs896865433 Chr.12: 128669519-128669519 on Build GRCh38
rs1040757819 Chr.12: 128669517-128669517 on Build GRCh38
rs779054139 Chr.12: 128669420-128669420 on Build GRCh38
rs540357593 Chr.12: 128669395-128669395 on Build GRCh38
rs769557725 Chr.12: 128669475-128669475 on Build GRCh38
rs778253530 Chr.12: 128669468-128669468 on Build GRCh38
rs150073399 Chr.12: 128669592-128669592 on Build GRCh38
rs753102735 Chr.12: 128669434-128669434 on Build GRCh38
rs906087987 Chr.12: 128669397-128669397 on Build GRCh38
rs768015459 Chr.12: 128669503-128669503 on Build GRCh38
rs953383821 Chr.12: 128669548-128669548 on Build GRCh38
rs943814459 Chr.12: 128669511-128669511 on Build GRCh38
rs145341430 Chr.12: 128669598-128669598 on Build GRCh38
rs139397925 Chr.12: 128669524-128669524 on Build GRCh38
rs4272850 Chr.12: 128669441-128669441 on Build GRCh38
rs561752570 Chr.12: 128669581-128669581 on Build GRCh38
rs377355130 Chr.12: 128669540-128669540 on Build GRCh38
rs568693738 Chr.12: 128669545-128669545 on Build GRCh38
rs150833144 Chr.12: 128669496-128669496 on Build GRCh38
rs201192757 Chr.12: 128669498-128669498 on Build GRCh38
rs202002692 Chr.12: 128669593-128669593 on Build GRCh38
rs373672782 Chr.12: 128669381-128669381 on Build GRCh38
rs1032979036 Chr.12: 128669437-128669437 on Build GRCh38
rs878980497 Chr.12: 128669379-128669379 on Build GRCh38
rs984505659 Chr.12: 128669549-128669549 on Build GRCh38
rs545813970 Chr.12: 128669473-128669473 on Build GRCh38
rs773936735 Chr.12: 128669504-128669504 on Build GRCh38
rs976296397 Chr.12: 128669528-128669528 on Build GRCh38
rs542076193 Chr.12: 128669525-128669525 on Build GRCh38
rs748979165 Chr.12: 128669502-128669502 on Build GRCh38
rs908877791 Chr.12: 128669580-128669580 on Build GRCh38
rs760516820 Chr.12: 128669552-128669552 on Build GRCh38

View Full Product Details