Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42161593-42161866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS39
Entrez Gene ID
23339
Gene Name
VPS39, HOPS complex subunit
Gene Aliases
TLP, VAM6, hVam6p
UniGene
Hs.88025
Gene Chromosome Location
Chr.15: 42158701-42208331 on Build GRCh38
Transcripts
NM_001301138.1 NM_015289.3 XM_011521403.2 XM_011521404.2 XM_017022036.1 XM_017022037.1 XM_017022038.1 XM_017022039.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6311156 c.2463C>A p.V821V Chr.15: 42161771-42161771 on Build GRCh38
4860626 c.2566A>G p.K856E Chr.15: 42161701-42161701 on Build GRCh38
6311157 c.2496C>A p.V832V Chr.15: 42161771-42161771 on Build GRCh38
961677 c.2522G>A p.C841Y Chr.15: 42161712-42161712 on Build GRCh38
1188982 c.2465A>G p.Q822R Chr.15: 42161769-42161769 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767315369 Chr.15: 42161664-42161664 on Build GRCh38
rs1056297739 Chr.15: 42161759-42161759 on Build GRCh38
rs16972779 Chr.15: 42161765-42161765 on Build GRCh38
rs1037191697 Chr.15: 42161661-42161661 on Build GRCh38
rs764937033 Chr.15: 42161708-42161708 on Build GRCh38
rs757004706 Chr.15: 42161699-42161699 on Build GRCh38
rs764057321 Chr.15: 42161822-42161822 on Build GRCh38
rs747516002 Chr.15: 42161619-42161619 on Build GRCh38
rs148082104 Chr.15: 42161654-42161654 on Build GRCh38
rs747523223 Chr.15: 42161758-42161758 on Build GRCh38
rs762035497 Chr.15: 42161626-42161626 on Build GRCh38
rs763153963 Chr.15: 42161720-42161722 on Build GRCh38
rs769935979 Chr.15: 42161630-42161630 on Build GRCh38
rs939193915 Chr.15: 42161747-42161747 on Build GRCh38
rs760648260 Chr.15: 42161667-42161667 on Build GRCh38
rs185398866 Chr.15: 42161697-42161697 on Build GRCh38
rs780459881 Chr.15: 42161753-42161753 on Build GRCh38
rs755452044 Chr.15: 42161761-42161761 on Build GRCh38
rs749522618 Chr.15: 42161795-42161795 on Build GRCh38
rs187923034 Chr.15: 42161810-42161810 on Build GRCh38
rs376424375 Chr.15: 42161787-42161787 on Build GRCh38
rs774161464 Chr.15: 42161657-42161657 on Build GRCh38
rs544380776 Chr.15: 42161825-42161825 on Build GRCh38
rs535901844 Chr.15: 42161620-42161620 on Build GRCh38
rs750086356 Chr.15: 42161760-42161760 on Build GRCh38
rs199811682 Chr.15: 42161771-42161771 on Build GRCh38
rs373700662 Chr.15: 42161819-42161819 on Build GRCh38
rs771821713 Chr.15: 42161803-42161803 on Build GRCh38
rs750080634 Chr.15: 42161712-42161712 on Build GRCh38
rs765313612 Chr.15: 42161661-42161676 on Build GRCh38
rs552920541 Chr.15: 42161666-42161666 on Build GRCh38
rs1004672123 Chr.15: 42161639-42161639 on Build GRCh38
rs758727646 Chr.15: 42161721-42161721 on Build GRCh38
rs748186599 Chr.15: 42161769-42161769 on Build GRCh38
rs759275978 Chr.15: 42161663-42161663 on Build GRCh38
rs773338079 Chr.15: 42161692-42161694 on Build GRCh38
rs763525614 Chr.15: 42161692-42161692 on Build GRCh38

View Full Product Details