Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48195134-48195393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSX5
Entrez Gene ID
6758
Gene Name
SSX family member 5
Gene Aliases
-
UniGene
Hs.166198
Gene Chromosome Location
Chr.X: 48177914-48196798 on Build GRCh38
Transcripts
NM_021015.3 NM_175723.1 XM_011543949.2 XM_017029758.1

SNP Detail

SNP ID Chromosome Location
rs782142959 Chr.X: 48195366-48195366 on Build GRCh38
rs782470132 Chr.X: 48195167-48195167 on Build GRCh38
rs375880452 Chr.X: 48195277-48195277 on Build GRCh38
rs782658686 Chr.X: 48195333-48195333 on Build GRCh38
rs781796534 Chr.X: 48195323-48195323 on Build GRCh38
rs782051544 Chr.X: 48195240-48195240 on Build GRCh38
rs782473942 Chr.X: 48195316-48195316 on Build GRCh38
rs781868079 Chr.X: 48195159-48195159 on Build GRCh38
rs782771404 Chr.X: 48195305-48195305 on Build GRCh38
rs4824675 Chr.X: 48195304-48195304 on Build GRCh38
rs781875347 Chr.X: 48195309-48195309 on Build GRCh38
rs781858128 Chr.X: 48195322-48195322 on Build GRCh38
rs782454324 Chr.X: 48195326-48195326 on Build GRCh38
rs782426008 Chr.X: 48195367-48195367 on Build GRCh38
rs782356547 Chr.X: 48195365-48195365 on Build GRCh38
rs782278926 Chr.X: 48195194-48195194 on Build GRCh38
rs370126682 Chr.X: 48195267-48195267 on Build GRCh38
rs781787831 Chr.X: 48195170-48195170 on Build GRCh38
rs199714668 Chr.X: 48195275-48195275 on Build GRCh38
rs111262473 Chr.X: 48195353-48195353 on Build GRCh38
rs782229511 Chr.X: 48195352-48195352 on Build GRCh38
rs781971787 Chr.X: 48195297-48195297 on Build GRCh38
rs782709899 Chr.X: 48195242-48195242 on Build GRCh38
rs373609454 Chr.X: 48195334-48195334 on Build GRCh38
rs370150651 Chr.X: 48195280-48195280 on Build GRCh38
rs147035705 Chr.X: 48195346-48195346 on Build GRCh38
rs782525924 Chr.X: 48195177-48195177 on Build GRCh38
rs377334775 Chr.X: 48195180-48195180 on Build GRCh38
rs147678026 Chr.X: 48195347-48195347 on Build GRCh38
rs782818998 Chr.X: 48195256-48195256 on Build GRCh38
rs782814183 Chr.X: 48195158-48195158 on Build GRCh38
rs149723790 Chr.X: 48195299-48195299 on Build GRCh38
rs782510142 Chr.X: 48195181-48195181 on Build GRCh38
rs372799428 Chr.X: 48195276-48195276 on Build GRCh38
rs782341619 Chr.X: 48195295-48195295 on Build GRCh38
rs377053689 Chr.X: 48195335-48195335 on Build GRCh38
rs199509152 Chr.X: 48195362-48195362 on Build GRCh38
rs782631773 Chr.X: 48195319-48195319 on Build GRCh38
rs782366639 Chr.X: 48195257-48195257 on Build GRCh38
rs782580397 Chr.X: 48195263-48195263 on Build GRCh38
rs782650039 Chr.X: 48195172-48195172 on Build GRCh38

View Full Product Details