Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 109865117-109865378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XPNPEP1
Entrez Gene ID
7511
Gene Name
X-prolyl aminopeptidase 1
Gene Aliases
APP1, SAMP, XPNPEP, XPNPEPL, XPNPEPL1
UniGene
Hs.390623
Gene Chromosome Location
Chr.10: 109864766-109923553 on Build GRCh38
Transcripts
NM_001167604.1 NM_001324128.1 NM_001324131.1 NM_001324132.1 NM_001324133.1 NM_001324134.1 NM_001324135.1 NM_001324136.1 NM_020383.3 XM_011540134.1 XM_017016613.1 XM_017016614.1 XM_017016615.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1321540 c.1986C>T p.I662I Chr.10: 109865199-109865199 on Build GRCh38
4474947 c.1825C>T p.L609F Chr.10: 109865231-109865231 on Build GRCh38
2060960 c.1968C>T p.I656I Chr.10: 109865217-109865217 on Build GRCh38
1321538 c.1910A>C p.D637A Chr.10: 109865275-109865275 on Build GRCh38
4474946 c.1954C>T p.L652F Chr.10: 109865231-109865231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746467816 Chr.10: 109865271-109865271 on Build GRCh38
rs770908422 Chr.10: 109865277-109865277 on Build GRCh38
rs538836885 Chr.10: 109865258-109865258 on Build GRCh38
rs751692225 Chr.10: 109865309-109865309 on Build GRCh38
rs777253890 Chr.10: 109865269-109865269 on Build GRCh38
rs771549174 Chr.10: 109865217-109865217 on Build GRCh38
rs371130746 Chr.10: 109865288-109865288 on Build GRCh38
rs371885350 Chr.10: 109865229-109865229 on Build GRCh38
rs567637506 Chr.10: 109865219-109865219 on Build GRCh38
rs892387906 Chr.10: 109865183-109865183 on Build GRCh38
rs778671374 Chr.10: 109865161-109865164 on Build GRCh38
rs773176228 Chr.10: 109865218-109865218 on Build GRCh38
rs750873254 Chr.10: 109865335-109865335 on Build GRCh38
rs752167837 Chr.10: 109865242-109865242 on Build GRCh38
rs1052432146 Chr.10: 109865174-109865174 on Build GRCh38
rs758022257 Chr.10: 109865348-109865350 on Build GRCh38
rs112546098 Chr.10: 109865283-109865283 on Build GRCh38
rs142133274 Chr.10: 109865299-109865299 on Build GRCh38
rs781606174 Chr.10: 109865141-109865141 on Build GRCh38
rs946579137 Chr.10: 109865216-109865216 on Build GRCh38
rs200865221 Chr.10: 109865253-109865253 on Build GRCh38
rs771639138 Chr.10: 109865265-109865265 on Build GRCh38
rs778389411 Chr.10: 109865208-109865208 on Build GRCh38
rs750831552 Chr.10: 109865247-109865247 on Build GRCh38
rs769440540 Chr.10: 109865286-109865286 on Build GRCh38
rs763353960 Chr.10: 109865310-109865310 on Build GRCh38
rs776501813 Chr.10: 109865228-109865228 on Build GRCh38
rs773834553 Chr.10: 109865235-109865235 on Build GRCh38
rs768090762 Chr.10: 109865315-109865315 on Build GRCh38
rs897642002 Chr.10: 109865284-109865284 on Build GRCh38
rs770216186 Chr.10: 109865190-109865190 on Build GRCh38
rs148080323 Chr.10: 109865261-109865261 on Build GRCh38
rs781482813 Chr.10: 109865245-109865245 on Build GRCh38
rs747717546 Chr.10: 109865214-109865214 on Build GRCh38
rs371362464 Chr.10: 109865171-109865171 on Build GRCh38
rs142010880 Chr.10: 109865278-109865278 on Build GRCh38
rs775846162 Chr.10: 109865296-109865296 on Build GRCh38
rs754959529 Chr.10: 109865145-109865145 on Build GRCh38
rs746159951 Chr.10: 109865154-109865154 on Build GRCh38
rs770774396 Chr.10: 109865226-109865226 on Build GRCh38
rs764262356 Chr.10: 109865238-109865238 on Build GRCh38
rs756542977 Chr.10: 109865338-109865338 on Build GRCh38
rs483072 Chr.10: 109865350-109865350 on Build GRCh38
rs757852233 Chr.10: 109865243-109865243 on Build GRCh38
rs375157786 Chr.10: 109865170-109865170 on Build GRCh38
rs752290467 Chr.10: 109865324-109865326 on Build GRCh38

View Full Product Details