Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98303232-98303467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABBR2
Entrez Gene ID
9568
Gene Name
gamma-aminobutyric acid type B receptor subunit 2
Gene Aliases
GABABR2, GPR51, GPRC3B, HG20, HRIHFB2099
UniGene
Hs.198612
Gene Chromosome Location
Chr.9: 98288082-98709197 on Build GRCh38
Transcripts
NM_005458.7 XM_005252316.4 XM_017015331.1 XM_017015332.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
199520 c.2318C>T p.T773M Chr.9: 98303335-98303335 on Build GRCh38
4452854 c.2236A>G p.T746A Chr.9: 98303417-98303417 on Build GRCh38
752329 c.2301G>T p.K767N Chr.9: 98303352-98303352 on Build GRCh38
357116 c.2267A>G p.Q756R Chr.9: 98303386-98303386 on Build GRCh38
5063443 c.2280C>T p.F760F Chr.9: 98303373-98303373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376785497 Chr.9: 98303310-98303310 on Build GRCh38
rs764328909 Chr.9: 98303406-98303406 on Build GRCh38
rs369087029 Chr.9: 98303445-98303445 on Build GRCh38
rs776659980 Chr.9: 98303326-98303326 on Build GRCh38
rs201747907 Chr.9: 98303358-98303358 on Build GRCh38
rs779722077 Chr.9: 98303438-98303438 on Build GRCh38
rs138346207 Chr.9: 98303335-98303335 on Build GRCh38
rs762171367 Chr.9: 98303410-98303410 on Build GRCh38
rs578253809 Chr.9: 98303293-98303293 on Build GRCh38
rs745534419 Chr.9: 98303280-98303280 on Build GRCh38
rs759681193 Chr.9: 98303352-98303352 on Build GRCh38
rs192861505 Chr.9: 98303446-98303446 on Build GRCh38
rs779677313 Chr.9: 98303302-98303302 on Build GRCh38
rs768304032 Chr.9: 98303325-98303325 on Build GRCh38
rs916651902 Chr.9: 98303437-98303437 on Build GRCh38
rs372381413 Chr.9: 98303285-98303285 on Build GRCh38
rs1013120257 Chr.9: 98303262-98303262 on Build GRCh38
rs774224900 Chr.9: 98303345-98303345 on Build GRCh38
rs757880673 Chr.9: 98303417-98303417 on Build GRCh38
rs747984740 Chr.9: 98303334-98303334 on Build GRCh38
rs754393016 Chr.9: 98303409-98303409 on Build GRCh38
rs761164200 Chr.9: 98303396-98303396 on Build GRCh38
rs375061314 Chr.9: 98303388-98303388 on Build GRCh38
rs778238324 Chr.9: 98303268-98303268 on Build GRCh38
rs764809039 Chr.9: 98303411-98303411 on Build GRCh38
rs540784101 Chr.9: 98303315-98303315 on Build GRCh38
rs80137447 Chr.9: 98303263-98303263 on Build GRCh38
rs376291087 Chr.9: 98303412-98303412 on Build GRCh38

View Full Product Details