Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43848521-43848790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD300LG
Entrez Gene ID
146894
Gene Name
CD300 molecule like family member g
Gene Aliases
CLM-9, CLM9, NEPMUCIN, TREM-4, TREM4
UniGene
Hs.147313
Gene Chromosome Location
Chr.17: 43846777-43863639 on Build GRCh38
Transcripts
NM_001168322.1 NM_001168323.1 NM_001168324.1 NM_145273.3 XM_005257075.2 XM_005257076.2 XM_006721721.2 XM_006721722.2 XM_011524384.2 XM_017024235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5164161 c.110G>T p.C37F Chr.17: 43848624-43848624 on Build GRCh38
979806 c.205G>T p.E69* Chr.17: 43848719-43848719 on Build GRCh38
5386441 c.71A>G p.E24G Chr.17: 43848585-43848585 on Build GRCh38
5833305 c.174C>T p.F58F Chr.17: 43848688-43848688 on Build GRCh38
3190455 c.121G>A p.E41K Chr.17: 43848635-43848635 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747302869 Chr.17: 43848593-43848593 on Build GRCh38
rs564442578 Chr.17: 43848570-43848570 on Build GRCh38
rs373847576 Chr.17: 43848693-43848693 on Build GRCh38
rs139674731 Chr.17: 43848594-43848594 on Build GRCh38
rs765766049 Chr.17: 43848731-43848731 on Build GRCh38
rs373515876 Chr.17: 43848592-43848592 on Build GRCh38
rs778447487 Chr.17: 43848757-43848757 on Build GRCh38
rs767101356 Chr.17: 43848631-43848631 on Build GRCh38
rs753226601 Chr.17: 43848738-43848738 on Build GRCh38
rs762941051 Chr.17: 43848710-43848710 on Build GRCh38
rs768136580 Chr.17: 43848711-43848711 on Build GRCh38
rs776536925 Chr.17: 43848595-43848595 on Build GRCh38
rs140028124 Chr.17: 43848726-43848726 on Build GRCh38
rs145473485 Chr.17: 43848759-43848759 on Build GRCh38
rs745606836 Chr.17: 43848692-43848692 on Build GRCh38
rs767002311 Chr.17: 43848727-43848727 on Build GRCh38
rs146351601 Chr.17: 43848578-43848578 on Build GRCh38
rs775408229 Chr.17: 43848598-43848598 on Build GRCh38
rs772294550 Chr.17: 43848581-43848581 on Build GRCh38
rs771133413 Chr.17: 43848668-43848668 on Build GRCh38
rs761264036 Chr.17: 43848720-43848720 on Build GRCh38
rs754194502 Chr.17: 43848637-43848637 on Build GRCh38
rs769728837 Chr.17: 43848596-43848596 on Build GRCh38
rs755304047 Chr.17: 43848641-43848641 on Build GRCh38
rs761580079 Chr.17: 43848621-43848621 on Build GRCh38
rs760783261 Chr.17: 43848665-43848665 on Build GRCh38
rs371238467 Chr.17: 43848690-43848690 on Build GRCh38
rs532808839 Chr.17: 43848604-43848604 on Build GRCh38
rs753314239 Chr.17: 43848643-43848643 on Build GRCh38
rs780898715 Chr.17: 43848725-43848725 on Build GRCh38
rs35373935 Chr.17: 43848569-43848569 on Build GRCh38
rs971896234 Chr.17: 43848615-43848615 on Build GRCh38
rs367991783 Chr.17: 43848749-43848749 on Build GRCh38
rs773806292 Chr.17: 43848716-43848716 on Build GRCh38
rs904244227 Chr.17: 43848767-43848767 on Build GRCh38
rs779322180 Chr.17: 43848574-43848574 on Build GRCh38
rs938572546 Chr.17: 43848673-43848673 on Build GRCh38
rs1026496781 Chr.17: 43848611-43848611 on Build GRCh38
rs148841815 Chr.17: 43848768-43848768 on Build GRCh38
rs781414083 Chr.17: 43848763-43848763 on Build GRCh38
rs115182680 Chr.17: 43848654-43848654 on Build GRCh38
rs762387492 Chr.17: 43848599-43848599 on Build GRCh38
rs72836561 Chr.17: 43848758-43848758 on Build GRCh38
rs144430828 Chr.17: 43848614-43848614 on Build GRCh38
rs187296668 Chr.17: 43848653-43848653 on Build GRCh38

View Full Product Details