Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65665310-65665575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND4A
Entrez Gene ID
10260
Gene Name
DENN domain containing 4A
Gene Aliases
IRLB, MYCPBP
UniGene
Hs.654567
Gene Chromosome Location
Chr.15: 65659123-65792293 on Build GRCh38
Transcripts
NM_001144823.2 NM_001320835.1 NM_005848.4 XM_005254120.4 XM_005254121.3 XM_005254122.4 XM_006720371.3 XM_011521157.2 XM_017021862.1 XM_017021863.1 XM_017021864.1 XM_017021865.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4640009 c.5326C>T p.Q1776* Chr.15: 65665375-65665375 on Build GRCh38
6876498 c.5243C>T p.P1748L Chr.15: 65665458-65665458 on Build GRCh38
6015827 c.5116C>T p.R1706C Chr.15: 65665456-65665456 on Build GRCh38
6642391 c.5192C>T p.P1731L Chr.15: 65665380-65665380 on Build GRCh38
4771918 c.5149_5150delTT p.L1717fs*39 Chr.15: 65665422-65665423 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs78890197 Chr.15: 65665460-65665460 on Build GRCh38
rs757437909 Chr.15: 65665455-65665455 on Build GRCh38
rs764864062 Chr.15: 65665381-65665381 on Build GRCh38
rs192164013 Chr.15: 65665513-65665513 on Build GRCh38
rs779019250 Chr.15: 65665456-65665456 on Build GRCh38
rs1050977036 Chr.15: 65665480-65665480 on Build GRCh38
rs1006172986 Chr.15: 65665409-65665409 on Build GRCh38
rs752574715 Chr.15: 65665535-65665538 on Build GRCh38
rs185363567 Chr.15: 65665489-65665489 on Build GRCh38
rs756734941 Chr.15: 65665426-65665426 on Build GRCh38
rs745915201 Chr.15: 65665484-65665484 on Build GRCh38
rs749880851 Chr.15: 65665387-65665387 on Build GRCh38
rs746923662 Chr.15: 65665506-65665506 on Build GRCh38
rs757858899 Chr.15: 65665396-65665396 on Build GRCh38
rs376126692 Chr.15: 65665453-65665453 on Build GRCh38
rs760420499 Chr.15: 65665370-65665370 on Build GRCh38
rs771991613 Chr.15: 65665353-65665353 on Build GRCh38
rs778269892 Chr.15: 65665433-65665433 on Build GRCh38
rs780144803 Chr.15: 65665502-65665502 on Build GRCh38
rs11293131 Chr.15: 65665469-65665469 on Build GRCh38
rs534439517 Chr.15: 65665468-65665468 on Build GRCh38
rs761508769 Chr.15: 65665380-65665380 on Build GRCh38
rs758838925 Chr.15: 65665397-65665397 on Build GRCh38
rs374087484 Chr.15: 65665408-65665408 on Build GRCh38
rs1025440601 Chr.15: 65665485-65665485 on Build GRCh38
rs768086262 Chr.15: 65665405-65665405 on Build GRCh38
rs61751113 Chr.15: 65665379-65665379 on Build GRCh38
rs1023965261 Chr.15: 65665515-65665515 on Build GRCh38
rs775430902 Chr.15: 65665364-65665364 on Build GRCh38
rs952403798 Chr.15: 65665541-65665541 on Build GRCh38
rs1009868463 Chr.15: 65665404-65665404 on Build GRCh38
rs753226756 Chr.15: 65665420-65665420 on Build GRCh38
rs750885824 Chr.15: 65665383-65665383 on Build GRCh38

View Full Product Details