Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 38689139-38689368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FREM2
Entrez Gene ID
341640
Gene Name
FRAS1 related extracellular matrix protein 2
Gene Aliases
-
UniGene
Hs.253994
Gene Chromosome Location
Chr.13: 38687036-38887131 on Build GRCh38
Transcripts
NM_207361.5 XM_017020554.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1207520 c.1886G>T p.W629L Chr.13: 38689230-38689230 on Build GRCh38
6666835 c.1820C>T p.T607M Chr.13: 38689164-38689164 on Build GRCh38
947270 c.1857T>C p.H619H Chr.13: 38689201-38689201 on Build GRCh38
6666837 c.1804T>A p.S602T Chr.13: 38689148-38689148 on Build GRCh38
3704622 c.2012C>A p.T671K Chr.13: 38689356-38689356 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910157803 Chr.13: 38689239-38689239 on Build GRCh38
rs959161171 Chr.13: 38689220-38689220 on Build GRCh38
rs769953468 Chr.13: 38689159-38689159 on Build GRCh38
rs199905904 Chr.13: 38689194-38689194 on Build GRCh38
rs115928688 Chr.13: 38689164-38689164 on Build GRCh38
rs370751010 Chr.13: 38689201-38689201 on Build GRCh38
rs758214399 Chr.13: 38689200-38689200 on Build GRCh38
rs764816586 Chr.13: 38689306-38689306 on Build GRCh38
rs766908727 Chr.13: 38689297-38689297 on Build GRCh38
rs771066976 Chr.13: 38689211-38689211 on Build GRCh38
rs781385468 Chr.13: 38689180-38689181 on Build GRCh38
rs749358845 Chr.13: 38689257-38689257 on Build GRCh38
rs780054707 Chr.13: 38689244-38689244 on Build GRCh38
rs774584521 Chr.13: 38689260-38689260 on Build GRCh38
rs754650884 Chr.13: 38689196-38689196 on Build GRCh38
rs772360572 Chr.13: 38689268-38689268 on Build GRCh38
rs200412879 Chr.13: 38689263-38689263 on Build GRCh38
rs752354093 Chr.13: 38689315-38689315 on Build GRCh38
rs762144113 Chr.13: 38689168-38689168 on Build GRCh38
rs971647391 Chr.13: 38689337-38689337 on Build GRCh38
rs142526257 Chr.13: 38689323-38689323 on Build GRCh38
rs976499456 Chr.13: 38689177-38689177 on Build GRCh38
rs147736905 Chr.13: 38689273-38689273 on Build GRCh38
rs145673360 Chr.13: 38689182-38689182 on Build GRCh38
rs1037243942 Chr.13: 38689270-38689270 on Build GRCh38
rs199661318 Chr.13: 38689223-38689223 on Build GRCh38
rs116802472 Chr.13: 38689265-38689265 on Build GRCh38
rs763026321 Chr.13: 38689165-38689165 on Build GRCh38
rs549157910 Chr.13: 38689329-38689329 on Build GRCh38
rs759061968 Chr.13: 38689299-38689299 on Build GRCh38
rs895211917 Chr.13: 38689321-38689321 on Build GRCh38
rs774621627 Chr.13: 38689167-38689167 on Build GRCh38
rs768812595 Chr.13: 38689259-38689259 on Build GRCh38
rs770004708 Chr.13: 38689235-38689235 on Build GRCh38
rs140036522 Chr.13: 38689181-38689181 on Build GRCh38
rs768034718 Chr.13: 38689180-38689180 on Build GRCh38
rs751385106 Chr.13: 38689333-38689333 on Build GRCh38
rs41292751 Chr.13: 38689285-38689285 on Build GRCh38
rs376624387 Chr.13: 38689179-38689179 on Build GRCh38
rs897320496 Chr.13: 38689272-38689272 on Build GRCh38
rs941602944 Chr.13: 38689255-38689255 on Build GRCh38
rs778594343 Chr.13: 38689197-38689197 on Build GRCh38
rs752479278 Chr.13: 38689198-38689198 on Build GRCh38

View Full Product Details