Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REV1
Entrez Gene ID
51455
Gene Name
REV1, DNA directed polymerase
Gene Aliases
AIBP80, REV1L
Chromosome Location
Chr.2: 99400475 - 99490123 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037872.2 NP_001032961.1 1 - 2 244 73
NM_001321454.1 NP_001308383.1 1 - 2 244 73
NM_001321455.1 NP_001308384.1 1 - 2 244 73
NM_001321458.1 NP_001308387.1 1 - 2 244 73
NM_001321459.1 NP_001308388.1 1 - 2 244 73
NM_001321460.1 NP_001308389.1 1 - 2 244 73
NM_016316.3 NP_057400.1 1 - 2 244 73
NR_135649.1 1 - 2 244 73
NR_135650.1 1 - 2 244 73
NR_135651.1 1 - 2 244 73
NR_135652.1 1 - 2 244 73
NR_135653.1 1 - 2 244 73
XM_017004312.1 XP_016859801.1 1 - 2 239 73
GenBank mRNA AB047646.1 - 1 - 2 169 73
AF151538.1 - 1 - 2 197 73
AF206019.1 - 1 - 2 206 73
AF357886.1 - 1 - 2 162 73
AK307719.1 - 1 - 2 155 73
BC037734.1 - 1 - 2 225 73
DR000409.1 - 1 - 2 135 73
HY010913.1 - 1 - 2 245 73

View Full Product Details