Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAC1
Entrez Gene ID
403955
Gene Name
Rac family small GTPase 1
Gene Aliases
RAC2
Chromosome Location
Chr.6: 11726259 - 11758532 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003274.2 NP_001003274.1 1 - 1 101 105
XM_005621021.1 XP_005621078.1 - 104 105
XM_022419478.1 XP_022275186.1 - 95 105
GenBank mRNA X56390.1 - - 101 105

View Full Product Details