Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72002461-72002956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL18BP
Entrez Gene ID
10068
Gene Name
interleukin 18 binding protein
Gene Aliases
IL18BPa
UniGene
Hs.591967
Gene Chromosome Location
Chr.11: 71997944-72002919 on Build GRCh38
Transcripts
NM_001039659.1 NM_001039660.1 NM_001145055.1 NM_001145057.1 NM_005699.3 NM_173042.2 NM_173044.2 XM_017017059.1 XM_017017060.1 XM_017017061.1 XM_017017062.1 XM_017017063.1 NM_001286561.1 NM_006185.3 NR_104476.1 XM_006718564.1 XM_011545054.1 XM_011545055.1 XM_011545056.2 XM_011545057.2 XM_011545058.1 XM_011545059.2 XM_011545060.2 XM_011545061.2 XM_011545062.2 XM_011545063.2 XM_011545064.1 XM_011545065.1 XM_011545066.1

Gene Symbol
NUMA1
Entrez Gene ID
4926
Gene Name
nuclear mitotic apparatus protein 1
Gene Aliases
NMP-22, NUMA
UniGene
Hs.325978
Gene Chromosome Location
Chr.11: 72002864-72080693 on Build GRCh38
Transcripts
NM_001039659.1 NM_001039660.1 NM_001145055.1 NM_001145057.1 NM_005699.3 NM_173042.2 NM_173044.2 XM_017017059.1 XM_017017060.1 XM_017017061.1 XM_017017062.1 XM_017017063.1 NM_001286561.1 NM_006185.3 NR_104476.1 XM_006718564.1 XM_011545054.1 XM_011545055.1 XM_011545056.2 XM_011545057.2 XM_011545058.1 XM_011545059.2 XM_011545060.2 XM_011545061.2 XM_011545062.2 XM_011545063.2 XM_011545064.1 XM_011545065.1 XM_011545066.1

SNP Detail

SNP ID Chromosome Location
rs1040771898 Chr.11: 72002679-72002679 on Build GRCh38
rs910508533 Chr.11: 72002866-72002866 on Build GRCh38
rs984612539 Chr.11: 72002865-72002865 on Build GRCh38
rs529816161 Chr.11: 72002659-72002659 on Build GRCh38
rs755106939 Chr.11: 72002720-72002721 on Build GRCh38
rs751554109 Chr.11: 72002602-72002603 on Build GRCh38
rs930562455 Chr.11: 72002650-72002650 on Build GRCh38
rs5743678 Chr.11: 72002852-72002852 on Build GRCh38
rs566683507 Chr.11: 72002699-72002699 on Build GRCh38
rs749724160 Chr.11: 72002846-72002846 on Build GRCh38
rs945499147 Chr.11: 72002882-72002882 on Build GRCh38
rs139225739 Chr.11: 72002512-72002512 on Build GRCh38
rs558942289 Chr.11: 72002785-72002785 on Build GRCh38
rs956126702 Chr.11: 72002747-72002747 on Build GRCh38
rs1024352475 Chr.11: 72002827-72002827 on Build GRCh38
rs569811260 Chr.11: 72002871-72002871 on Build GRCh38
rs547025486 Chr.11: 72002674-72002675 on Build GRCh38
rs924483926 Chr.11: 72002592-72002592 on Build GRCh38
rs751036360 Chr.11: 72002649-72002649 on Build GRCh38
rs550616026 Chr.11: 72002621-72002621 on Build GRCh38
rs537945669 Chr.11: 72002868-72002868 on Build GRCh38
rs755844999 Chr.11: 72002552-72002552 on Build GRCh38
rs943226346 Chr.11: 72002867-72002867 on Build GRCh38
rs959607460 Chr.11: 72002755-72002755 on Build GRCh38
rs968012916 Chr.11: 72002795-72002795 on Build GRCh38
rs966263072 Chr.11: 72002577-72002577 on Build GRCh38
rs919416690 Chr.11: 72002850-72002850 on Build GRCh38
rs78932475 Chr.11: 72002922-72002922 on Build GRCh38
rs374681765 Chr.11: 72002622-72002622 on Build GRCh38
rs935353630 Chr.11: 72002600-72002600 on Build GRCh38
rs903934857 Chr.11: 72002906-72002906 on Build GRCh38
rs919052189 Chr.11: 72002619-72002619 on Build GRCh38
rs905392205 Chr.11: 72002633-72002634 on Build GRCh38
rs529615222 Chr.11: 72002819-72002819 on Build GRCh38
rs779520414 Chr.11: 72002912-72002912 on Build GRCh38
rs14537 Chr.11: 72002907-72002907 on Build GRCh38
rs142149472 Chr.11: 72002841-72002841 on Build GRCh38
rs147624645 Chr.11: 72002766-72002766 on Build GRCh38
rs971179660 Chr.11: 72002839-72002839 on Build GRCh38
rs968597643 Chr.11: 72002701-72002701 on Build GRCh38
rs997724053 Chr.11: 72002714-72002714 on Build GRCh38
rs562353186 Chr.11: 72002639-72002639 on Build GRCh38
rs190921526 Chr.11: 72002923-72002923 on Build GRCh38
rs532517978 Chr.11: 72002530-72002530 on Build GRCh38
rs991102958 Chr.11: 72002498-72002498 on Build GRCh38
rs898045482 Chr.11: 72002671-72002671 on Build GRCh38
rs949837224 Chr.11: 72002538-72002538 on Build GRCh38
rs990006027 Chr.11: 72002781-72002781 on Build GRCh38
rs915269187 Chr.11: 72002783-72002783 on Build GRCh38
rs889020201 Chr.11: 72002660-72002660 on Build GRCh38
rs1042888777 Chr.11: 72002885-72002885 on Build GRCh38
rs1007597290 Chr.11: 72002673-72002673 on Build GRCh38
rs1031601080 Chr.11: 72002728-72002728 on Build GRCh38
rs5743677 Chr.11: 72002686-72002686 on Build GRCh38
rs573460986 Chr.11: 72002496-72002496 on Build GRCh38
rs777929000 Chr.11: 72002824-72002824 on Build GRCh38
rs1000969227 Chr.11: 72002719-72002719 on Build GRCh38
rs919117247 Chr.11: 72002845-72002845 on Build GRCh38
rs1045113448 Chr.11: 72002621-72002622 on Build GRCh38
rs367746979 Chr.11: 72002711-72002711 on Build GRCh38
rs976065476 Chr.11: 72002590-72002590 on Build GRCh38
rs915344740 Chr.11: 72002606-72002606 on Build GRCh38
rs939550308 Chr.11: 72002843-72002843 on Build GRCh38
rs1021135182 Chr.11: 72002700-72002700 on Build GRCh38
rs951754366 Chr.11: 72002488-72002488 on Build GRCh38
rs1056727614 Chr.11: 72002666-72002670 on Build GRCh38
rs946824081 Chr.11: 72002608-72002608 on Build GRCh38
rs887429175 Chr.11: 72002682-72002682 on Build GRCh38
rs564926390 Chr.11: 72002523-72002523 on Build GRCh38
rs1017478534 Chr.11: 72002549-72002549 on Build GRCh38
rs1033921722 Chr.11: 72002738-72002738 on Build GRCh38
rs901811753 Chr.11: 72002881-72002881 on Build GRCh38

View Full Product Details