Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 76349870-76350376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THAP12
Entrez Gene ID
5612
Gene Name
THAP domain containing 12
Gene Aliases
DAP4, P52rIPK, PRKRIR, THAP0
UniGene
Hs.503315
Gene Chromosome Location
Chr.11: 76349956-76380965 on Build GRCh38
Transcripts
NM_004705.3 NR_130898.1

SNP Detail

SNP ID Chromosome Location
rs1028283295 Chr.11: 76350315-76350315 on Build GRCh38
rs879394581 Chr.11: 76349905-76349905 on Build GRCh38
rs962272185 Chr.11: 76349907-76349907 on Build GRCh38
rs935906444 Chr.11: 76350142-76350143 on Build GRCh38
rs566221325 Chr.11: 76350224-76350224 on Build GRCh38
rs770364616 Chr.11: 76349972-76349976 on Build GRCh38
rs1036687294 Chr.11: 76350222-76350222 on Build GRCh38
rs188213775 Chr.11: 76350192-76350192 on Build GRCh38
rs906938237 Chr.11: 76350199-76350199 on Build GRCh38
rs564212275 Chr.11: 76349972-76349972 on Build GRCh38
rs948520336 Chr.11: 76350158-76350158 on Build GRCh38
rs1019414637 Chr.11: 76350070-76350070 on Build GRCh38
rs183488201 Chr.11: 76350033-76350033 on Build GRCh38
rs995190526 Chr.11: 76350278-76350278 on Build GRCh38
rs889384955 Chr.11: 76350316-76350316 on Build GRCh38
rs762094290 Chr.11: 76350019-76350019 on Build GRCh38
rs989994753 Chr.11: 76350143-76350143 on Build GRCh38
rs546425900 Chr.11: 76349949-76349949 on Build GRCh38
rs192384495 Chr.11: 76350241-76350241 on Build GRCh38
rs977391774 Chr.11: 76350108-76350108 on Build GRCh38
rs764967615 Chr.11: 76350018-76350018 on Build GRCh38
rs777021019 Chr.11: 76349969-76349970 on Build GRCh38
rs374516491 Chr.11: 76350255-76350255 on Build GRCh38
rs995116776 Chr.11: 76349946-76349946 on Build GRCh38
rs1045577267 Chr.11: 76350175-76350175 on Build GRCh38
rs939790110 Chr.11: 76350205-76350205 on Build GRCh38
rs753872063 Chr.11: 76349906-76349906 on Build GRCh38
rs555608617 Chr.11: 76350143-76350143 on Build GRCh38
rs567698991 Chr.11: 76350038-76350038 on Build GRCh38
rs966189298 Chr.11: 76350094-76350094 on Build GRCh38
rs953647918 Chr.11: 76350032-76350032 on Build GRCh38
rs577718956 Chr.11: 76350307-76350307 on Build GRCh38
rs569594571 Chr.11: 76350106-76350106 on Build GRCh38
rs535790453 Chr.11: 76350176-76350176 on Build GRCh38
rs924637015 Chr.11: 76350122-76350122 on Build GRCh38

View Full Product Details