Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 82914632-82915137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDIAS
Entrez Gene ID
220042
Gene Name
DNA damage induced apoptosis suppressor
Gene Aliases
C11orf82, noxin
UniGene
Hs.165607
Gene Chromosome Location
Chr.11: 82901695-82934659 on Build GRCh38
Transcripts
NM_145018.3 XM_011544835.1 XM_011544836.2 XM_011544838.2 XM_017017352.1 XM_017017353.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3453906 c.46C>T p.Q16* Chr.11: 82914784-82914784 on Build GRCh38
274200 c.22C>A p.L8I Chr.11: 82914760-82914760 on Build GRCh38
3752777 c.34G>A p.V12I Chr.11: 82914772-82914772 on Build GRCh38
3453907 c.107C>T p.S36F Chr.11: 82914845-82914845 on Build GRCh38
268777 c.63A>G p.I21M Chr.11: 82914801-82914801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775082867 Chr.11: 82914735-82914735 on Build GRCh38
rs750930355 Chr.11: 82914752-82914752 on Build GRCh38
rs747965063 Chr.11: 82914705-82914705 on Build GRCh38
rs139177881 Chr.11: 82915080-82915080 on Build GRCh38
rs938068444 Chr.11: 82915049-82915049 on Build GRCh38
rs917014160 Chr.11: 82915092-82915092 on Build GRCh38
rs368903831 Chr.11: 82914703-82914703 on Build GRCh38
rs764873036 Chr.11: 82914728-82914728 on Build GRCh38
rs974606746 Chr.11: 82915039-82915039 on Build GRCh38
rs373687907 Chr.11: 82914912-82914912 on Build GRCh38
rs746987452 Chr.11: 82914800-82914800 on Build GRCh38
rs180790055 Chr.11: 82914950-82914950 on Build GRCh38
rs1028212895 Chr.11: 82915066-82915066 on Build GRCh38
rs74752163 Chr.11: 82914714-82914714 on Build GRCh38
rs553910788 Chr.11: 82914992-82914992 on Build GRCh38
rs548481924 Chr.11: 82914733-82914733 on Build GRCh38
rs937050457 Chr.11: 82915089-82915089 on Build GRCh38
rs779222473 Chr.11: 82914704-82914704 on Build GRCh38
rs777593915 Chr.11: 82914799-82914799 on Build GRCh38
rs773354817 Chr.11: 82914709-82914709 on Build GRCh38
rs756192318 Chr.11: 82914693-82914693 on Build GRCh38
rs555798954 Chr.11: 82914818-82914818 on Build GRCh38
rs200274419 Chr.11: 82914768-82914768 on Build GRCh38
rs1032767791 Chr.11: 82914922-82914922 on Build GRCh38
rs775172518 Chr.11: 82914841-82914841 on Build GRCh38
rs1041908325 Chr.11: 82914781-82914781 on Build GRCh38
rs769426543 Chr.11: 82914716-82914716 on Build GRCh38
rs779312498 Chr.11: 82914784-82914784 on Build GRCh38
rs998740771 Chr.11: 82915105-82915105 on Build GRCh38
rs117929011 Chr.11: 82915070-82915070 on Build GRCh38
rs569378998 Chr.11: 82914699-82914699 on Build GRCh38
rs758987657 Chr.11: 82914727-82914727 on Build GRCh38
rs992447713 Chr.11: 82915090-82915090 on Build GRCh38
rs766917044 Chr.11: 82914866-82914866 on Build GRCh38
rs745412190 Chr.11: 82914810-82914810 on Build GRCh38
rs762632200 Chr.11: 82914842-82914842 on Build GRCh38
rs142883872 Chr.11: 82914746-82914746 on Build GRCh38
rs903032672 Chr.11: 82914896-82914896 on Build GRCh38
rs981020159 Chr.11: 82915073-82915073 on Build GRCh38
rs750889159 Chr.11: 82914679-82914679 on Build GRCh38
rs1027933893 Chr.11: 82914949-82914949 on Build GRCh38
rs754361149 Chr.11: 82914871-82914871 on Build GRCh38
rs957118029 Chr.11: 82914933-82914933 on Build GRCh38
rs139641669 Chr.11: 82914757-82914757 on Build GRCh38
rs147443808 Chr.11: 82914751-82914751 on Build GRCh38
rs573538532 Chr.11: 82914905-82914905 on Build GRCh38
rs150528544 Chr.11: 82914801-82914801 on Build GRCh38
rs759715868 Chr.11: 82914872-82914872 on Build GRCh38
rs199579071 Chr.11: 82914847-82914847 on Build GRCh38
rs771864976 Chr.11: 82914712-82914712 on Build GRCh38
rs768652012 Chr.11: 82914687-82914687 on Build GRCh38
rs896467291 Chr.11: 82915078-82915078 on Build GRCh38
rs752922430 Chr.11: 82914797-82914797 on Build GRCh38
rs962402425 Chr.11: 82914973-82914973 on Build GRCh38
rs765386482 Chr.11: 82914903-82914903 on Build GRCh38
rs149655535 Chr.11: 82914846-82914846 on Build GRCh38
rs1045661171 Chr.11: 82915091-82915091 on Build GRCh38
rs760893299 Chr.11: 82914855-82914855 on Build GRCh38
rs773046419 Chr.11: 82914718-82914718 on Build GRCh38
rs781039063 Chr.11: 82914807-82914807 on Build GRCh38
rs781283390 Chr.11: 82914803-82914806 on Build GRCh38
rs537271353 Chr.11: 82914962-82914962 on Build GRCh38
rs758176091 Chr.11: 82914798-82914798 on Build GRCh38
rs770388916 Chr.11: 82914723-82914723 on Build GRCh38
rs776202115 Chr.11: 82914724-82914724 on Build GRCh38
rs577309703 Chr.11: 82914952-82914952 on Build GRCh38
rs940161295 Chr.11: 82914708-82914708 on Build GRCh38
rs903062476 Chr.11: 82915095-82915095 on Build GRCh38
rs143540019 Chr.11: 82914760-82914760 on Build GRCh38
rs926935019 Chr.11: 82914736-82914736 on Build GRCh38
rs972631887 Chr.11: 82914698-82914698 on Build GRCh38
rs761020338 Chr.11: 82914722-82914722 on Build GRCh38
rs1024579965 Chr.11: 82914834-82914834 on Build GRCh38
rs940211218 Chr.11: 82914988-82914988 on Build GRCh38
rs975084225 Chr.11: 82914938-82914938 on Build GRCh38
rs61902276 Chr.11: 82914772-82914772 on Build GRCh38
rs920046592 Chr.11: 82915041-82915041 on Build GRCh38
rs375533789 Chr.11: 82914701-82914701 on Build GRCh38

View Full Product Details