Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 95818685-95819220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP57
Entrez Gene ID
9702
Gene Name
centrosomal protein 57
Gene Aliases
MVA2, PIG8, TSP57
UniGene
Hs.101014
Gene Chromosome Location
Chr.11: 95790461-95832693 on Build GRCh38
Transcripts
NM_001243776.1 NM_001243777.1 NM_014679.4 XM_006718945.2 XM_006718946.2 XM_017018592.1 XM_017018593.1 XM_017018594.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6846026 c.671G>T p.R224M Chr.11: 95818876-95818876 on Build GRCh38
239296 c.676C>T p.R226C Chr.11: 95818881-95818881 on Build GRCh38
6223602 c.699G>T p.E233D Chr.11: 95818904-95818904 on Build GRCh38
430113 c.692C>G p.A231G Chr.11: 95818897-95818897 on Build GRCh38
6864403 c.631G>A p.E211K Chr.11: 95818863-95818863 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200535773 Chr.11: 95818838-95818838 on Build GRCh38
rs767039480 Chr.11: 95818802-95818802 on Build GRCh38
rs545967011 Chr.11: 95818907-95818907 on Build GRCh38
rs141039043 Chr.11: 95819001-95819001 on Build GRCh38
rs143711180 Chr.11: 95818882-95818882 on Build GRCh38
rs988705929 Chr.11: 95819187-95819187 on Build GRCh38
rs560205172 Chr.11: 95819015-95819015 on Build GRCh38
rs758284146 Chr.11: 95818859-95818859 on Build GRCh38
rs570978492 Chr.11: 95819050-95819050 on Build GRCh38
rs562503341 Chr.11: 95818983-95818983 on Build GRCh38
rs755140070 Chr.11: 95818875-95818875 on Build GRCh38
rs564157453 Chr.11: 95818930-95818930 on Build GRCh38
rs200137467 Chr.11: 95818856-95818856 on Build GRCh38
rs190912541 Chr.11: 95818997-95818997 on Build GRCh38
rs750363422 Chr.11: 95818857-95818857 on Build GRCh38
rs903450982 Chr.11: 95818815-95818815 on Build GRCh38
rs138222051 Chr.11: 95818837-95818837 on Build GRCh38
rs963446452 Chr.11: 95818711-95818711 on Build GRCh38
rs910295771 Chr.11: 95819111-95819111 on Build GRCh38
rs937529119 Chr.11: 95819009-95819009 on Build GRCh38
rs374700509 Chr.11: 95818833-95818833 on Build GRCh38
rs1036346628 Chr.11: 95819092-95819093 on Build GRCh38
rs527816583 Chr.11: 95819019-95819019 on Build GRCh38
rs552716201 Chr.11: 95819045-95819045 on Build GRCh38
rs969211120 Chr.11: 95818911-95818911 on Build GRCh38
rs574499415 Chr.11: 95818751-95818751 on Build GRCh38
rs199507017 Chr.11: 95818735-95818735 on Build GRCh38
rs778056271 Chr.11: 95818892-95818892 on Build GRCh38
rs889261108 Chr.11: 95818773-95818773 on Build GRCh38
rs540998751 Chr.11: 95819141-95819142 on Build GRCh38
rs896065083 Chr.11: 95819034-95819034 on Build GRCh38
rs746863051 Chr.11: 95818860-95818862 on Build GRCh38
rs186589069 Chr.11: 95819145-95819145 on Build GRCh38
rs1040563223 Chr.11: 95818801-95818801 on Build GRCh38
rs770187092 Chr.11: 95818887-95818887 on Build GRCh38
rs912475749 Chr.11: 95818934-95818934 on Build GRCh38
rs1019434910 Chr.11: 95819164-95819164 on Build GRCh38
rs553842178 Chr.11: 95819147-95819147 on Build GRCh38
rs775289872 Chr.11: 95818821-95818822 on Build GRCh38
rs550502984 Chr.11: 95819092-95819092 on Build GRCh38
rs1014969549 Chr.11: 95818709-95818709 on Build GRCh38
rs370448879 Chr.11: 95818932-95818932 on Build GRCh38
rs771347163 Chr.11: 95818916-95818916 on Build GRCh38
rs151075559 Chr.11: 95818948-95818948 on Build GRCh38
rs1054649966 Chr.11: 95819154-95819154 on Build GRCh38
rs375237035 Chr.11: 95818946-95818946 on Build GRCh38
rs142615007 Chr.11: 95818862-95818862 on Build GRCh38
rs73531050 Chr.11: 95818981-95818981 on Build GRCh38
rs34620235 Chr.11: 95819015-95819016 on Build GRCh38
rs758512770 Chr.11: 95818813-95818813 on Build GRCh38
rs144903440 Chr.11: 95818881-95818881 on Build GRCh38
rs751672542 Chr.11: 95818874-95818874 on Build GRCh38
rs934935323 Chr.11: 95819152-95819152 on Build GRCh38
rs745634452 Chr.11: 95818813-95818814 on Build GRCh38
rs944886989 Chr.11: 95818794-95818794 on Build GRCh38
rs988422740 Chr.11: 95818947-95818947 on Build GRCh38
rs765009552 Chr.11: 95818855-95818855 on Build GRCh38
rs182738434 Chr.11: 95819137-95819137 on Build GRCh38
rs778589075 Chr.11: 95818789-95818789 on Build GRCh38
rs961996507 Chr.11: 95818960-95818960 on Build GRCh38
rs893115088 Chr.11: 95819169-95819169 on Build GRCh38
rs777770670 Chr.11: 95819125-95819125 on Build GRCh38
rs371398882 Chr.11: 95818829-95818829 on Build GRCh38
rs768644523 Chr.11: 95818948-95818948 on Build GRCh38
rs775153753 Chr.11: 95818803-95818803 on Build GRCh38
rs760336031 Chr.11: 95818810-95818810 on Build GRCh38
rs940952870 Chr.11: 95819064-95819064 on Build GRCh38
rs771817239 Chr.11: 95818814-95818814 on Build GRCh38
rs868405509 Chr.11: 95818908-95818908 on Build GRCh38
rs538496392 Chr.11: 95819054-95819054 on Build GRCh38
rs965247204 Chr.11: 95819173-95819173 on Build GRCh38

View Full Product Details