Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 95821788-95822300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP57
Entrez Gene ID
9702
Gene Name
centrosomal protein 57
Gene Aliases
MVA2, PIG8, TSP57
UniGene
Hs.101014
Gene Chromosome Location
Chr.11: 95790461-95832693 on Build GRCh38
Transcripts
NM_001243776.1 NM_001243777.1 NM_014679.4 XM_006718945.2 XM_006718946.2 XM_017018592.1 XM_017018593.1 XM_017018594.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1357995 c.778delA p.K262fs*31 Chr.11: 95821949-95821949 on Build GRCh38
6135074 c.698G>C p.R233T Chr.11: 95821896-95821896 on Build GRCh38
7449192 c.709G>T p.E237* Chr.11: 95821907-95821907 on Build GRCh38
2061835 c.721A>G p.N241D Chr.11: 95821892-95821892 on Build GRCh38
5446984 c.778A>T p.K260* Chr.11: 95821949-95821949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923018567 Chr.11: 95821851-95821851 on Build GRCh38
rs758901369 Chr.11: 95821825-95821825 on Build GRCh38
rs61903295 Chr.11: 95821969-95821969 on Build GRCh38
rs934272401 Chr.11: 95822134-95822134 on Build GRCh38
rs530427449 Chr.11: 95822066-95822066 on Build GRCh38
rs1046407529 Chr.11: 95822270-95822270 on Build GRCh38
rs941528018 Chr.11: 95821857-95821857 on Build GRCh38
rs76411609 Chr.11: 95821894-95821894 on Build GRCh38
rs765532194 Chr.11: 95821949-95821949 on Build GRCh38
rs768269976 Chr.11: 95821935-95821935 on Build GRCh38
rs779531903 Chr.11: 95821923-95821923 on Build GRCh38
rs974368664 Chr.11: 95822118-95822118 on Build GRCh38
rs765028167 Chr.11: 95821855-95821855 on Build GRCh38
rs757692899 Chr.11: 95822020-95822020 on Build GRCh38
rs773606683 Chr.11: 95822000-95822000 on Build GRCh38
rs762912636 Chr.11: 95821878-95821878 on Build GRCh38
rs528203509 Chr.11: 95822239-95822239 on Build GRCh38
rs750192070 Chr.11: 95821872-95821872 on Build GRCh38
rs766330674 Chr.11: 95821888-95821888 on Build GRCh38
rs553879690 Chr.11: 95822204-95822204 on Build GRCh38
rs762013265 Chr.11: 95821948-95821949 on Build GRCh38
rs376366996 Chr.11: 95821946-95821946 on Build GRCh38
rs770564750 Chr.11: 95822095-95822095 on Build GRCh38
rs1039887699 Chr.11: 95822083-95822083 on Build GRCh38
rs546452798 Chr.11: 95822268-95822268 on Build GRCh38
rs933041172 Chr.11: 95822202-95822202 on Build GRCh38
rs577605522 Chr.11: 95822194-95822194 on Build GRCh38
rs372063775 Chr.11: 95821890-95821890 on Build GRCh38
rs563096432 Chr.11: 95822051-95822051 on Build GRCh38
rs11021328 Chr.11: 95822148-95822148 on Build GRCh38
rs376006876 Chr.11: 95822001-95822001 on Build GRCh38
rs941532209 Chr.11: 95822067-95822067 on Build GRCh38
rs778132447 Chr.11: 95821902-95821902 on Build GRCh38
rs746321531 Chr.11: 95822024-95822024 on Build GRCh38
rs1001489205 Chr.11: 95821934-95821934 on Build GRCh38
rs149293164 Chr.11: 95821922-95821922 on Build GRCh38
rs756320195 Chr.11: 95822009-95822009 on Build GRCh38
rs895424805 Chr.11: 95821949-95821952 on Build GRCh38
rs773014425 Chr.11: 95821949-95821949 on Build GRCh38
rs764244846 Chr.11: 95822015-95822015 on Build GRCh38
rs895386299 Chr.11: 95822256-95822256 on Build GRCh38
rs143492267 Chr.11: 95821821-95821821 on Build GRCh38
rs748882845 Chr.11: 95822078-95822078 on Build GRCh38
rs776571098 Chr.11: 95821853-95821853 on Build GRCh38
rs779479034 Chr.11: 95822021-95822021 on Build GRCh38
rs761465423 Chr.11: 95821945-95821945 on Build GRCh38
rs888598885 Chr.11: 95821833-95821833 on Build GRCh38
rs758981182 Chr.11: 95822027-95822027 on Build GRCh38
rs892441206 Chr.11: 95822164-95822164 on Build GRCh38
rs964634775 Chr.11: 95822071-95822074 on Build GRCh38
rs117602509 Chr.11: 95821986-95821986 on Build GRCh38
rs368470481 Chr.11: 95821962-95821962 on Build GRCh38
rs763298656 Chr.11: 95822029-95822030 on Build GRCh38
rs181776180 Chr.11: 95821981-95821981 on Build GRCh38
rs1038714604 Chr.11: 95821820-95821820 on Build GRCh38
rs1051299243 Chr.11: 95822163-95822163 on Build GRCh38
rs982513458 Chr.11: 95822044-95822044 on Build GRCh38
rs755015433 Chr.11: 95821892-95821892 on Build GRCh38
rs370418539 Chr.11: 95822026-95822026 on Build GRCh38
rs756302353 Chr.11: 95821898-95821898 on Build GRCh38
rs1054397171 Chr.11: 95822243-95822243 on Build GRCh38
rs537497503 Chr.11: 95821903-95821903 on Build GRCh38
rs548614373 Chr.11: 95822166-95822166 on Build GRCh38
rs540668844 Chr.11: 95822203-95822203 on Build GRCh38
rs767525367 Chr.11: 95821994-95821994 on Build GRCh38
rs189859991 Chr.11: 95821968-95821968 on Build GRCh38
rs979007914 Chr.11: 95821976-95821976 on Build GRCh38
rs11742 Chr.11: 95821924-95821924 on Build GRCh38
rs916891532 Chr.11: 95822007-95822007 on Build GRCh38

View Full Product Details