Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 100859438-100859953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP42
Entrez Gene ID
143872
Gene Name
Rho GTPase activating protein 42
Gene Aliases
GRAF3
UniGene
Hs.741465
Gene Chromosome Location
Chr.11: 100687676-100993941 on Build GRCh38
Transcripts
NM_152432.2 XM_011542615.2 XM_011542616.2 XM_011542617.2 XM_017017237.1 XM_017017238.1

SNP Detail

SNP ID Chromosome Location
rs149389795 Chr.11: 100859855-100859855 on Build GRCh38
rs192934500 Chr.11: 100859889-100859889 on Build GRCh38
rs770256541 Chr.11: 100859568-100859568 on Build GRCh38
rs1017451544 Chr.11: 100859526-100859526 on Build GRCh38
rs367588102 Chr.11: 100859769-100859769 on Build GRCh38
rs992401715 Chr.11: 100859583-100859583 on Build GRCh38
rs572304857 Chr.11: 100859515-100859515 on Build GRCh38
rs1006614086 Chr.11: 100859540-100859540 on Build GRCh38
rs945720738 Chr.11: 100859915-100859915 on Build GRCh38
rs761806771 Chr.11: 100859562-100859562 on Build GRCh38
rs970823051 Chr.11: 100859865-100859865 on Build GRCh38
rs933139156 Chr.11: 100859756-100859756 on Build GRCh38
rs953347162 Chr.11: 100859536-100859536 on Build GRCh38
rs646392 Chr.11: 100859482-100859482 on Build GRCh38
rs555060491 Chr.11: 100859864-100859864 on Build GRCh38
rs1033860181 Chr.11: 100859894-100859894 on Build GRCh38
rs914249491 Chr.11: 100859728-100859728 on Build GRCh38
rs918047089 Chr.11: 100859569-100859569 on Build GRCh38
rs1029610347 Chr.11: 100859829-100859829 on Build GRCh38
rs897824974 Chr.11: 100859836-100859836 on Build GRCh38
rs768948348 Chr.11: 100859556-100859556 on Build GRCh38
rs990191882 Chr.11: 100859727-100859727 on Build GRCh38
rs914177853 Chr.11: 100859906-100859906 on Build GRCh38
rs539855964 Chr.11: 100859565-100859565 on Build GRCh38
rs11224490 Chr.11: 100859922-100859922 on Build GRCh38
rs187482748 Chr.11: 100859555-100859555 on Build GRCh38
rs972829065 Chr.11: 100859890-100859890 on Build GRCh38
rs954215902 Chr.11: 100859481-100859481 on Build GRCh38
rs958497967 Chr.11: 100859898-100859898 on Build GRCh38
rs962405525 Chr.11: 100859563-100859563 on Build GRCh38
rs374009403 Chr.11: 100859510-100859510 on Build GRCh38
rs184565901 Chr.11: 100859899-100859899 on Build GRCh38
rs904282079 Chr.11: 100859740-100859740 on Build GRCh38
rs773574454 Chr.11: 100859581-100859581 on Build GRCh38
rs926843923 Chr.11: 100859616-100859616 on Build GRCh38
rs771455624 Chr.11: 100859666-100859666 on Build GRCh38
rs559688865 Chr.11: 100859713-100859713 on Build GRCh38
rs780221159 Chr.11: 100859508-100859508 on Build GRCh38
rs889004621 Chr.11: 100859776-100859776 on Build GRCh38
rs532111368 Chr.11: 100859747-100859747 on Build GRCh38
rs916907276 Chr.11: 100859717-100859717 on Build GRCh38
rs201466895 Chr.11: 100859920-100859920 on Build GRCh38
rs552019947 Chr.11: 100859754-100859754 on Build GRCh38
rs948412449 Chr.11: 100859720-100859720 on Build GRCh38
rs752213544 Chr.11: 100859666-100859669 on Build GRCh38
rs114387514 Chr.11: 100859811-100859811 on Build GRCh38
rs945745914 Chr.11: 100859733-100859733 on Build GRCh38
rs936956988 Chr.11: 100859715-100859715 on Build GRCh38
rs1006147808 Chr.11: 100859780-100859780 on Build GRCh38
rs763568067 Chr.11: 100859652-100859652 on Build GRCh38
rs980964683 Chr.11: 100859601-100859601 on Build GRCh38

View Full Product Details