Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 124037978-124038487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR10G7
Entrez Gene ID
390265
Gene Name
olfactory receptor family 10 subfamily G member 7
Gene Aliases
OR11-283
UniGene
Hs.553761
Gene Chromosome Location
Chr.11: 124038066-124039001 on Build GRCh38
Transcripts
NM_001004463.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3998289 c.636G>C p.L212L Chr.11: 124038366-124038366 on Build GRCh38
6733641 c.568G>T p.D190Y Chr.11: 124038434-124038434 on Build GRCh38
1293330 c.840T>C p.P280P Chr.11: 124038162-124038162 on Build GRCh38
924293 c.648C>A p.S216S Chr.11: 124038354-124038354 on Build GRCh38
6531332 c.884A>G p.K295R Chr.11: 124038118-124038118 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763765989 Chr.11: 124038301-124038301 on Build GRCh38
rs147011748 Chr.11: 124038420-124038420 on Build GRCh38
rs773966921 Chr.11: 124038428-124038428 on Build GRCh38
rs781769794 Chr.11: 124038444-124038444 on Build GRCh38
rs757153234 Chr.11: 124038266-124038266 on Build GRCh38
rs575557746 Chr.11: 124038071-124038071 on Build GRCh38
rs140026196 Chr.11: 124038427-124038427 on Build GRCh38
rs776340538 Chr.11: 124038354-124038354 on Build GRCh38
rs974847062 Chr.11: 124038024-124038024 on Build GRCh38
rs769355217 Chr.11: 124038401-124038401 on Build GRCh38
rs751136418 Chr.11: 124038077-124038077 on Build GRCh38
rs376708837 Chr.11: 124038300-124038300 on Build GRCh38
rs780871212 Chr.11: 124038100-124038100 on Build GRCh38
rs149133950 Chr.11: 124038365-124038365 on Build GRCh38
rs747619519 Chr.11: 124038457-124038457 on Build GRCh38
rs140206954 Chr.11: 124038272-124038272 on Build GRCh38
rs114086709 Chr.11: 124038172-124038172 on Build GRCh38
rs748216010 Chr.11: 124038275-124038275 on Build GRCh38
rs386758387 Chr.11: 124038420-124038421 on Build GRCh38
rs374379935 Chr.11: 124038062-124038062 on Build GRCh38
rs755284254 Chr.11: 124038029-124038029 on Build GRCh38
rs747090731 Chr.11: 124038175-124038175 on Build GRCh38
rs372508264 Chr.11: 124038388-124038388 on Build GRCh38
rs761554342 Chr.11: 124038357-124038357 on Build GRCh38
rs774866321 Chr.11: 124038217-124038217 on Build GRCh38
rs959090142 Chr.11: 124038361-124038361 on Build GRCh38
rs528549128 Chr.11: 124038207-124038207 on Build GRCh38
rs751456010 Chr.11: 124038367-124038367 on Build GRCh38
rs368033716 Chr.11: 124038044-124038044 on Build GRCh38
rs562656605 Chr.11: 124038421-124038421 on Build GRCh38
rs748354779 Chr.11: 124038369-124038369 on Build GRCh38
rs747442781 Chr.11: 124038442-124038442 on Build GRCh38
rs1004646192 Chr.11: 124038010-124038010 on Build GRCh38
rs865835329 Chr.11: 124038460-124038460 on Build GRCh38
rs755802892 Chr.11: 124038171-124038171 on Build GRCh38
rs748390521 Chr.11: 124038205-124038205 on Build GRCh38
rs755763606 Chr.11: 124038246-124038246 on Build GRCh38
rs199551954 Chr.11: 124038349-124038349 on Build GRCh38
rs895821309 Chr.11: 124038183-124038183 on Build GRCh38
rs199934441 Chr.11: 124038209-124038209 on Build GRCh38
rs754592994 Chr.11: 124038087-124038087 on Build GRCh38
rs372451457 Chr.11: 124038223-124038223 on Build GRCh38
rs184333829 Chr.11: 124038176-124038176 on Build GRCh38
rs533864228 Chr.11: 124038304-124038304 on Build GRCh38
rs750627105 Chr.11: 124038434-124038434 on Build GRCh38
rs1032068353 Chr.11: 124038398-124038398 on Build GRCh38
rs150532549 Chr.11: 124038461-124038461 on Build GRCh38
rs769977171 Chr.11: 124038206-124038206 on Build GRCh38
rs12291067 Chr.11: 124038342-124038342 on Build GRCh38
rs772517733 Chr.11: 124038297-124038297 on Build GRCh38
rs747457854 Chr.11: 124038399-124038399 on Build GRCh38
rs759372225 Chr.11: 124038466-124038466 on Build GRCh38
rs200190231 Chr.11: 124038418-124038418 on Build GRCh38
rs758323652 Chr.11: 124038316-124038316 on Build GRCh38
rs777501224 Chr.11: 124038109-124038109 on Build GRCh38
rs867513894 Chr.11: 124038234-124038234 on Build GRCh38
rs368320970 Chr.11: 124038016-124038016 on Build GRCh38
rs573282156 Chr.11: 124038364-124038364 on Build GRCh38
rs748463459 Chr.11: 124038194-124038208 on Build GRCh38
rs779241710 Chr.11: 124038328-124038328 on Build GRCh38
rs781415894 Chr.11: 124038032-124038036 on Build GRCh38
rs189411659 Chr.11: 124038382-124038382 on Build GRCh38
rs947951769 Chr.11: 124038174-124038174 on Build GRCh38
rs757847611 Chr.11: 124038067-124038067 on Build GRCh38
rs142080661 Chr.11: 124038264-124038264 on Build GRCh38
rs767034136 Chr.11: 124038163-124038163 on Build GRCh38
rs140667003 Chr.11: 124038325-124038325 on Build GRCh38
rs927624601 Chr.11: 124038197-124038197 on Build GRCh38
rs763444511 Chr.11: 124038148-124038148 on Build GRCh38
rs773507107 Chr.11: 124038138-124038138 on Build GRCh38
rs142972044 Chr.11: 124038184-124038185 on Build GRCh38
rs146835072 Chr.11: 124038287-124038287 on Build GRCh38
rs28414940 Chr.11: 124038465-124038465 on Build GRCh38
rs747047250 Chr.11: 124038185-124038185 on Build GRCh38
rs150421981 Chr.11: 124038326-124038326 on Build GRCh38
rs201156002 Chr.11: 124038381-124038381 on Build GRCh38
rs141478608 Chr.11: 124038414-124038414 on Build GRCh38
rs148603378 Chr.11: 124038235-124038235 on Build GRCh38
rs11219419 Chr.11: 124038366-124038366 on Build GRCh38
rs772278444 Chr.11: 124038034-124038034 on Build GRCh38
rs765022860 Chr.11: 124038360-124038360 on Build GRCh38
rs754133814 Chr.11: 124038046-124038046 on Build GRCh38
rs747845511 Chr.11: 124038107-124038107 on Build GRCh38
rs1029651371 Chr.11: 124038103-124038103 on Build GRCh38
rs769432723 Chr.11: 124038459-124038459 on Build GRCh38
rs756156993 Chr.11: 124038371-124038371 on Build GRCh38
rs368332049 Chr.11: 124038164-124038164 on Build GRCh38
rs751044514 Chr.11: 124038152-124038152 on Build GRCh38
rs768779066 Chr.11: 124038242-124038242 on Build GRCh38
rs754504836 Chr.11: 124038153-124038153 on Build GRCh38
rs964616204 Chr.11: 124038078-124038078 on Build GRCh38
rs538806093 Chr.11: 124038324-124038324 on Build GRCh38
rs571498330 Chr.11: 124038322-124038322 on Build GRCh38
rs760167866 Chr.11: 124038346-124038346 on Build GRCh38
rs780277686 Chr.11: 124038439-124038439 on Build GRCh38
rs750311471 Chr.11: 124038311-124038311 on Build GRCh38
rs768066719 Chr.11: 124038352-124038352 on Build GRCh38
rs753983047 Chr.11: 124038375-124038375 on Build GRCh38
rs987224723 Chr.11: 124038051-124038051 on Build GRCh38
rs59358830 Chr.11: 124038392-124038392 on Build GRCh38
rs752771848 Chr.11: 124038320-124038320 on Build GRCh38
rs371534846 Chr.11: 124038049-124038049 on Build GRCh38
rs188316662 Chr.11: 124038270-124038270 on Build GRCh38
rs757360950 Chr.11: 124038423-124038423 on Build GRCh38
rs766911838 Chr.11: 124038149-124038149 on Build GRCh38
rs772586163 Chr.11: 124038341-124038341 on Build GRCh38
rs143478590 Chr.11: 124038132-124038132 on Build GRCh38
rs556293335 Chr.11: 124038055-124038055 on Build GRCh38
rs758704940 Chr.11: 124038436-124038436 on Build GRCh38
rs775046629 Chr.11: 124038299-124038299 on Build GRCh38
rs774256000 Chr.11: 124038419-124038419 on Build GRCh38
rs760978609 Chr.11: 124038035-124038035 on Build GRCh38
rs1006094232 Chr.11: 124038030-124038030 on Build GRCh38
rs770679526 Chr.11: 124038464-124038464 on Build GRCh38
rs747137233 Chr.11: 124038114-124038114 on Build GRCh38
rs149592392 Chr.11: 124038282-124038282 on Build GRCh38
rs750401537 Chr.11: 124038268-124038268 on Build GRCh38
rs746205816 Chr.11: 124038385-124038385 on Build GRCh38
rs151111393 Chr.11: 124038267-124038267 on Build GRCh38
rs180880238 Chr.11: 124038309-124038309 on Build GRCh38
rs776252261 Chr.11: 124038404-124038404 on Build GRCh38
rs747708955 Chr.11: 124038412-124038412 on Build GRCh38
rs763706606 Chr.11: 124038262-124038262 on Build GRCh38
rs564168235 Chr.11: 124038029-124038033 on Build GRCh38
rs997119184 Chr.11: 124038099-124038099 on Build GRCh38
rs776944826 Chr.11: 124038125-124038125 on Build GRCh38
rs566479670 Chr.11: 124038295-124038295 on Build GRCh38
rs746270292 Chr.11: 124038334-124038334 on Build GRCh38
rs371655001 Chr.11: 124038384-124038384 on Build GRCh38
rs750945977 Chr.11: 124038064-124038064 on Build GRCh38
rs199697577 Chr.11: 124038022-124038022 on Build GRCh38
rs200733022 Chr.11: 124038045-124038045 on Build GRCh38
rs775683985 Chr.11: 124038112-124038112 on Build GRCh38
rs752203237 Chr.11: 124038240-124038240 on Build GRCh38
rs776659432 Chr.11: 124038199-124038199 on Build GRCh38
rs768954231 Chr.11: 124038120-124038120 on Build GRCh38
rs1044988464 Chr.11: 124038179-124038179 on Build GRCh38
rs765648153 Chr.11: 124038135-124038135 on Build GRCh38
rs765110770 Chr.11: 124038310-124038310 on Build GRCh38
rs770256479 Chr.11: 124038257-124038257 on Build GRCh38
rs774780147 Chr.11: 124038296-124038296 on Build GRCh38
rs749258039 Chr.11: 124038110-124038110 on Build GRCh38
rs779903828 Chr.11: 124038319-124038319 on Build GRCh38
rs553750424 Chr.11: 124038104-124038104 on Build GRCh38

View Full Product Details