Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 126262419-126262928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM118B
Entrez Gene ID
79607
Gene Name
family with sequence similarity 118 member B
Gene Aliases
-
UniGene
Hs.276315
Gene Chromosome Location
Chr.11: 126211414-126264540 on Build GRCh38
Transcripts
NM_024556.3 XM_005271665.3 XM_011542977.2 XM_011542978.2 XM_017018285.1 XM_017018286.1 NM_001177842.1 NM_003139.3 XM_017018179.1

Gene Symbol
SRPRA
Entrez Gene ID
6734
Gene Name
SRP receptor alpha subunit
Gene Aliases
DP, SRPR, Sralpha
UniGene
Hs.368376
Gene Chromosome Location
Chr.11: 126255151-126268982 on Build GRCh38
Transcripts
NM_024556.3 XM_005271665.3 XM_011542977.2 XM_011542978.2 XM_017018285.1 XM_017018286.1 NM_001177842.1 NM_003139.3 XM_017018179.1

SNP Detail

SNP ID Chromosome Location
rs989903090 Chr.11: 126262592-126262592 on Build GRCh38
rs895159493 Chr.11: 126262481-126262481 on Build GRCh38
rs867068066 Chr.11: 126262469-126262469 on Build GRCh38
rs1042990397 Chr.11: 126262707-126262707 on Build GRCh38
rs868379033 Chr.11: 126262551-126262551 on Build GRCh38
rs778457493 Chr.11: 126262657-126262657 on Build GRCh38
rs933310634 Chr.11: 126262675-126262675 on Build GRCh38
rs961302539 Chr.11: 126262598-126262598 on Build GRCh38
rs1003056473 Chr.11: 126262550-126262550 on Build GRCh38
rs1033686389 Chr.11: 126262465-126262465 on Build GRCh38
rs569424554 Chr.11: 126262516-126262516 on Build GRCh38
rs910943588 Chr.11: 126262635-126262635 on Build GRCh38
rs1006704001 Chr.11: 126262887-126262887 on Build GRCh38
rs11603806 Chr.11: 126262496-126262496 on Build GRCh38
rs903980998 Chr.11: 126262455-126262455 on Build GRCh38
rs114906929 Chr.11: 126262846-126262846 on Build GRCh38
rs538915712 Chr.11: 126262847-126262847 on Build GRCh38
rs867903630 Chr.11: 126262548-126262548 on Build GRCh38
rs483668 Chr.11: 126262690-126262690 on Build GRCh38
rs1050954097 Chr.11: 126262682-126262682 on Build GRCh38
rs958489946 Chr.11: 126262835-126262835 on Build GRCh38
rs373743291 Chr.11: 126262778-126262778 on Build GRCh38
rs895213406 Chr.11: 126262758-126262758 on Build GRCh38
rs772036732 Chr.11: 126262699-126262699 on Build GRCh38
rs958541934 Chr.11: 126262576-126262576 on Build GRCh38
rs530565852 Chr.11: 126262604-126262604 on Build GRCh38
rs965058528 Chr.11: 126262672-126262672 on Build GRCh38
rs753629172 Chr.11: 126262508-126262508 on Build GRCh38
rs1013513776 Chr.11: 126262484-126262484 on Build GRCh38
rs558520586 Chr.11: 126262851-126262851 on Build GRCh38
rs952534859 Chr.11: 126262880-126262880 on Build GRCh38
rs993760364 Chr.11: 126262444-126262444 on Build GRCh38
rs976425433 Chr.11: 126262686-126262686 on Build GRCh38
rs925457195 Chr.11: 126262733-126262733 on Build GRCh38
rs973008986 Chr.11: 126262600-126262600 on Build GRCh38
rs770522646 Chr.11: 126262625-126262625 on Build GRCh38
rs1000902552 Chr.11: 126262457-126262457 on Build GRCh38
rs1042547467 Chr.11: 126262449-126262449 on Build GRCh38
rs770106203 Chr.11: 126262869-126262870 on Build GRCh38
rs936808900 Chr.11: 126262750-126262750 on Build GRCh38
rs985213640 Chr.11: 126262634-126262634 on Build GRCh38
rs201747634 Chr.11: 126262824-126262824 on Build GRCh38
rs967734836 Chr.11: 126262613-126262613 on Build GRCh38
rs775218911 Chr.11: 126262814-126262814 on Build GRCh38
rs575377144 Chr.11: 126262879-126262879 on Build GRCh38
rs116795411 Chr.11: 126262806-126262806 on Build GRCh38
rs923287706 Chr.11: 126262638-126262638 on Build GRCh38
rs566956106 Chr.11: 126262795-126262795 on Build GRCh38
rs1025331482 Chr.11: 126262557-126262557 on Build GRCh38
rs1055150834 Chr.11: 126262751-126262751 on Build GRCh38
rs1034064314 Chr.11: 126262822-126262822 on Build GRCh38
rs1025764958 Chr.11: 126262461-126262463 on Build GRCh38
rs1026868845 Chr.11: 126262853-126262853 on Build GRCh38
rs971070018 Chr.11: 126262486-126262486 on Build GRCh38

View Full Product Details