Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1721411-1721848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK11A
Entrez Gene ID
728642
Gene Name
cyclin dependent kinase 11A
Gene Aliases
CDC2L2, CDC2L3, CDK11-p110, CDK11-p46, CDK11-p58, PITSLRE, p58GTA
UniGene
Hs.651228
Gene Chromosome Location
Chr.1: 1702383-1724565 on Build GRCh38
Transcripts
NM_001313896.1 NM_001313982.1 NM_024011.3 NM_033529.3 NR_132739.1 NR_132740.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7143745 c.161G>C p.R54T Chr.1: 1721662-1721662 on Build GRCh38
5844357 c.140C>T p.S47F Chr.1: 1721683-1721683 on Build GRCh38
4475201 c.196C>T p.P66S Chr.1: 1721627-1721627 on Build GRCh38
48277 c.145G>C p.E49Q Chr.1: 1721678-1721678 on Build GRCh38
2088846 c.154G>T p.E52* Chr.1: 1721669-1721669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761308164 Chr.1: 1721592-1721598 on Build GRCh38
rs188565066 Chr.1: 1721671-1721671 on Build GRCh38
rs776650584 Chr.1: 1721702-1721702 on Build GRCh38
rs758160511 Chr.1: 1721569-1721569 on Build GRCh38
rs368601641 Chr.1: 1721756-1721756 on Build GRCh38
rs780694487 Chr.1: 1721661-1721661 on Build GRCh38
rs564761761 Chr.1: 1721751-1721751 on Build GRCh38
rs576656860 Chr.1: 1721765-1721765 on Build GRCh38
rs748452843 Chr.1: 1721689-1721689 on Build GRCh38
rs779436006 Chr.1: 1721682-1721682 on Build GRCh38
rs796263331 Chr.1: 1721587-1721589 on Build GRCh38
rs774681561 Chr.1: 1721558-1721558 on Build GRCh38
rs374169207 Chr.1: 1721625-1721625 on Build GRCh38
rs770617385 Chr.1: 1721596-1721596 on Build GRCh38
rs535469099 Chr.1: 1721524-1721524 on Build GRCh38
rs570621826 Chr.1: 1721450-1721450 on Build GRCh38
rs765075112 Chr.1: 1721566-1721566 on Build GRCh38
rs539981326 Chr.1: 1721458-1721458 on Build GRCh38
rs762879797 Chr.1: 1721710-1721710 on Build GRCh38
rs561679412 Chr.1: 1721735-1721735 on Build GRCh38
rs769222111 Chr.1: 1721676-1721676 on Build GRCh38
rs555696071 Chr.1: 1721541-1721541 on Build GRCh38
rs774820411 Chr.1: 1721606-1721606 on Build GRCh38
rs1041857027 Chr.1: 1721728-1721728 on Build GRCh38
rs377733353 Chr.1: 1721559-1721559 on Build GRCh38
rs1137006 Chr.1: 1721607-1721607 on Build GRCh38
rs754267453 Chr.1: 1721682-1721693 on Build GRCh38
rs540105321 Chr.1: 1721690-1721690 on Build GRCh38
rs763961101 Chr.1: 1721723-1721723 on Build GRCh38
rs533141562 Chr.1: 1721807-1721807 on Build GRCh38
rs770387234 Chr.1: 1721588-1721588 on Build GRCh38
rs750404819 Chr.1: 1721562-1721562 on Build GRCh38
rs114172122 Chr.1: 1721552-1721552 on Build GRCh38
rs376262877 Chr.1: 1721577-1721577 on Build GRCh38
rs181359165 Chr.1: 1721672-1721672 on Build GRCh38
rs1026133068 Chr.1: 1721819-1721819 on Build GRCh38
rs958440268 Chr.1: 1721473-1721473 on Build GRCh38
rs866177599 Chr.1: 1721770-1721770 on Build GRCh38
rs370759338 Chr.1: 1721608-1721608 on Build GRCh38
rs185839325 Chr.1: 1721806-1721806 on Build GRCh38
rs200953503 Chr.1: 1721701-1721701 on Build GRCh38
rs755604551 Chr.1: 1721755-1721755 on Build GRCh38
rs942749671 Chr.1: 1721593-1721593 on Build GRCh38
rs201926274 Chr.1: 1721622-1721622 on Build GRCh38
rs36039898 Chr.1: 1721565-1721565 on Build GRCh38
rs760857763 Chr.1: 1721691-1721691 on Build GRCh38
rs763706850 Chr.1: 1721547-1721554 on Build GRCh38
rs763905836 Chr.1: 1721653-1721653 on Build GRCh38
rs746418138 Chr.1: 1721549-1721549 on Build GRCh38
rs773403139 Chr.1: 1721623-1721623 on Build GRCh38
rs372225055 Chr.1: 1721673-1721673 on Build GRCh38
rs767669345 Chr.1: 1721560-1721560 on Build GRCh38
rs16825265 Chr.1: 1721589-1721589 on Build GRCh38
rs778777791 Chr.1: 1721525-1721525 on Build GRCh38
rs751016482 Chr.1: 1721580-1721581 on Build GRCh38
rs1059832 Chr.1: 1721654-1721654 on Build GRCh38
rs755345301 Chr.1: 1721750-1721751 on Build GRCh38
rs373127485 Chr.1: 1721575-1721575 on Build GRCh38
rs754420872 Chr.1: 1721465-1721466 on Build GRCh38
rs775954898 Chr.1: 1721548-1721548 on Build GRCh38
rs750018859 Chr.1: 1721739-1721739 on Build GRCh38
rs556693732 Chr.1: 1721461-1721461 on Build GRCh38
rs903387267 Chr.1: 1721736-1721736 on Build GRCh38
rs769117676 Chr.1: 1721591-1721591 on Build GRCh38
rs749715548 Chr.1: 1721821-1721821 on Build GRCh38
rs751412452 Chr.1: 1721726-1721726 on Build GRCh38
rs753342989 Chr.1: 1721752-1721752 on Build GRCh38
rs763393094 Chr.1: 1721553-1721553 on Build GRCh38
rs565553918 Chr.1: 1721631-1721631 on Build GRCh38
rs1013074319 Chr.1: 1721484-1721484 on Build GRCh38
rs761505045 Chr.1: 1721730-1721730 on Build GRCh38
rs370345461 Chr.1: 1721580-1721580 on Build GRCh38
rs184721773 Chr.1: 1721587-1721587 on Build GRCh38
rs796339356 Chr.1: 1721750-1721751 on Build GRCh38
rs570095997 Chr.1: 1721468-1721468 on Build GRCh38
rs780758425 Chr.1: 1721583-1721583 on Build GRCh38
rs368753784 Chr.1: 1721626-1721626 on Build GRCh38
rs755740687 Chr.1: 1721746-1721746 on Build GRCh38
rs766922547 Chr.1: 1721672-1721674 on Build GRCh38
rs752555902 Chr.1: 1721567-1721567 on Build GRCh38

View Full Product Details